Gallery and Gallery Plus discrete analyzers reduce hands-on time and simplify laboratory testing. Designed to increase efficiency for photometric (colorimetric and enzymatic) and electrochemical (pH and conductivity) analysis, these systems provide fast, reproducible results in a compact, benchtop design. The discrete cell technology and automated analysis allow you to measure multiple analytes simultaneously with low detection levels. These analyzers can accommodate up to 108 samples and up to 42 reagents, giving them the ability to perform up to 350 tests/hour using precise, predefined workflows, minimizing manual errors, and providing technicians with up to 3 hours of walkaway time.
Gallery and Gallery Plus discrete analyzers are fully automated, sample-oriented, random-access analyzers built for wet chemical analysis. Many different tests can be run from a single sample simultaneously and the systems offer continuous access to samples, reagents and cuvettes without interrupting the analytical workflow, which further increases throughput and saves more time. Measurements cover a wide range of concentrations with excellent reproducibility and detection limits in the parts-per-billion (ppb) range. Our state-of-the-art, high-throughput discrete analyzers feature:
 Unique, disposable cuvettes
Unique, disposable cuvettes
Gallery systems use low-cost, disposable DECACELL cuvettes. A single set of cuvettes includes 10 reaction cells, mounted together for true discrete analysis. They eliminate sample carryover and the need for washing, increasing throughput, eliminating contamination and ensuring reliable results. They also require smaller (2–240 µL) sample and reagent volumes, reducing waste generation, disposal costs and, ultimately, cost per analysis up to 20x.
True, direct-read analysis
True, direct-read analysis
These high-throughput, direct-read discrete analyzers can process up to 350 tests/hour and feature an optional ECM unit that enables parallel pH and conductivity measurements. They include 12 filter positions that cover a spectral range from 340–880 nm and up to 20 different chemical parameters per sample. Measurements cover a wide range of concentrations with sensitivity to the ppb level and excellent reproducibility.
Ready-to-use methods and reagents
Ready-to-use methods and reagents
Each Gallery discrete analyzer comes with pre-defined, easy-to-use methods, but you can also create and run application-specific methods of your own. More than 50 ready-to-use enzymatic and colorimetric reagents eliminate time-consuming reagent preparation and enable operators to attend to other tasks.
Automated instrument processes
Automated instrument processes
Simply load the samples, insert the reagent vials and leave the analyzer do its work for up to 3 hours of walkaway time. Instrument calibration is easily and automatically performed from a single stock standard. Additionally, the analyzer’s smart dilution capabilities mean that over-range samples are automatically diluted by the system to increase both throughput and accuracy. Gallery systems also include automatic startup and shutdown protocols.
Workflow-based software and data management
Workflow-based software and data management
Our intuitive user interface is suitable for all experience levels. The software is open and flexible.  It can be programmed to adopt third party reagent and segmented flow photometric test methods.  The software also includes LIMS connectivity, user administration, QC charts that flag out-of-specification control results and a real-time QC program with multiple user-defined Westgard rules. A built-in barcode reader provides traceability of all samples and reagents in the system.  Methods can easily be shared between laboratories across the globe by simply scanning a barcode.​ 
See how Gallery discrete analyzers automate photometric and electrochemical analysis to provide fast, reproducible results in a compact, benchtop design.
Thermo Scientific Gallery Discrete Analyzer
Learn more about the Gallery discrete analyzer and see its features in action.
Thermo Scientific Gallery Plus Discrete Analyzer
See the Gallery Plus discrete analyzer at work and learn more about its features.
Both our Gallery and Gallery Plus discrete analyzers are available with an electrochemical measurement (ECM) unit, which enables rapid, parallel electrochemical analysis in addition to photometric analysis of multiple analytes for a variety of applications.
Gallery Discrete Analyzer
Gallery Plus Discrete Analyzer
Sample capacity
90 samples
108 samples
Reagent capacity
30 positions
42 positions
Reagents per test
Add up to 4 reagents
Tests/hour
200 tests/hr
350 tests/hr
System dimensions
75 cm W × 70 cm D × 62 cm H (closed)
75 cm W × 70 cm D × 130 cm H (open)
94 cm W × 70 cm D × 62 cm H (closed)
94 cm W × 70 cm D × 130 cm H (open)
Full specifications
Full specifications
Which discrete analyzer is best for my lab?
Download this helpful discrete analyzer selection guide to learn about the top 10 features to consider when choosing the right discrete analyzer for your wet chemical analysis, including hardware, software, throughput, and cost per analysis.
Download guide ›
Click on the corresponding link below to request a quote for the purchase of your desired discrete analyzer system.
Simplified wine analysis
Thermo Scientific Gallery discrete analyzers make wine analysis simple, accurate and reliable. Our ready-to-use system reagents are specifically developed for rapid and cost-effective analysis of juice and wine.
High-throughput environmental analysis
Learn how our discrete analyzers help you comply with the U.S. EPA Clean Water Act (CWA) by automating photometric and electrochemical analysis to provide fast, reproducible results.
Industrial water analysis
To eliminate corrosion and scaling that can damage valuable industrial components and to ensure regulatory compliance, reliable water analysis must be performed continuously.
Increasing efficiency at a Napa Valley winery
Learn how Starmont winery used automated testing to reduce testing time and enable employees to complete other vital tasks during the busy harvest season.
View now
Solutions for industrial and wastewater processing
Take a look at this infographic to see our solutions to the top three analytical challenges in industrial water and wastewater processing.
View the solution
On-demand webinars
Looking for more automated wet chemical analysis resources?
Visit our filterable and searchable resources library for product information, application notes, case studies, videos and more about our discrete analyzers.
Access more resources ›
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific™ products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
Style Sheet for Komodo Tabs
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD Wide-format style fixes
CMD SchemaApp code