Saving lives and preventing disaster
Radiation, chemical and narcotics incidents expose all elements of society to fatal risks. Our vast range of identification and detection devices save lives and prevent disaster - any mission, any scenario. Thermo Scientific instruments help emergency and first responder personnel keep themselves and our world safer.
Thermo Scientific First Defender instrument in the field
Handheld analyzers
From routine security monitoring and surveillance to emergency response situations, integrated analytical instruments help safety and security professionals detect and mitigate hazardous materials, narcotics, and chemical weapons and agents to keep the public and themselves safe.
TruNarc Handheld Narcotics Analyzer
The Thermo Scientific TruNarc Handheld Narcotics Analyzer enables officers, customs, border control, and other personnel to scan more than 500 suspected controlled substances in a single, definitive test.
Thermo Scientific TruNarc Handheld Narcotics Analyzer
Gemini Analyzer with LowDoseID
The Thermo Scientific Gemini Analyzer with LowDoseID integrates both FTIR and Raman spectroscopies in a rugged, lightweight, handheld solution.
Image of Thermo Scientific Gemini Analyzer with LowDoseID
FirstDefender RM Chemical Identification System
The Thermo Scientific FirstDefender RM Chemical Identification System enables hazmat, law enforcement, military, and other first responders to obtain accurate identification of chemicals, explosives and hazardous materials in seconds, even through sealed translucent containers.
Thermo Scientific FirstDefender RM Chemical Identification System
FirstDefender RMX Handheld Chemical Identification Analyzer
The Thermo Scientific FirstDefender RMX Handheld Chemical Identification Analyzer helps enable military and civilian first responders to quickly identify unknown solid and liquid chemicals down range including explosives, toxic industrial chemicals, chemical weapons, narcotics, precursors, white powders, and more.
Gloved hand holding Thermo Scientific FirstDefender RMX Handheld Chemical Identification Analyzer
TruDefender FTX Handheld FTIR Analyzer
Thermo Scientific TruDefender FTX Handheld FTIR Analyzer for chemical identification harnesses the power of FTIR spectroscopy to provide fast, actionable data to mitigate potential threats and protect hazmat teams and the community.
Thermo Scientific™ TruDefender™ FTX/FTXi Handheld FTIR analyzer for Chemical Identification
Defender Omega Handheld Raman Analyzer
Perform safe, fast identification of explosives, chemical warfare agents, narcotics, and other chemicals with the Thermo Scientific Defender Omega Handheld Raman Analyzer.
Image of Thermo Scientific Defender Omega Handheld Raman Analyzer
1064Defender Raman Analyzer
The Thermo Scientific 1064Defender Raman Analyzer performs touchless, point and shoot scanning for narcotics identification and chemical identification.
Image of Thermo Scientific 1064Defender Raman Analyzer
Radiation detectors with natural background rejection (NBR)
Radiation detectors utilize technology that separates atoms into positive ions and electrons, which are then attracted to electrodes, causing a current to flow, and an electric signal to be generated. These electric signals can be measured as the amount of radiation. However, in real-world, field settings, naturally occurring radiation exists in varying levels from the earth and atmosphere, which can obscure and confuse targeted detection efforts. 
Natural Background Rejection (NBR) is a technology used to eliminate fluctuating natural background levels while measuring radiation. This proprietary and patented technology is used to quickly differentiate between natural and artificial radiation by stripping away any natural background radiation that is registering, delivering you a more accurate result of artificial radiation levels. Operators using instruments without NBR often set their alarm thresholds higher to eliminate the nuisances of false alarms or ignore alarms due to their frequency potentially missing out on hidden or shielded sources. Using an instrument with NBR allows you to keep your alarm threshold extremely low without the need to worry about false alarms from non-threatening sources so when the alarm goes off, you know it is time to take action and act.
RadEye G Series Personal Dose Rate Meters
The Thermo Scientific RadEye G Series Personal Dose Rate Meters provide first responders with quick, reliable gamma and dose rate measurements.
Thermo Scientific RadEye G Series Personal Dose Rate Meters
RadHalo RDP and FM Spectroscopic Area Monitors
The Thermo Scientific RadHalo RDP and FM Spectroscopic Area Monitors detect and identify radiation on location or from miles away.
Thermo Scientific RadHalo Monitors visible
PackEye Radiation Detection Backpack
The Thermo Scientific PackEye Radiation Detection Backpack provides your survey teams with a tool for effectively addressing the problems of orphaned sources, radiological contamination, and maliciously introduced sources.
Thermo Scientific™ PackEye Radiation Detection Backpack
RIIDEye Radiation Isotope Identifier
The Thermo Scientific RIIDEye X and RIIDEye M Handheld Radioisotope Identifiers, with patented Quadratic Compression Conversion (QCC) technology, are well suited to support users from homeland security operations to contamination monitoring and remediation.
Thermo Scientific RIIDEye radiation isotope identifier
RadEye AB100 Alpha-Beta Contamination Monitor
Safeguard your employees and help ensure a secure work environment with the Thermo Scientific RadEye AB100 Alpha-Beta Contamination Monitor. The RadEye AB100 monitor detects alpha/beta surface contamination in a range of environments using versatile operation modes so employees never risk exceeding exposure limits.
Thermo Scientific RadEye AB100 Alpha-Beta Contamination Monitor
Dosimetry Instruments
lab worker near radiation-emitting equipment
Dosimetry instruments measure and monitor the radiation dose absorbed in the body resulting from exposure to ionizing radiation. Some jobs regularly expose workers to limited amounts of radiation, and workers are required to wear a dosimeter in order to monitor and track cumulative radiation exposure against a prescribed annual radiation dose limit.  Safety and security personnel, especially those tasked with threat detection, can be protected by monitoring any exposure using dosimetry technology.  Similarly, working environments where radiation exposure could occur can be monitored over a large area using environmental dosimetry systems.
EPD TruDose Electronic Personal Dosimeter (EPD)
The Thermo Scientific EPD TruDose Electronic Personal Dosimeter (EPD) monitors Gamma and Beta radiation in any environment where workers need to monitor their exposure to radiation, with improved accuracy and simplified operation over previous generations of EPDs.
Thermo Scientific™ EPD TruDose Electronic Personal Dosimeter visible
TLD readers, cards and materials
Thermo Scientific TLD readers, cards and materials provide cost-effective measurements of the radiation dose absorbed by individual TLD elements for medical, nuclear, research, and health physics applications.
Thermo Scientific dosimetry readers
Dosimetry Services
Thermo Scientific Dosimetry Services offers you accurate measurement of radiation dose exposure in the workplace, ensuring worker safety. Badges are sent to the employer for employees to wear during a certain period. When the time periodtime is up, badges are returned to the Thermo Fisher Scientific facility to be analyzed.  Online reports are then generated and available to the employer.
Thermo Scientific Dosimetry Services brochure
For the presumptive identification or detection of narcotics, hazardous materials, chemical agents, explosives, and radiological threats only.
1x1 image pixel for data collection