SF-900 III
  • Low hydrolysate concentration — reduces cell culture variability
  • Animal origin-free — facilitates the regulatory process
  • Optimized to support Sf9 and Sf21 cells — achieves Sf9 densities of 10-14 X106 cells/ml
Experience the difference with Sf-900 III SFM
Gibco Sf-900 III SFM is specifically designed to offer high performance and yield using Sf9 and Sf21 cells in both baculovirus and stable expression systems in a serum-free environment. This medium contains no components of animal origin, thus minimizing the opportunity for introduction of adventitious agents and facilitating the regulatory process. It will support Sf9 cell growth to densities of 10 to 14 × 106 cells/ml (Figure 1) and offers:
  • A protein-free formulation with a low hydrolysate concentration, for improved lot-to-lot consistency and reliability
  • Freedom to add the supplements you need for flexibility and control over your process
  • Convenient packaging in Gibco bottles (500mL and 1000mL) and Universal Bags (10L and 20L)
  • Compatibility with available pre-adapted cells for faster turnaround
Sf9 cell growth in Sf-900 III SFM compared to other insect cell culture media
Figure 1. Sf9 cell growth in Sf-900 III SFM compared to other insect cell culture media. Sf9 cells were sequentially adapted to all three tested animal origin–free (AOF) media (Sf-900 III SFM, EX-CELL® TiterHigh™ medium, ESF AF medium) and Sf-900 II SFM. Once the cells were adapted to each test medium (a total of 8 passages post-recovery), growth curves were determined by seeding 30 mL of medium in 125 mL shake flasks at a seeding density of 4 × 105 viable cells/mL. Each test condition represents the average of two lots of the respective medium with two replicates each (a total of four replicates per test condition).
Ordering information
Catalog # Name Size Price (USD) Qty
12658035 Sf-900™ III SFM, 10 L Each
1,182.65
Online Exclusive
1,246.00
Save 63.35 (5%)
12659017 Sf9 cells in Sf-900™ III SFM Each
813.65
Online Exclusive
870.00
Save 56.35 (6%)
12658001 Sf-900™ III SFM, 20 L Each
2,004.65
Online Exclusive
2,465.00
Save 460.35 (19%)
12658027 Sf-900™ III SFM, 1000 mL Each
139.65
Online Exclusive
147.00
Save 7.35 (5%)
12682019 Sf21 cells in Sf-900™ III SFM Each
856.00
12658019 Sf-900™ III SFM, 500 mL Each
80.65
Online Exclusive
84.50
Save 3.85 (5%)
For Research Use Only. Not for use in diagnostic procedures.