ExpiSf CD Medium
Experience the highest consistency and protein expression with a chemically defined formulation.
ExpiSf CD Medium is a GMP-manufactured, chemically defined, yeastolate-free, serum-free, protein-free, animal origin-free medium specifically designed for high-density growth, transfection, and baculovirus infection of suspension-adapted Sf9 (Spodoptera frugiperda) insect cells.
Delivers high reproducibility and lot-to-lot reliability.
Facilitates high-level expression of baculovirus-infected cultures, achieving protein yields three times higher than traditional insect expression platforms when used with other components of the ExpiSf Expression System.
Sf-900 III SFM
An animal origin-free formulation to support high density growth and protein expression
Optimized to support Sf9 and Sf21 cells—achieves Sf9 densities of 10-14 X 106 cells/ml
Sf9 and Sf21 cells pre-adapted to Sf-900 SFM available providing faster turnaround.
Contains lower hydrolysate concentration compared to other commercially available serum free media
Animal origin-free — facilitates regulatory processes.
Sf-900 II SFM
A protein-free medium optimized to support and maximize Sf9 and Sf21 cell growth in suspension applications
Optimized for recombinant gene expression in both baculovirus and stable insect expression systems
Express Five SFM
A protein-free medium optimized to support maximum cell growth
Optimized for recombinant gene expression by the baculovirus expression by the baculovirus expression vector system (BEVS) in the BTI-TN-5B1-4 (High Five)Trichoplusia ni cell line.
Table 1. Gibco insect cell culture media.
ProductOptimized forProtein freeSerum freeAnimal origin freePreadapted cell lines available
ExpiSf CD Medium
Suspension ExpiSf9, Sf9, Sf21 cells 
(Spodoptera frugiperda)
XXXExpiSF9 Cells (A35243)
Sf-900 III SFM
Suspension Sf9, Sf21 cells 
(Spodoptera frugiperda)
XX Sf9 Cells
(12659-017)
Sf21 Cells
(12682-019)
Sf-900 II SFM*Suspension Sf9, Sf21 cells
(Spodoptera frugiperda), TN368 cells (Trichoplusia ni)
XX Sf9 Cells
(11496-015)
Sf21 Cells
(11497-013)
Express Five  SFM*
Suspension BT1-TN-5B1-4 insect cellsXX High Five™ Cells
(B855-02)
All four media used for the growth and maintenance of cells in the baculovirus expression vector systems (BEVS) for adherent or suspension culture. Also for large-scale production of recombinant proteins expressed by BEVS.
*Drug Master File Available.
Note: Cell lines from different sources, and different clones of the same cell line, may have highly specific nutritional requirements and may therefore prefer one medium over another. More than one medium formulation (if available) should be evaluated to determine the best option.
Catalog # Name Size Price (USD) Qty
12658035 Sf-900™ III SFM, 10 L Each
1,182.65
Online Exclusive
1,246.00
Save 63.35 (5%)
12659017 Sf9 cells in Sf-900™ III SFM Each
813.65
Online Exclusive
870.00
Save 56.35 (6%)
A3767804 ExpiSf™ CD Medium, 10 L Each
1,268.65
Online Exclusive
1,308.00
Save 39.35 (3%)
A3767803 ExpiSf™ CD Medium, 6 x 1000 mL Each
807.65
Online Exclusive
832.00
Save 24.35 (3%)
A3767802 ExpiSf™ CD Medium, 1000 mL Each
151.65
Online Exclusive
159.00
Save 7.35 (5%)
A3767801 ExpiSf™ CD Medium, 500 mL Each
85.65
Online Exclusive
87.75
Save 2.10 (2%)
12658027 Sf-900™ III SFM, 1000 mL Each
139.65
Online Exclusive
147.00
Save 7.35 (5%)
12682019 Sf21 cells in Sf-900™ III SFM Each
856.00
10486025 Express Five™ SFM Each
143.00
10902088 Sf-900™ II SFM, 1000 mL Each
126.65
Online Exclusive
136.00
Save 9.35 (7%)
10902187 Sf-900™ II SFM, 20 L Each
1,918.65
Online Exclusive
1,978.00
Save 59.35 (3%)
10902179 Sf-900™ II SFM, 10 L Each
1,088.65
Online Exclusive
1,122.00
Save 33.35 (3%)
10902161 Sf-900™ II SFM, 5 L Each
655.65
Online Exclusive
676.00
Save 20.35 (3%)
10902096 Sf-900™ II SFM, 500 mL Each
72.65
Online Exclusive
76.25
Save 3.60 (5%)
12658019 Sf-900™ III SFM, 500 mL Each
80.65
Online Exclusive
84.50
Save 3.85 (5%)
For Research Use Only. Not for use in diagnostic procedures.