Microvascular Endothelial Cell Culture Systems
  • > 500,000 cells/vial, cryopreserved at the end of 3' culture
  • > 70% viability
  • > 16 population doublings HMVECad
  • > 16 population doublings HMVECnd
  • Optimized for superior performance over the leading competitors
For a comprehensive list of cells, technical resources and related technologies visit the Cell Resource Center.
Request More Information
Phase contrast images of Human Microvascular Endothelial Cells (HMVECad) growing in culture
HMVECad grown in Medium 131 with attachment factor ( M-131-500) with the addition of Microvascular Growth Supplement ( MVGS). First culture after thaw.
HMVECad
Day 3 - HMVECad
 HMVECad
Day 5 - HMVECad
 HMVECad
Day 7 - HMVECad
Ordering Information
Catalog # Name Size Price (USD) Qty
S00525 Microvascular Growth Supplement (MVGS) Each
154.00
R01510 Gentamicin/Amphotericin Solution Each
117.00
R001100 Trypsin/EDTA Solution (TE) Each
39.65
Online Exclusive
41.30
Save 1.65 (4%)
R002100 Trypsin Neutralizer Solution Each
38.65
Online Exclusive
41.28
Save 2.63 (6%)
For Research Use Only. Not for use in diagnostic procedures.