Smooth Muscle Cell Culture Systems
Primary Human Aortic Smooth Muscle Cells (HASMC)
Primary Human Coronary Artery Smooth Muscle Cells (HCASMC)
Primary Human Pulmonary Artery Smooth Muscle Cells (HPASMC)
  • > 500,000 cells/vial, cryopreserved at the end of 3' culture
  • > 70% viability
  • > 16 population doublings HASMC
  • > 16 population doublings HCASMC
  • > 16 population doublings HPASMC
  • Optimized for superior performance over the leading competitors
Request More Information
Phase contrast image of Primary Human Aortic Smooth Muscle Cells (HASMC) in culture
HASMC grown in Medium 231 ( M-231-500) and Smooth Muscle Growth Supplement ( SMGS). First culture after thaw.
Phase contrast image of Primary Human Aortic Smooth Muscle Cells (HASMC) in culture
Day 5 - HASMC
Phase contrast image of Primary Human Aortic Smooth Muscle Cells (HASMC) in culture
Day 7 - HAMSC
Phase contrast image of Primary Human Aortic Smooth Muscle Cells (HASMC) in culture
Day 9 - HAMSC
Ordering Information
Catalog # Name Size Price (USD) Qty
R01510 Gentamicin/Amphotericin Solution Each
117.00
R001100 Trypsin/EDTA Solution (TE) Each
41.30
R002100 Trypsin Neutralizer Solution Each
38.65
Online Exclusive
41.28
Save 2.63 (6%)
M231500 Human Vascular Smooth Muscle Cell Basal Medium (formerly Medium 231) Each
102.65
Online Exclusive
109.00
Save 6.35 (6%)
S0085 Smooth Muscle Differentiation Supplement (SMDS) Each
79.25
S00725 Smooth Muscle Growth Supplement (SMGS) Each
147.65
Online Exclusive
155.00
Save 7.35 (5%)
For Research Use Only. Not for use in diagnostic procedures.