Aquanex Ultrapure Water Purification System
Count on Thermo Scientific Aquanex Ultrapure Water Purification System for convenience, flexibility and reliable water purity. Spend more time on your research and less time maintaining your equipment.
Ease of use and intelligent operation
Designed with ease of use in mind
Quick-connect fixtures make installation easy. Dispense from the unit or remove the pipette-style dispenser to fill larger containers. Change components with ease by removing the magnetic front panel for quick access – no screws or complicated fixtures; unit automatically recognizes new cartridges.
Aquanex ease of use
Reliable water quality at your fingertips
Secured log-in feature and seamless event logging, monitoring and reporting. High Flow smart dispensing enhances user experience with industry leading accuracy. Automatic Leak Detection and system shut off for safer lab environment.
 Sustainability
Select Water Saver, Pack Saver or Dynamic Mode for your lab operation. Water Saver Mode can help you save water when using high-quality feed water.
Aquanex sustainability
Aquanex features
Aquanex touch smart
Aquanex Touch Smart Dispenser
  • Intelligent, easy-to-handle pipette-style dispenser
  • Remove from the stand to fill larger containers
  • Intuitive touchscreen display allows for easy operation
Aquanex user interface
Aquanex System User Interface
  • The intuitive, full color touchscreen display is easy to read and simple to operate
  • The large display allows you to clearly view water quality, type and tank fill from a distance
Aquanex maintenance
Easy maintenance and self-service design
  • UV lamp, RO assembly, and ultrafilter consumables are easily accessed by removing the magnetic front panel
  • No screws or complicated tools are required to change consumables
Applications
The perfect fit for your applications
Laboratory scientists count on the consistent performance of Type I water. The intelligent, reliable Aquanex system provides reliable performance to meet the purification needs of a range of industries.
aquanex applications
Maintenance and Consumables
Aquanex consumables
System consumables
Name
Catalog number
Estimated lifetime*
When to replace
Thermo Scientific Reverse Osmosis Cartridge
50157608
Up to 12 months
"Change RO Assembly" alert on display
DI cartridges exhausting quickly
Thermo Scientific Aquanex DI Cartridge Set
7501050
Up to 12 months
"Change DI Packs" alert on display
"Water Quality Alert" on display
Thermo Scientific Barnstead Replacement Ultrafilters
50133980
Up to 24 months
"Change Ultrafilter" alert on display
Low flow from dispenser
Pyrogen in product water that don't improve with disinfection
Thermo Scientific Barnstead Replacement UV Lamps
09.2002
Up to 24 months
"Change UV" alert on display
Higher TOC levels observed
UV lamp is burned out
Thermo Scientific CO2 Absorber + Sterile Filter
06.5002
Up to 12 months
"Change CO2 Vent Filter" alert on display
Color indicator changed from white to purple
Dispenser consumables
Thermo Scientific Barnstead Point of Dispense 1.0 um Filters
50157375
Up to 12 months
"Change POU Final Filter" alert on display
Low flow from dispenser
When DI cartridges are replaced
When bacterial growth is suspected in the POU filter
Videos
Ordering Information
Equipment
Consumables
Accessories
For Laboratory Use.