Search Thermo Fisher Scientific
Nalgene round laboratory bottles are iconic, and used in labs throughout the world. The narrow mouth provides more controlled pouring than wide-mouth bottles. The wide mouth designs are easier to fill and are recommended for solid, semi-solid and viscous liquid materials. Nalgene bottles and their closures are engineered to work together to provide leakproof* containment for lab reagents and samples.
Choose from our rugged lab-quality bottles and large lab bottles or lower cost lighter-weight economy bottles. Thermo Scientific Nalgene bottles are made only from the highest quality plastic materials that meet pharmaceutical, laboratory and food grade standards; our plastic resins are selected to minimize additives and reduce potential leachables.
Nalgene bottles are manufactured in our ISO 13485:2016 registered manufacturing facilities. Most of our plastic resin materials are DMF registered and meet a number of regulatory specifications including: USP Class VI, EP monographs, EU food-contact directives, CONEG, RoHS, CA Prop 65, SARA Title III Sec. 313, and 21 CFR pt 177. Nalgene containers are made from a wide variety of plastic types to meet the needs of virtually any laboratory application.
For help selecting the Nalgene bottle that's perfect for your lab applications, use our online selection guide. For personal assistance, contact the Nalgene technical support team.
Wide-Mouth Round PPCO Lab Quality Bottle (2105-series) | Wide-Mouth Round LDPE Lab Quality Bottle (2103-series) | Wide-Mouth Round HDPE Lab Quality Bottle (2104-series) | Wide-Mouth Round Amber HDPE Lab Quality Bottle (2106-series) | Large Wide-Mouth Round HDPE Bottle (2120-series) | Large Wide-Mouth Round PPCO Bottle (2120-series) |
Traditional reusable lab bottle for long-term use and critical applications, autoclavable | Traditional reusable lab bottle for long-term use and critical applications, squeezably flexible | Traditional reusable lab bottle for long-term use and critical applications | Amber color protects light-sensitive solutions. Traditional reusable lab bottle for long-term use and critical applications | Larger size, rugged lab quality construction | Larger size, rugged lab quality construction, Autoclavable |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
PPCO bottle PP Closure | LDPE bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | HDPE bottle white PP Closure | PPCO bottle white PP Closure |
Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber bottle | Milky white translucent | Milky white translucent |
30mL–1L | 30mL–1L | 30mL–1.5L | 30mL–1L | 2L–4L | 2L–4L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Not liquid leakproof | Not liquid leakproof |
No graduations | No graduations | No graduations | No graduations | No graduations | No graduations |
Temp range -40oC to 121oC | Temp range -100oC to 80oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -40oC to 121oC |
Autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Autoclavable |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use |
BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Also available bulk packed, unassembled | Also available bulk packed, unassembled | Also available bulk packed, unassembled | Also available bulk packed, unassembled |
Wide-Mouth Round PPCO Lab Quality Bottle (2105-series) | Wide-Mouth Round LDPE Lab Quality Bottle (2103-series) | Wide-Mouth Round HDPE Lab Quality Bottle (2104-series) | Wide-Mouth Round Amber HDPE Lab Quality Bottle (2106-series) | Large Wide-Mouth Round HDPE Bottle (2120-series) | Large Wide-Mouth Round PPCO Bottle (2120-series) | |
Acids, dilute or weak | E | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E | E |
Aldehydes | G | G | G | G | G | G |
Bases/alkali | E | E | E | E | E | E |
Esters | G | G | G | G | G | G |
Hydrocarbons, aliphatic | G | F | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Wide-Mouth Round HDPE Economy Bottle (2189-series) | Wide-Mouth Round HDPE Economy Bottle Bulk Pack (332189-series) | Wide-Mouth Round HDPE Amber Economy Bottle (DS2185-series) | Wide-Mouth Round PPCO Economy Bottle (2187-series) |
Lower cost, lighter weight, thinner wall for light-duty applications | Bulk packed for additional cost savings, lighter weight, thinner wall for light-duty applications | Amber color protects light-sensitive solutions. Lower cost, lighter weight, thinner wall for light-duty applications | Lower cost, lighter weight, thinner wall for light-duty applications, autoclavable |
![]() | ![]() | ![]() | ![]() |
HDPE bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | PPCO bottle PP Closure |
Milky white translucent | Milky white translucent | Opaque amber bottle | Milky white translucent |
30mL–1L | 60mL–1L | 30mL–1L | 30mL–1L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
No graduations | No graduations | No graduations | No graduations |
Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -40oC to 121oC |
Not autoclavable | Not autoclavable | Not autoclavable | Autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use |
BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Also available bulk packed, unassembled | Also available bulk packed, unassembled | Also available bulk packed, unassembled | Also available bulk packed, unassembled |
Wide-Mouth Round HDPE Economy Bottle (2189-series) | Wide-Mouth Round HDPE Economy Bottle Bulk Pack (332189-series) | Wide-Mouth Round HDPE Amber Economy Bottle (DS2185-series) | Wide-Mouth Round PPCO Economy Bottle (2187-series) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G |
Alcohols, aliphatic | E | E | E | E |
Aldehydes | G | G | G | G |
Bases/alkali | E | E | E | E |
Esters | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Narrow-Mouth Round PPCO Lab Quality Bottle (2006-series) | Narrow-Mouth Round HDPE Lab Quality Bottle (2002-series) | Narrow-Mouth Round Amber Lab Quality Bottle (2004-series) | Narrow-Mouth Round LDPE Lab Quality Bottle (2003-series) | Narrow-Mouth Round FEP Bottle (1600-series) | Narrow-Mouth Round PFA Bottle (1630-series) |
Good chemical resistance, autoclavable | Good chemical resistance, most popular reagent bottle | Amber color protects light-sensitive solutions, good chemical resistance | Squeezably flexible, good chemical resistance | Exceptional chemical resistance and inertness, wide temp range, high cost | Exceptional chemical resistance and inertness, widest temp range, high cost |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
PPCO bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | LDPE bottle PP Closure | FEP bottle ETFE Closure | PFA bottle PFA Closure |
Milky white translucent | Milky white translucent | Opaque amber bottle | Milky white translucent | Hazy white translucent | Hazy white translucent |
Semi-rigid | Semi-rigid | Semi-rigid | Flexible | Flexible | Flexible |
4mL–1L | 4mL–1L | 4mL–1L | 8mL–1L | 30mL–2L | 30mL–1L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
No graduations | No graduations | No graduations | No graduations | No graduations | No graduations |
Temp range -40oC to 121oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 80oC | Temp range -270oC to 150oC | Temp range -270oC to 250oC |
Autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Autoclavable | Autoclavable |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1550 for food use | No food use compliance |
BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled |
Narrow-Mouth Round PPCO Lab Quality Bottle (2006-series) | Narrow-Mouth Round HDPE Lab Quality Bottle (2002-series) | Narrow-Mouth Round Amber Lab Quality Bottle (2004-series) | Narrow-Mouth Round LDPE Lab Quality Bottle (2003-series) | Narrow-Mouth Round FEP Bottle (1600-series) | Narrow-Mouth Round PFA Bottle (1630-series) | |
Acids, dilute or weak | E | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | E | E |
Alcohols, aliphatic | E | E | E | E | E | E |
Aldehydes | G | G | G | G | E | E |
Bases/alkali | E | E | E | E | E | E |
Esters | G | G | G | G | E | E |
Hydrocarbons, aliphatic | G | G | G | F | E | E |
Hydrocarbons, aromatic | N | N | N | N | E | E |
Hydrocarbons, halogenated | N | N | N | N | E | E |
Ketones, aromatic | N | N | N | N | E | E |
Oxidizing agents, strong | F | F | F | F | E | E |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Narrow-Mouth Round PPCO Economy Bottle (2087-series) | Narrow-Mouth Round HDPE Economy Bottle (2089-series) | Narrow-Mouth Round HDPE Economy Bottle Bulk Pack (332089-series) | Narrow-Mouth Round Amber Economy Bottle (DS2085-series) |
Lower cost, lighter weight, thinner wall for light-duty applications, autoclavable | Lower cost, lighter weight, thinner wall for light-duty applications | Bulk packed for additional cost savings, lighter weight, thinner wall for light-duty applications | Amber color protects light-sensitive solutions. Lower cost, lighter weight, thinner wall for light-duty applications |
![]() | ![]() | ![]() | ![]() |
PPCO bottle PP Closure | HDPE bottle PP Closure | HDPE bottle PP closure | Amber HDPE bottle amber PP Closure |
Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber bottle |
30mL–1L | 30mL–1L | 125mL–1L | 30mL–1L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
No graduations | No graduations | No graduations | No graduations |
Temp range -40oC to 121oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled |
Narrow-Mouth Round Economy Bottle (2087-series) | Narrow-Mouth Round Economy Bottle (2089-series) | Narrow-Mouth Round Economy Bottle Bulk Pack (332089-series) | Narrow-Mouth Round Amber Economy Bottle (DS2085-series) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G |
Alcohols, aliphatic | E | E | E | E |
Aldehydes | G | G | G | G |
Bases/alkali | E | E | E | E |
Esters | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Narrow-Mouth Large Round PPCO Bottle (2203-series) | Large Narrow-Mouth Round Amber Bottle (2204-series) | Large Narrow-Mouth Round LDPE Bottle (2202-series) | Large Narrow-Mouth Round PC Bottle (2205-series) |
Good chemical resistance, autoclavable | Amber color protects light-sensitive solutions. Good chemical resistance, autoclavable | Good chemical resistance, lower cost | Excellent clarity for viewing contents, superior impact resistance, autoclavable |
![]() | ![]() | ![]() | ![]() |
PPCO bottle PP Closure | Amber PPCO bottle amber or white closure | LDPE bottle PP Closure | PC bottle PP Closure |
Milky white translucent | Opaque amber bottle | Milky white translucent | Clear |
2L–8L | 2L–8L | 2L–8L | 500mL–8L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
No graduations | No graduations | No graduations | No graduations |
Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -100oC to 80oC | Temp range -135oC to 135oC |
Autoclavable | Autoclavable | Not autoclavable | Autoclavable |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1580 for food use |
BPA-Free | BPA-Free | BPA-Free | Contains BPA |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Narrow-Mouth Large Round Bottle (2203-series) | Narrow-Mouth Large Round Amber Bottle (2204-series) | Narrow-Mouth Round Lab Quality Bottle (2202-series) | Large Narrow-Mouth Round Bottle (2205-series) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | N |
Alcohols, aliphatic | E | E | E | G |
Aldehydes | G | G | G | F |
Bases/alkali | E | E | E | N |
Esters | G | G | G | N |
Hydrocarbons, aliphatic | G | G | F | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Wide-Mouth Round PPCO Lab Quality Bottle (2105-series) | Wide-Mouth Round LDPE Lab Quality Bottle (2103-series) | Wide-Mouth Round HDPE Lab Quality Bottle (2104-series) | Wide-Mouth Round Amber HDPE Lab Quality Bottle (2106-series) | Large Wide-Mouth Round HDPE Bottle (2120-series) | Large Wide-Mouth Round PPCO Bottle (2120-series) |
Traditional reusable lab bottle for long-term use and critical applications, autoclavable | Traditional reusable lab bottle for long-term use and critical applications, squeezably flexible | Traditional reusable lab bottle for long-term use and critical applications | Amber color protects light-sensitive solutions. Traditional reusable lab bottle for long-term use and critical applications | Larger size, rugged lab quality construction | Larger size, rugged lab quality construction, Autoclavable |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
PPCO bottle PP Closure | LDPE bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | HDPE bottle white PP Closure | PPCO bottle white PP Closure |
Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber bottle | Milky white translucent | Milky white translucent |
30mL–1L | 30mL–1L | 30mL–1.5L | 30mL–1L | 2L–4L | 2L–4L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Not liquid leakproof | Not liquid leakproof |
No graduations | No graduations | No graduations | No graduations | No graduations | No graduations |
Temp range -40oC to 121oC | Temp range -100oC to 80oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -40oC to 121oC |
Autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Autoclavable |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use |
BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Also available bulk packed, unassembled | Also available bulk packed, unassembled | Also available bulk packed, unassembled | Also available bulk packed, unassembled |
Wide-Mouth Round PPCO Lab Quality Bottle (2105-series) | Wide-Mouth Round LDPE Lab Quality Bottle (2103-series) | Wide-Mouth Round HDPE Lab Quality Bottle (2104-series) | Wide-Mouth Round Amber HDPE Lab Quality Bottle (2106-series) | Large Wide-Mouth Round HDPE Bottle (2120-series) | Large Wide-Mouth Round PPCO Bottle (2120-series) | |
Acids, dilute or weak | E | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E | E |
Aldehydes | G | G | G | G | G | G |
Bases/alkali | E | E | E | E | E | E |
Esters | G | G | G | G | G | G |
Hydrocarbons, aliphatic | G | F | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Wide-Mouth Round HDPE Economy Bottle (2189-series) | Wide-Mouth Round HDPE Economy Bottle Bulk Pack (332189-series) | Wide-Mouth Round HDPE Amber Economy Bottle (DS2185-series) | Wide-Mouth Round PPCO Economy Bottle (2187-series) |
Lower cost, lighter weight, thinner wall for light-duty applications | Bulk packed for additional cost savings, lighter weight, thinner wall for light-duty applications | Amber color protects light-sensitive solutions. Lower cost, lighter weight, thinner wall for light-duty applications | Lower cost, lighter weight, thinner wall for light-duty applications, autoclavable |
![]() | ![]() | ![]() | ![]() |
HDPE bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | PPCO bottle PP Closure |
Milky white translucent | Milky white translucent | Opaque amber bottle | Milky white translucent |
30mL–1L | 60mL–1L | 30mL–1L | 30mL–1L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
No graduations | No graduations | No graduations | No graduations |
Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -40oC to 121oC |
Not autoclavable | Not autoclavable | Not autoclavable | Autoclavable |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use |
BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Also available bulk packed, unassembled | Also available bulk packed, unassembled | Also available bulk packed, unassembled | Also available bulk packed, unassembled |
Wide-Mouth Round HDPE Economy Bottle (2189-series) | Wide-Mouth Round HDPE Economy Bottle Bulk Pack (332189-series) | Wide-Mouth Round HDPE Amber Economy Bottle (DS2185-series) | Wide-Mouth Round PPCO Economy Bottle (2187-series) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G |
Alcohols, aliphatic | E | E | E | E |
Aldehydes | G | G | G | G |
Bases/alkali | E | E | E | E |
Esters | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Narrow-Mouth Round PPCO Lab Quality Bottle (2006-series) | Narrow-Mouth Round HDPE Lab Quality Bottle (2002-series) | Narrow-Mouth Round Amber Lab Quality Bottle (2004-series) | Narrow-Mouth Round LDPE Lab Quality Bottle (2003-series) | Narrow-Mouth Round FEP Bottle (1600-series) | Narrow-Mouth Round PFA Bottle (1630-series) |
Good chemical resistance, autoclavable | Good chemical resistance, most popular reagent bottle | Amber color protects light-sensitive solutions, good chemical resistance | Squeezably flexible, good chemical resistance | Exceptional chemical resistance and inertness, wide temp range, high cost | Exceptional chemical resistance and inertness, widest temp range, high cost |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
PPCO bottle PP Closure | HDPE bottle PP Closure | Amber HDPE bottle amber PP closure | LDPE bottle PP Closure | FEP bottle ETFE Closure | PFA bottle PFA Closure |
Milky white translucent | Milky white translucent | Opaque amber bottle | Milky white translucent | Hazy white translucent | Hazy white translucent |
Semi-rigid | Semi-rigid | Semi-rigid | Flexible | Flexible | Flexible |
4mL–1L | 4mL–1L | 4mL–1L | 8mL–1L | 30mL–2L | 30mL–1L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
No graduations | No graduations | No graduations | No graduations | No graduations | No graduations |
Temp range -40oC to 121oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 80oC | Temp range -270oC to 150oC | Temp range -270oC to 250oC |
Autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Autoclavable | Autoclavable |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1550 for food use | No food use compliance |
BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled |
Narrow-Mouth Round PPCO Lab Quality Bottle (2006-series) | Narrow-Mouth Round HDPE Lab Quality Bottle (2002-series) | Narrow-Mouth Round Amber Lab Quality Bottle (2004-series) | Narrow-Mouth Round LDPE Lab Quality Bottle (2003-series) | Narrow-Mouth Round FEP Bottle (1600-series) | Narrow-Mouth Round PFA Bottle (1630-series) | |
Acids, dilute or weak | E | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | E | E |
Alcohols, aliphatic | E | E | E | E | E | E |
Aldehydes | G | G | G | G | E | E |
Bases/alkali | E | E | E | E | E | E |
Esters | G | G | G | G | E | E |
Hydrocarbons, aliphatic | G | G | G | F | E | E |
Hydrocarbons, aromatic | N | N | N | N | E | E |
Hydrocarbons, halogenated | N | N | N | N | E | E |
Ketones, aromatic | N | N | N | N | E | E |
Oxidizing agents, strong | F | F | F | F | E | E |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Narrow-Mouth Round PPCO Economy Bottle (2087-series) | Narrow-Mouth Round HDPE Economy Bottle (2089-series) | Narrow-Mouth Round HDPE Economy Bottle Bulk Pack (332089-series) | Narrow-Mouth Round Amber Economy Bottle (DS2085-series) |
Lower cost, lighter weight, thinner wall for light-duty applications, autoclavable | Lower cost, lighter weight, thinner wall for light-duty applications | Bulk packed for additional cost savings, lighter weight, thinner wall for light-duty applications | Amber color protects light-sensitive solutions. Lower cost, lighter weight, thinner wall for light-duty applications |
![]() | ![]() | ![]() | ![]() |
PPCO bottle PP Closure | HDPE bottle PP Closure | HDPE bottle PP closure | Amber HDPE bottle amber PP Closure |
Milky white translucent | Milky white translucent | Milky white translucent | Opaque amber bottle |
30mL–1L | 30mL–1L | 125mL–1L | 30mL–1L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
No graduations | No graduations | No graduations | No graduations |
Temp range -40oC to 121oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC | Temp range -100oC to 120oC |
Autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-Free | BPA-Free | BPA-Free | BPA-Free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled | Also available in bulk, unassembled |
Narrow-Mouth Round Economy Bottle (2087-series) | Narrow-Mouth Round Economy Bottle (2089-series) | Narrow-Mouth Round Economy Bottle Bulk Pack (332089-series) | Narrow-Mouth Round Amber Economy Bottle (DS2085-series) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G |
Alcohols, aliphatic | E | E | E | E |
Aldehydes | G | G | G | G |
Bases/alkali | E | E | E | E |
Esters | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Narrow-Mouth Large Round PPCO Bottle (2203-series) | Large Narrow-Mouth Round Amber Bottle (2204-series) | Large Narrow-Mouth Round LDPE Bottle (2202-series) | Large Narrow-Mouth Round PC Bottle (2205-series) |
Good chemical resistance, autoclavable | Amber color protects light-sensitive solutions. Good chemical resistance, autoclavable | Good chemical resistance, lower cost | Excellent clarity for viewing contents, superior impact resistance, autoclavable |
![]() | ![]() | ![]() | ![]() |
PPCO bottle PP Closure | Amber PPCO bottle amber or white closure | LDPE bottle PP Closure | PC bottle PP Closure |
Milky white translucent | Opaque amber bottle | Milky white translucent | Clear |
2L–8L | 2L–8L | 2L–8L | 500mL–8L |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
No graduations | No graduations | No graduations | No graduations |
Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range -100oC to 80oC | Temp range -135oC to 135oC |
Autoclavable | Autoclavable | Not autoclavable | Autoclavable |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1580 for food use |
BPA-Free | BPA-Free | BPA-Free | Contains BPA |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Narrow-Mouth Large Round Bottle (2203-series) | Narrow-Mouth Large Round Amber Bottle (2204-series) | Narrow-Mouth Round Lab Quality Bottle (2202-series) | Large Narrow-Mouth Round Bottle (2205-series) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | N |
Alcohols, aliphatic | E | E | E | G |
Aldehydes | G | G | G | F |
Bases/alkali | E | E | E | N |
Esters | G | G | G | N |
Hydrocarbons, aliphatic | G | G | F | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20 oC |
G | Little or no damage after 30 days constant exposure at 20oC |
F | Some effect after 7 days constant exposure at 20oC |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except oxidizing acids; for oxidizing acids, see "Oxidizing acids, strong"
Fluorinated inside and out for improved barrier properties and reduced solvent adsorption and permeation. Thermo Scientific Nalgene Fluorinated Narrow-Mouth HDPE Bottles with Closure are a cost effective alternative to costly fluoropolymer containers. Fluorination enhances long-term container performance, prevents permeation loss and yields lower extractables when used with organic solvents. Available in narrow-mouth and wide-mouth formats.
For assistance choosing products appropriate for your application, please speak with a Nalgene Technical Support Representative team by phone at +1.585.586.8800 or (1.800.625.4327 US toll free), or email your request to
technicalsupport@thermofisher.com.
In Austria, France, Germany, Ireland, Switzerland and the United Kingdom please contact technical support by phone at +800 1234 9696 (toll free) or +49.6184.90.6321, or email your request to techsupport.labproducts.eu@thermofisher.com.
Regulatory Support: For regulatory documentation of product or material claims, please contact Nalgene Regulatory Support at RocRegSupport@thermofisher.com.
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.