Learn the fundamentals of protein expression using the Gibco ExpiCHO Expression System with detailed overviews of the unique system components and key workflow steps for which optimal techniques will help maximize results. This protocol guide includes protocol videos, a protein expression protocol calculator, required material checklists, visual step-by-step guides, and helpful tips.
At the end of this protocol guide, you should be able to:
  • Outline the full ExpiCHO Expression System workflow, identify key components of the system, and know when, how, and why to use each one
  • Apply optimal techniques for thawing and subculturing mammalian suspension‑adapted cells like Gibco ExpiCHO-S Cells to help maximize cell health and protein expression
  • Apply optimal techniques for transfecting mammalian suspension‑adapted cells like Gibco ExpiCHO-S Cells to help maximize cell health and protein expression (available only in the Protein Expression Learning Labs)
  • Demonstrate best practices for efficient protein harvest as well as accurate detection and quantitation of protein yield (available only in the Protein Expression Learning Labs)
  • Identify workflow steps for which optimization will have the most impact to help improve future expression results (available only in the Protein Expression Learning Labs)
Discover the Protein Expression Learning Labs
Experience the full ExpiCHO expression system protocol guide in an immersive and interactive lab environment.
Overview and components of the ExpiCHO Expression System Kit
ExpiCHO expression system overview
The ExpiCHO Expression System is a high-yield transient protein expression system based on suspension-adapted Chinese hamster ovary (CHO) cells. ExpiCHO-S Cells have been specifically adapted to high-density suspension culture in Gibco ExpiCHO Expression Medium. ExpiCHO-S Cells are supplied frozen in ExpiCHO Expression Medium and can be thawed directly in it. This serum-free and chemically defined medium is formulated to support transfection and protein expression in ExpiCHO-S Cells, so there is no need to switch to a different medium. The ExpiCHO Expression System Kit provides enough cells, culture medium, and reagents to transfect 1 L of cell culture. Other than ExpiCHO-S Cells, all components of the kit are animal origin–free (AOF).
product photo
Properties of ExpiCHO-S Cells
  • Rapid recovery after thawing
  • Adapted to high-density suspension growth in ExpiCHO Expression Medium
  • Doubling time of approximately 17 hours
  • Maximum cell density of approximately 2 x 107 cells/mL in shake flask culture
  • Consistent, high-titer transient protein expression performance over 20 passages
Components of ExpiCHO Expression System Kit (Cat. No. A29133)
Component
Quantity
Cat. No.
Storage
ExpiCHO-S Cells (1 x 107 cells/mL)
2 x 1 mL
A29127
Liquid nitrogen*
ExpiCHO Expression Medium
1 L
A2910001
2°C to 8°C (protect from light)
ExpiFectamine CHO Transfection Kit for
1 L of culture:
ExpiFectamine CHO Reagent
ExpiCHO Feed
ExpiFectamine CHO Enhancer
1 kit
3 x 1.1 mL
3 x 110 mL
6 mL
A29129
2°C to 8°C
2°C to 8°C (protect from light)
2°C to 8°C
OptiPRO SFM (complexation medium)
100 mL
12309050
2°C to 8°C (protect from light)
Antibody-Expressing Positive Control Vector
(at 1 mg/mL in TE buffer, pH 8)**
150 µg
A14662
–20°C
* Store frozen cells in liquid nitrogen until ready to use. Do not store cells at –80°C.
** TE buffer, pH 8.0: 10 mM Tris-HCl and 1 mM EDTA, pH 8.0.
Protein expression essentials checklist
Protein expression essentials checklist
The following checklist provides a concise list of recommended consumables, instruments, and equipment to guide you in starting you’re your protein expression
Guidelines for growing and maintaining ExpiCHO-S Cells
General cell handling
  • Important: All solutions and equipment that come into contact with the cells must be sterile. Always use proper sterile technique and work in a laminar flow hood or biosafety cabinet (BSC).
  • Important: Store the frozen cells in liquid nitrogen until ready to use. Do not store the cells at –80°C.
  • Avoid short-term, extreme temperature changes. When storing cells in liquid nitrogen, allow the cells to remain in liquid nitrogen for 3–4 days prior to the first thaw.
  • For all cell manipulations, mix the cells with gentle swirling. Avoid vigorous shaking and pipetting.
  • Before starting your experiments, be sure to have cells established and have some frozen stocks on hand.
  • Upon receipt, grow and freeze multiple vials of ExpiCHO-S Cells to ensure that you have an adequate supply of early passage cells.
  • Allow freshly thawed cells to recover in culture for two or more passages post-thaw prior to transfection.
  • ExpiCHO-S Cells are adapted to high-density growth conditions with a doubling time of approximately 17 hours. They have a broad log phase growth window spanning from approximately 4 x 106 to 1.5 x 107 cells/mL with a maximum density of approximately 2 x 107 cells/mL in shake flask cultures.
  • For general maintenance, passage ExpiCHO-S Cells when they reach a density of approximately 4 x 106 to 6 x 106 viable cells per mL in early log phase growth, typically every 3–4 days.
  • Note: Cells that are subcultured at densities outside of this early log phase growth window may have longer doubling times with lower titers over time. Modify the initial seeding density to attain the target cell density of 4 x 106 to 6 x 106 viable cells per mL at the time of subculturing.
  • Use a hemocytometer with the trypan blue exclusion method or an automated cell counter to determine cell viability. Log phase cultures should be >95% viable.
  • When thawing or subculturing cells, transfer them into pre-warmed medium.
Medium
  • ExpiCHO Expression Medium is formulated with Gibco GlutaMAX-I Supplement. Therefore, ExpiCHO Expression Medium does not require supplementation for suspension growth and transfection applications.
Important: ExpiCHO Expression Medium is sensitive to light. For optimal results, keep it protected from light during use and storage.
Thawing and establishing ExpiCHO-S Cells
Use the following protocol for thawing ExpiCHO-S Cells to initiate cell culture.
IMPORTANT: Store the frozen cells in liquid nitrogen until ready to use. The cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells per mL in 90% ExpiCHO Expression Medium and 10% DMSO. Thaw the cells directly into ExpiCHO Expression Medium that has been warmed to 37°C.
Avoid short-term, extreme temperature changes. Allow the cells to remain in liquid nitrogen for 3–4 days prior to thawing.
product photo
Required materials
Step-by-step protocol for thawing and establishing ExpiCHO-S Cells
Thaw cells (Day 1)
1. Remove the vial of cells from liquid nitrogen and swirl it in a 37°C water bath for 1–2 min to rapidly thaw the cells until only a small amount of ice remains. Note: Do not fully submerge the vial in the water bath.
2. Decontaminate the external surface of the vial by wiping it with 70% ethanol before opening the vial in the BSC or laminar flow hood.
Add cells to medium (Day 1)
3. Carefully pipette the entire contents of the vial into the 125 mL Erlenmeyer shake flask containing 30 mL pre‑warmed ExpiCHO Expression Medium.
Incubate cells
4. Incubate the cells at 37°C on the orbital shaker with ≥80% relative humidity and 8% CO2.
Shaking speed
19 mm shaking diameter: 125 ± 5 rpm
25 mm shaking diameter: 120 ± 5 rpm
50 mm shaking diameter: 95 ± 5 rpm
Count cells and determine viability (Day 3)
5. On Day 3 post-thaw, determine viable cell density and percent cell viability. Cell viability should be ≥90%.
6. Continue to monitor cell density and viability until the culture has reached 4 x 106 to 6 x 106 viable cells per mL, typically three or four days post-thaw, then proceed to subculturing (next segment).
Subculturing ExpiCHO-S Cells
Helpful tips for subculturing ExpiCHO-S Cells
Since ExpiCHO-S Cells are adapted to high‑density suspension culture in ExpiCHO Expression Medium, we recommend allowing cells to reach a minimum density of 4 x 106 to 6 x 106 viable cells per mL at the subculturing stage. You may observe some clumping during routine cell culture maintenance at higher cell densities, but this will not diminish the performance of the cells in subsequent steps.
Required materials
Step-by-step protocol for subculturing ExpiCHO-S Cells
Passage cells (Day 1)
1. Based on the measured viable cell density, calculate the volume of cell suspension required to seed a new shake flask according to the recommended seeding densities in Table 1 and the recommended culture volumes in Table 2.
2. Transfer the calculated volume of cells to pre-warmed ExpiCHO Expression Medium in the Erlenmeyer shake flask(s).
Subculturing and harvest
3. Incubate flask(s) on the orbital shaker in the 37°C incubator with ≥80% relative humidity and 8% CO2 until culture(s) reach a density of 4 x 106 to 6 x 106 viable cells per mL.
Note: Cells that are subcultured at densities outside of this early log-phase growth window may show longer doubling times and lower titers over time. If necessary, modify the initial seeding density to attain the target cell density of 4 x 106 to 6 x 106 viable cells per mL at the subculturing stage.
4. Repeat Steps 1–3 to maintain or expand the cells for transfection.
Table 2. Recommended volumes and shaking speeds for routine cell culture maintenance.
Flask size
125 mL
250 mL
500 mL
1L
2 L
3 L
Culture volume (mL)
30–35
60–70
120–140
240–280
500–560
600–1,000
Shaking speed*
19 mm shaking diameter: 125 ± 5 rpm
25 mm shaking diameter: 120 ± 5 rpm
50 mm shaking diameter: 95 ± 5 rpm
90 ± 5 rpm
85 ± 5 rpm
80 ± 5 rpm
Flask type
Vented, non-baffled
For Research Use Only. Not for use in diagnostic procedures.