Search Thermo Fisher Scientific
The Applied Biosystems 3500 Series Genetic Analyzer is designed to maintain the unsurpassed application versatility that life science researchers expect from Applied Biosystems instruments.
Looking for more out of your instrument? Our newest genetic analyzer, the SeqStudio Flex, delivers the reliable performance you need while leveraging design and technological improvements to offer increased flexibility, connectivity, serviceability, and easier operation for users of all skill levels.
Explore the benefits of SeqStudio Flex ›
It's accurate
Based on one of the most widely used, widely trusted sequencing methodologies available—Sanger sequencing—the 3500 Series Genetic Analyzer is designed to deliver the accuracy you demand.
It works the way you do
Designed with you in mind, the 3500 Series Genetic Analyzer gives you automated operation, easy-to-install consumables, electronic tracking of instrument performance and maintenance tasks, and integrated analysis software—so you can focus on the outcome.
It's flexible
The 3500 platform can run a wide variety of Sanger sequencing and fragment analysis applications, including de novo sequencing and resequencing (mutational profiling), microsatellite analysis, MLPA, AFLP, LOH, and SNP validation or screening.
It's backed by award-winning support
Keeping your lab operational is critical. Our technical and field service support staffs respond quickly to your calls, so that you can continue to respond to those who depend on you for results.
The Applied Biosystems 3500 and 3500xL Genetic Analyzers are supplied as follows:
Capillary array | |
![]() |
|
Cathode buffer container | |
![]() |
|
Performance optimized polymer | |
![]() |
|
Anode buffer container | |
![]() |
|
Putting you in the driver's seat | |
![]() | 3500 Data Collection Software offers an intuitive dashboard design including:
|
Operates on standard power | |
![]() | The 3500 Series is easy to install and operate:
|
Feature | Benefit |
---|---|
RFID | Instrument automatically checks for the use of correct product and expiration dates |
SAE | Security, audit and electronic signature capability to help compliance with governing regulations |
8-capillary or 24-capillary models | Select the appropriate instrument to meet your laboratory throughput needs |
Flexibility | Allows you to adhere to your lab's development requirements—sequencing or fragment analysis applications |
Aneuploidy Detection by QF-PCR of STR Markers on the Applied Biosystems 3500xL Genetic Analyzer
BigDye Direct Cycle Sequencing Kit simplifies workflow for characterizing melanoma mutations in research samples
Development of a Workflow to Detect Sequence Variants in the BRCA1 and BRCA2 Genes
Development of an Interlaboratory-Verified Sequencing Workflow for KRAS Variant Identification
DNA Resequencing Analysis of X-linked Variants Using the Applied Biosystems 3500xL Genetic Analyzer
Fungal DNA sequencing from laser capture microdissection samples
High coverage gene expression profiling on the Applied Biosystems 3500xL Genetic Analyzer (1.6 MB)
ISSR Genotyping of Endangered Plants Using an Optimized Workflow
Resequencing Workflow for MLH1 and MSH2 Variant Analysis
SNP Gentyping Using SNaPshot Multiplex Kit and 3500 Series Genetic Analyzer
Uniparental disomy (UPD) analysis of chromosome 15 (3.2 MB)
Beyond repair to proactive care
The AB Assurance Plan is our premium repair plan, designed to maximize instrument performance and help ensure availability of critical systems. The AB Assurance Plan will help keep your laboratory running smoothly with preventive maintenance, proactive instrument monitoring and—should one of your instruments require repair—fast response.
Qualification services
In regulated industries, compliance with government and international standards requires documented verification that your systems are installed and functioning according to their manufacturers’ specifications. Whether you need Installation Qualification and Operation Qualification (IQ/OQ) or Instrument Performance Qualification (PQ), our qualification services provide audit-quality data to share with your regulated body and verify that your instrument is working according to the manufacturer's specifications.
Computer System Validation (CSV)
An instrument qualification service is only the starting point to your validation. Our Compliance Services professionals partner with you and can help you get into production faster, while controlling CSV costs and getting you audit-ready faster.
AB Assurance with qualification services* | AB Assurance with system performance check* | |
---|---|---|
On-site response time for repairs (business days) | Guaranteed 2 business days† | Guaranteed 2 business days |
Remote instrument monitoring and diagnostics | ![]() | ![]() |
Priority phone and email access to instrument support | ![]() | ![]() |
Application technical support | ![]() | ![]() |
Hearing Loss Research Panel | ![]() | ![]() |
Instrument operating software | ![]() | ![]() |
Instrument qualification services | ![]() | |
Instrument system performance check | ![]() | |
Planned maintenance | ![]() | ![]() |
Parts, labor and travel | ![]() | ![]() |
*Terms and conditions apply. Contact your local service representative for terms and conditions applicable to your region.
†Guaranteed service timeline applies only to North America region.
Trade in your Applied Biosystems™ 3100 or 3130 Series Genetic Analyzer for a new 3500 Series Genetic Analyzer at an incredible price.
Instruments
Buffer & conditioning reagents
For Research Use Only. Not for use in diagnostic procedures.