Unlike the polydisperse dextrans (Fluorescent and Biotinylated Dextrans—Section 14.5), fluorescent protein tracers have molecular weights that are reasonably well defined (bovine serum albumin (BSA) ~66,000 daltons; ovalbumin ~45,000; codfish parvalbumin 12,328 daltons, casein ~23,600 daltons; monomeric subunit B of cholera toxin ~12,000 daltons; horseradish peroxidase (HRP) ~40,000 daltons; soybean trypsin inhibitor ~21,500 daltons; Phaseolus vulgaris leucoagglutinin (PHA-L) ~126,000 daltons). Some of their applications are similar to those of dextran tracers, although protein conjugates may be more susceptible to proteolysis. Through creative modifications of the proteins, however, researchers have been able to use these conjugates to target receptors, to detect protease activity and to measure intracellular pH.
Albumin Conjugates
Fluorescent BSA, Ovalbumin and Parvalbumin
For use as protein tracers, we supply bovine serum albumin (BSA) fluorescently labeled with:
In addition to these BSA conjugates, we prepare fluorescein and Texas Red conjugates of ovalbumin (O23020, O23021), as well as the intensely fluorescent Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594 and Alexa Fluor 647 conjugates (O34781, O34782, O34783, O34784). Ovalbumin is a protein with a relatively low molecular weight (~45,000 daltons) that is primarily useful as an antigen for activation of dendritic cells, macrophages and B cells.ref
We also offer the Alexa Fluor 488 conjugate of codfish parvalbumin (P23012), which is the smallest fluorescent protein tracer in this section. Parvalbumin, a stable and protease-resistant protein with a molecular weight of 12,328 daltons, is found in the skeletal muscle of vertebrates and the endocrine glands of mammals.ref
Dinitrophenylated Bovine Serum Albumin
We offer BSA that has been conjugated to multiple dinitrophenyl hapten molecules (DNP-BSA, A23018), a reagent that is commonly used to study Fc receptor–mediated immune function ref and the IgE- and IgG-mediated responses to crosslinking of DNP-specific antibodies.ref Our dinitrophenylated BSA typically has ~25 dinitrophenyl substituents per molecule of BSA. For detection of DNP-BSA, we offer unlabeled rabbit anti-DNP antibody prepared against DNP-conjugated keyhole limpet hemocyanin (anti–DNP-KLH antibody) and the Alexa Fluor 488 dye, fluorescein and biotin-XX antibody conjugates (A6430, A11097, A6423, A6435; Anti-Dye and Anti-Hapten Antibodies—Section 7.4), as well as a Qdot 555 rat monoclonal anti-dinitrophenol antibody (Q17421MP).
DQ BSA, DQ Ovalbumin and DQ Collagen
DQ substrates, including the DQ Green BSA and DQ Red BSA (D12050, D12051) and DQ ovalbumin (D12053) are based on the strong fluorescence quenching effect observed when proteins are heavily conjugated with BODIPY dyes or, in the case of DQ collagen conjugates (D12052, D12060), with multiple fluorescein dyes. Upon hydrolysis of the proteins to single dye–labeled peptides by proteases, this quenching is relieved (Figure 14.7.1). This technology provides an easy continuous assay for numerous proteases (Detecting Peptidases and Proteases—Section 10.4). When complexed with an anti-BSA rabbit IgG antibody (A11133), the DQ BSA products are phagocytosed into live cells via the Fc receptor (Probes for Following Receptor Binding and Phagocytosis—Section 16.1, Figure 14.7.2), where they are broken down into fluorescent peptides ref (Figure 14.7.1). DQ Green BSA, which gives bright green fluorescence upon proteolysis, and DQ Red BSA, which yields red-fluorescent hydrolysis products, are packaged in sets of five vials, each containing 1 mg of the substrate. Our Fc OxyBURST Green assay reagent (F2902, Probes for Following Receptor Binding and Phagocytosis—Section 16.1), which is based on a similar concept, comprises BSA that has been conjugated to a dihydrofluorescein derivative and then complexed with anti-BSA IgG. Upon binding to the Fc receptor, the immune complex is internalized and oxidized to a fluorescent product within the phagovacuole.ref
We have found our highly quenched DQ ovalbumin (D12053) to be particularly useful for antigen processing and presentation studies. Ovalbumin conjugates are internalized via the mannose receptor–mediated endocytosis pathway and are recognized by antigen-presenting cells. BSA that was heavily labeled with fluorescein (FITC) has been used for these studies.ref However, fluorescence of the intracellular hydrolysis products of FITC BSA is significantly decreased by the acidity of the phagovacuole. In contrast, the bright fluorescence of the BODIPY FL fluorophore in DQ ovalbumin is completely independent of the pH. The fluorescence generated after DQ collagen (D12052) is hydrolyzed by cellular collagenase has been used to visualize the migratory pathway followed by tumor cells during invasion of a gelatin matrix and to image proteolysis by living breast cancer cells.ref
Enzyme detection
Figure 14.7.1
Principle of enzyme detection via the disruption of intramolecular self-quenching. Enzyme-catalyzed hydrolysis of the heavily labeled and almost totally quenched substrates provided in our EnzChek Protease Assay Kits (E6638, E6639), EnzChek Ultra Amylase Assay Kit (E33651), EnzChek Gelatinase/Collagenase Assay Kit (E12055), EnzChek Elastase Kit (E12056), EnzChek Lysozyme Assay Kit (E22013)—as well as the stand-alone quenched substrates DQ BSA (D12050, D12051), DQ collagen (D12052, D12060), DQ ovalbumin (D12053) and DQ gelatin (D12054)—relieves the intramolecular self-quenching, yielding brightly fluorescent reaction products.
Immune complex BSA conjugate 
Figure 14.7.2 Immune complex of DQ BSA conjugate (D12050, D12051) with an anti–bovine serum albumin (BSA) antibody (A11133) for the fluorescent detection of the Fc receptor–mediated phagocytosis pathway. The DQ BSA is a derivative of BSA that is labeled to such a high degree with either the green-fluorescent BODIPY FL or red-fluorescent BODIPY TR-X dye that the fluorescence is self-quenched. Upon binding to an Fc receptor, the nonfluorescent immune complex is internalized and the protein is subsequently hydrolyzed to fluorescent peptides within the phagovacuole.
Injectable Contrast Reagents for Small Animal in vivo Imaging Applications
The study of vascularization—often in tumors—requires a probe that will adequately distribute throughout the system under study without being cleared from the tissue prior to the time needed to perform imaging. For this purpose, bovine serum albumin (BSA) and human serum transferrin labeled with near-infrared–fluorescent Alexa Fluor 680 (S34788, S34790) and Alexa Fluor 750 (S34789, S34791) dyes are offered as injectable contrast agents in small animal in vivo imaging.ref These SAIVI injectable contrast agents have been optimized for emission intensity and tested as reagents for the in vivo imaging of regions of blood pooling and vascularization in an inflammatory disease model in mice. Infrared fluorescent microspheres (Microspheres—Section 6.5) can also be used in a similar manner.
Casein Conjugates
BODIPY FL casein and BODIPY TR-X casein, which are components of our EnzChek Protease Assay Kits (E6638, E6639), are substrates for metallo-, serine, acid and sulfhydryl proteases, including cathepsin, chymotrypsin, elastase, papain, pepsin, thermolysin and trypsin ref (Detecting Peptidases and Proteases—Section 10.4). These casein-based substrates are heavily labeled and therefore highly quenched conjugates; they typically exhibit <3% of the fluorescence of the corresponding free dyes. Protease-catalyzed hydrolysis relieves this quenching, yielding brightly fluorescent peptides (Figure 14.7.1).
The EnzChek Protease Assay Kit (E6638, E6639) includes:
Each kit provides sufficient reagents for ~100 assays using 2 mL assay volumes and standard fluorescence cuvettes or ~1000 assays using 200 µL assay volumes and 96-well microplates.
The EnzChek Protease Assay Kits, as well as the DQ BSA and DQ ovalbumin substrates mentioned above, have significant potential as:
  • Indicators of protease contamination in culture media
  • Fluorogenic substrates for circulating or secreted proteases in extracellular fluids
  • Nontoxic, pH-insensitive markers for phagocytic cells, which will ingest and eventually cleave the quenched casein substrates to yield fluorescent BODIPY FL– or BODIPY TR-X–labeled peptides
  • Microinjectable tracers to detect enhanced protease activity associated with cell activation and fusion
  • Nontoxic markers for assessing various cell-loading and cell-transfection techniques, including electroporation (Techniques for loading molecules into the cytoplasm—Table 14.1)
The peptide hydrolysis products of BODIPY FL casein exhibit green fluorescence that is optimally excited by the argon-ion laser at 488 nm, permitting flow sorting of the cells. The red-fluorescent BODIPY TR-X–labeled peptides, with excitation and emission spectra similar to those of the Texas Red fluorophore, should be useful for multilabeling experiments or measurements in the presence of green autofluorescence. Following intracellular hydrolysis, some of the lower molecular weight fluorescent peptides may diffuse into other organelles or pass thorough gap junctions.
Phaseolus vulgaris Leucoagglutinin: An Important Anterograde Tracer
Our Alexa Fluor 488, Alexa Fluor 594 and Alexa Fluor 647 conjugates (L11270, L32456, L32457) of Phaseolus vulgaris leucoagglutinin (PHA-L) are excellent fluorescent anterograde tracers.ref Iontophoretically injected PHA-L clearly demonstrates the morphological features of the filled neurons at the injection site and the labeled axons and axon terminals. Furthermore, PHA-L that has been transported is not degraded over long periods. PHA-L has been used in combination with our biotin dextran amines ref (BDA dextrans, Fluorescent and Biotinylated Dextrans—Section 14.5) or the fluorescent dextrans fluoro-ruby and mini-ruby ref (D1817, D3312; Fluorescent and Biotinylated Dextrans—Section 14.5) to demonstrate their similar transport properties and to show the distribution of anterograde-labeled fibers.ref PHA-L has been simultaneously injected with the retrograde tracer hydroxystilbamidine (H22845, Polar Tracers—Section 14.3) for double-labeling experiments.ref
Cholera Toxin Subunit B
Cholera toxin comprises two subunits, A and B, arranged in an AB5 conformation. Subunit A is an ADP-ribosyltransferase, which disrupts the proper signaling of G proteins and eventually leads to dehydration of the cell.ref The 12,000-dalton nontoxic subunit B ("choleragenoid"), which is assembled into a 60,000-dalton pentamer above pH 2,ref allows the protein complex to bind to cellular surfaces via the pentasaccharide chain of ganglioside GM1.ref All of Molecular Probes cholera toxin subunit B conjugates are prepared from recombinant cholera toxin subunit B, which is completely free of the toxic subunit A, thus eliminating any concern for toxicity or ADP-ribosylating activity.
Cholera toxin subunit B and its conjugates are established as superior tracers for retrograde labeling of neurons.ref Cholera toxin subunit B conjugates bind to the pentasaccharide chain of ganglioside GM1 on neuronal cell surfaces and are actively taken up and transported; alternatively, they can be injected by iontophoresis. Unlike the carbocyanine-based neuronal tracers such as DiI (D282, D3911, V22885; Tracers for Membrane Labeling—Section 14.4), cholera toxin subunit B conjugates can be used on tissue sections that will be fixed and frozen.ref Our Alexa Fluor 488 (C22841, C34775), Alexa Fluor 555 (C22843, C34776), Alexa Fluor 594 (C22842, C34777) and Alexa Fluor 647 (C34778) conjugates of cholera toxin subunit B combine this versatile tracer with the superior brightness of our Alexa Fluor dyes to provide sensitive and selective receptor labeling and neuronal tracing.ref We also offer biotin-XX (C34779) and horseradish peroxidase (C34780) conjugates of cholera toxin subunit B for use in combination with diaminobenzidine (DAB) oxidation,ref tyramide signal amplification (TSA) techniques (TSA and Other Peroxidase-Based Signal Amplification Techniques—Section 6.2) and Qdot nanocrystal–streptavidin conjugates.ref
Fluorescent cholera toxins are also markers of lipid rafts—regions of cell membranes high in ganglioside GM1 that are thought to be important in cell signaling.ref The Vybrant Lipid Raft Labeling Kits (V34403, V34404, V34405; Tracers for Membrane Labeling—Section 14.4) provide the key reagents, including fluorescent cholera toxin subunit B conjugates, for fluorescently labeling lipid rafts in vivo with our bright and extremely photostable Alexa Fluor dyes (photo).
Fluorescent Protein–Based Cytosol Marker
Fluorescent protein–based cytosolic marker BacMam GFP transduction control (B10383) incorporates all the generic advantages of BacMam 2.0 delivery technology (BacMam Gene Delivery and Expression Technology—Note 11.1) to produce nonspecific Green Fluorescent Protein (GFP) expression in a wide range of cell types. Once initiated, GFP expression lasts about 5 days in cell lines such as HeLa and CHO. In cells that divide more slowly or show contact inhibition, such as some stem cells, primary cells and neurons, we have observed useful levels of GFP expression for more than 2 weeks. Within this timeframe, labeled cells can be used for any purpose requiring stable incorporation of a passive fluorescent marker in live cells, including wound healing, migration or adhesion assays. In adoptive transfer applications, cells with undetectable levels of GFP fluorescence can be identified immunohistochemically using our anti-GFP antibodies (Anti-Dye and Anti-Hapten Antibodies—Section 7.4).
Phycobiliproteins
Phycobiliproteins make effective polar tracers because they are intensely fluorescent, are very soluble in water and have little tendency to bind nonspecifically to cells (Phycobiliproteins—Section 6.4). These intrinsically fluorescent biomolecules are stable proteins that are monodisperse and substantially larger than albumins, with molecular weights of 240,000 daltons (B- and R-phycoerythrin, P800, P801) and 104,000 daltons (allophycocyanin, A803, or crosslinked allophycocyanin, A819). Phycoerythrin has been conjugated to nuclear-localization peptide sequences to follow nuclear import processes.ref B-phycoerythrin can pass through pores in erythrocyte membranes created by streptolysin O.ref Single molecules of R-phycoerythrin have been imaged and tracked within mammalian cells.ref
Alexa Fluor 488 Soybean Trypsin Inhibitor
The high water solubility and chemical stability of soybean trypsin inhibitor (SBTI) may make its green-fluorescent Alexa Fluor 488 conjugate (T23011) a useful polar tracer. This conjugate has been used as a probe for the acrosome reaction of spermatozoa.ref
Ordering Information
Catalog # Name Size Price (USD) Qty
A34786 Albumin from Bovine Serum (BSA), Alexa Fluor™ 555 conjugate Each
342.65
Online Exclusive
353.00
Save 10.35 (3%)
A34787 Albumin from Bovine Serum (BSA), Alexa Fluor™ 680 conjugate Each
343.00
A13100 Albumin from Bovine Serum (BSA), Alexa Fluor™ 488 conjugate Each
337.65
Online Exclusive
348.00
Save 10.35 (3%)
A13101 Albumin from Bovine Serum (BSA), Alexa Fluor™ 594 conjugate Each
339.00
A34785 Albumin from Bovine Serum (BSA), Alexa Fluor™ 647 conjugate Each
348.00
A23018 DNP-BSA (Albumin from Bovine Serum (BSA), 2,4-Dinitrophenylated) Each
182.00
A23015 Albumin from Bovine Serum (BSA), FITC conjugate Each
187.65
Online Exclusive
193.00
Save 5.35 (3%)
A23016 Albumin from Bovine Serum (BSA), Tetramethylrhodamine conjugate Each
189.00
A23017 Albumin from Bovine Serum (BSA), Texas Red™ conjugate Each
188.65
Online Exclusive
194.00
Save 5.35 (3%)
A11133 Bovine Serum Albumin Polyclonal Antibody 500 µL
358.00
C22841 Cholera Toxin Subunit B (Recombinant), Alexa Fluor™ 488 Conjugate, 500 μg Each
945.65
Online Exclusive
984.00
Save 38.35 (4%)
C34775 Cholera Toxin Subunit B (Recombinant), Alexa Fluor™ 488 Conjugate, 100 μg Each
338.65
Online Exclusive
352.00
Save 13.35 (4%)
C22843 Cholera Toxin Subunit B (Recombinant), Alexa Fluor™ 555 Conjugate, 500 μg Each
953.65
Online Exclusive
1,002.00
Save 48.35 (5%)
C34776 Cholera Toxin Subunit B (Recombinant), Alexa Fluor™ 555 Conjugate, 100 μg Each
336.65
Online Exclusive
347.00
Save 10.35 (3%)
C22842 Cholera Toxin Subunit B (Recombinant), Alexa Fluor™ 594 Conjugate, 500 μg Each
962.00
C34777 Cholera Toxin Subunit B (Recombinant), Alexa Fluor™ 594 Conjugate, 100 μg Each
343.00
C34778 Cholera Toxin Subunit B (Recombinant), Alexa Fluor™ 647 Conjugate Each
328.65
Online Exclusive
338.00
Save 9.35 (3%)
C34779 Cholera Toxin Subunit B (Recombinant), Biotin-XX Conjugate Each
345.00
C34780 Cholera Toxin Subunit B (Recombinant), Horseradish Peroxidase Conjugate Each
330.00
D12060 DQ™ Collagen, type I From Bovine Skin, Fluorescein Conjugate Each
445.00
D12052 DQ™ Collagen, type IV From Human Placenta, Fluorescein Conjugate Each
446.00
D12050 DQ™ Green BSA - Special Packaging Each
576.00
D12053 DQ™ Ovalbumin - Special Packaging Each
562.65
Online Exclusive
582.00
Save 19.35 (3%)
D12051 DQ™ Red BSA - Special Packaging Each
565.65
Online Exclusive
586.00
Save 20.35 (3%)
E6638 EnzChek™ Protease Assay Kit, green fluorescence, 100-1000 assays Each
531.65
Online Exclusive
558.00
Save 26.35 (5%)
E6639 EnzChek™ Protease Assay Kit, red fluorescence, 100-1000 assays Each
548.00
L11270 Lectin PHA-L From Phaseolus vulgaris (red kidney bean), Alexa Fluor™ 488 Conjugate Each
196.00
L32456 Lectin PHA-L From Phaseolus vulgaris (red kidney bean), Alexa Fluor™ 594 Conjugate Each
197.00
O34781 Ovalbumin, Alexa Fluor™ 488 Conjugate Each
170.65
Online Exclusive
176.00
Save 5.35 (3%)
O34782 Ovalbumin, Alexa Fluor™ 555 Conjugate Each
166.00
O34784 Ovalbumin, Alexa Fluor™ 647 Conjugate Each
175.65
Online Exclusive
181.00
Save 5.35 (3%)
O23020 Ovalbumin, Fluorescein Conjugate Each
171.65
Online Exclusive
177.00
Save 5.35 (3%)
O23021 Ovalbumin, Texas Red™ Conjugate Each
167.00
P801 R-Phycoerythrin Each
375.00
B10383 BacMam GFP Transduction Control (BacMam 2.0) Each -
For Research Use Only. Not for use in diagnostic procedures.