decorative
Advance to clinical applications with CTS NK cell culture media and reagents
Natural killer (NK) cells are innate lymphocytes that kill virally infected or malignant cells. Unlike T cells, NK cells function in an antigen independent manner, responding to anything they perceive as "non-self", including malignant cells. NK cells are an attractive allogeneic immune modulator, as they have a well-known safety profile and lack the potential to cause graft-vs-host disease, a risk associated with CAR-T cell immunotherapy. This makes them an attractive cell type for development of off-the-shelf immunotherapies [1].
Pre-clinical and clinical studies have demonstrated safety and efficacy of allogeneic NK cells against various malignancies—both hematological and solid tumors, without the need for strict HLA-matching. Several clinical trials are currently ongoing which use NK or CAR-NK cells as off-the-shelf allogeneic therapeutics. However, many NK cells are needed in order to infiltrate solid tumors effectively. Generation of sufficient cell numbers for off-the-shelf allogeneic translational, cancer immunotherapy requires efficient scale-up culture conditions. For this, Thermo Fisher Scientific has developed Gibco Cell Therapy Systems (CTS) products across the cell therapy workflow to facilitate the transition from discovery to commercial manufacturing.
Processes and products for cell isolation in natural killer cell therapy
Human peripheral blood mononuclear cells (PBMCs) are a rich source of many immune cell types, including NK cells. Often, PBMCs are isolated from whole peripheral blood or leukapheresis preparation via density gradient separation, but this requires a lengthy, manual process, which can be prone to error and contamination.
Alternatively, the CTS Rotea Counterflow Centrifugation System offers a closed, automatable solution for PBMC isolation.
NK cells can be isolated from the purified PBMC preparation using immunomagnetic beads such as the Invitrogen Dynabeads Untouched Human NK Cells Kit.
NK cell culture media and reagents for cell expansion
Feeder-free NK cell expansion systems are the desirable choice for clinical applications due to their reduced safety risks. However, the historically lower expansion rates of feeder-free culture systems present a challenge to widespread clinical adoption and are a key reason why feeder-based expansion systems are frequently used today, despite the safety and regulatory risks they pose.
product photo
product photo
Gibco CTS NK-Xpander Medium was designed to proliferate a high yield of functional human NK cells to address the needs of translational and clinical cell therapy researchers with or without the use of feeder cells.
  • Innovative performance—This NK cell media is capable of expanding a high yield of human NK cells without the need for feeder cells; cells maintain CD56+ and CD16+ expression and demonstrate cytolytic capabilities
  • Reduction of regulatory risks—NK cell media designed to help meet regulatory compliance requirements for ancillary materials used in cell and gene therapy manufacturing; manufactured AOF (human AB serum supplementation  is recommended for best results); offers Drug Master File, Certificate of Origin, Certificate of Analysis, and Safety Data Sheet
  • Closed-system compatible format available off-the-shelf—NK cell media available in 500 mL bottles and 5 L bag kit formats for convenient process development and closed-system manufacturing scale-up
CTS NK-Xpander Medium is available in 5 L bags and 500 mL bottles.
graphed results
Figure 1. NK cell media, CTS NK-Xpander Medium, offers significantly higher fold expansion than all other media systems tested, including a specialty NK culture medium (Supplier 1) (p<0.0001) with average expansion greater than 1,500-fold after 14 days of small-scale culture (n=20 donors).
Gibco PeproTech growth factors and cytokines for NK cell therapy
Thermo Fisher Scientific offers growth factors, cytokines, and other NK cell-related recombinant proteins in the Gibco PeproTech portfolio to effectively maintain and expand human NK cells. 
Learn more about how PeproTech and PeproGMP recombinant proteins can support your NK cell therapy applications.
Learn more
product photo
Order cell expansion products for NK cell therapy
Genetic modification in natural killer cell therapy
Lentiviral methods
NK cells are often genetically modified to improve NK cell function, persistence, and trafficking to tumor sites. Lentiviral vectors can be used to make genetic modifications such as insertion of chimeric antigen receptors. The Gibco CTS LV-MAX Lentiviral Production System offers a scalable and high-yield lentiviral vector production platform that is based on a suspension culture of HEK 293-derived Viral Production Cells adapted to a special chemically defined, serum-free, and protein-free CTS LV-MAX Production Medium.
The CTS LV-MAX Lentiviral Production System offers
  • High titer—Greater than 1 x 108 TU/mL (unconcentrated LVVs-GFP)
  • Scalable suspension system—From 96 deep-well plate to 2 L bioreactor
  • Serum-free—Eliminating animal-origin components and associated risks
  • Easy, robust culture and transfection protocols—Fits into current transient production workflows
Non-viral gene editing methods
As you move your genome editing projects from basic research to clinical settings, high-quality cell therapy ancillary materials and documentation are critical. We now offer our Gibco CTS TrueCut Cas9 Protein for clinical research that requires Cas9 nuclease that is manufactured with stringent specifications.
  • Exceptional performance—Consistently high performance in all tested cell lines, including >90% editing in iPSCs
  • Higher stringency manufacturing—Manufactured in compliance for Ancillary Materials for Cell, Gene, and Tissue-Based Products
  • Extensive safety testing—Including sterility, endotoxin, mycoplasma, residual host nucleic acids, and proteins
See data
Discover a closed, scalable electroporation system for your NK cell therapy development and manufacturing
product photo
Gibco CTS Xenon Electroporation System
  • High speed and large volume
  • Scalable, proven performance and viability
  • Process flexibility
  • Efficient nonviral transfection
  • Closed-system processing
Learn more
Washing and concentrating cells in natural killer cell therapy
NK cell therapy culture volumes can often reach final volumes of 10–20 L, which must be concentrated and undergo buffer exchange prior to final product formulation. The Gibco CTS Rotea Counterflow Centrifugation System is a closed, automatable alternative to commonly used manual centrifugation processes and offers exceptional flexibility in cell therapy development and manufacturing to enable scaling from research through commercial manufacturing.
Discover a closed, scalable cell processing system for your NK cell therapy research and manufacturing
product photo
Gibco CTS Rotea Counterflow Centrifugation System
  • Process flexibility
  • High cell recovery and viability
  • Low output volumes
  • Research through commercial manufacturing
Learn more
Order cell washing and concentration products for NK cell therapy
Cell characterization in natural killer cell therapy
NK cells are traditionally identified by the presence of CD56 and absence of CD3 on their cell surface via flow cytometry. In addition to CD56, NK cells can be further characterized into a variety of subsets through many different cell surface markers including CD16 and the KIR family of receptors. Check out these tools that can be used for NK cell analysis including flow cytometry, functional assays, and immunoassays. For additional characterization information, visit our NK cell overview page
For the most advanced cell analysis, the Attune CytPix Flow Cytometer combines acoustic focusing fluidics for high sensitivity and high throughput with a high-speed camera.
Cryopreservation in NK cell therapy
NK cell therapies typically require a cryopreservation step to help ensure that the cellular product is preserved without compromising viability and efficacy. Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) offer precise, repeatable freezing results that protect precious samples from intracellular freezing and are equipped with enhanced data traceability features to support audit requirements and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.
NK cell culture media and reagents webinars
Video Player is loading.
Current Time 0:00
Duration 1:01:44
Loaded: 0%
Stream Type LIVE
Remaining Time 1:01:44
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    Video: Serum-free expansion of NK cells with K562-based feeder systems for clinical applications
    Dr. Dean Lee of Nationwide Children’s Hospital discusses his work with NK cells, including optimization of culture conditions.
    Video Player is loading.
    Current Time 0:00
    Duration 29:08
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 29:08
    1x
      • Chapters
      • descriptions off, selected
      • subtitles off, selected
      • en (Main), selected
      Video: Feeder-free expansion and closed wash and concentration of a clinically relevant number of human NK cells
      Dr. Erica Heipertz introduces CTS NK-Xpander Medium in a feeder-free system.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
        • Chapters
        • descriptions off, selected
        • subtitles off, selected
          Video: Large scale expansion and processing of clinically relevant numbers of functional human NK cells
          Dr. Erica Heipertz discusses scale-up of human NK cell populations in various bioreactor systems.
          References
          Cell therapy resources
          Support