Conceptual art of three different bottles of FBS overlayed on a cell
In general, FBS is a mixture of a wide range of components, including growth factors, hormones, lipids, and many others. However, certain experimental applications require more control over serum composition. Examples of cell culture applications requiring specialty serum include those involving tet-inducible gene expression systems, those requiring very specific concentrations of certain molecules, or those involving the use of stem cells.
Choose from a large selection of serum products designed for specific experimental needs and sensitive cell culture, including stem cell research, cancer research, reporter assays, immunoassays, and more.
Selection guide: Specialty FBS
FBS typeIcon showing an arrow pointing out to the right from a circle
Removes unwanted components
Stopwatch icon
Speeds up your workflow
Speaker icon
Reduces background noise
Cell icon
Proven to work with demanding cells
Clipboard with checkmark icon
Enables more control in your research
Icon showing two arrows around a circle
Delivers consistency
Dialyzed FBS
Charcoal-stripped FBS
Tet-system approved FBS 
MaxSpec FBS  
Ultra-low IgG FBS
Exosome-depleted FBS
MSC-qualified FBS  
ESC-qualified FBS  
Choose specialty FBS to control your experimental conditions
MaxSpec FBS
Artistic rendering of multiple eukaryotic cells
Gibco MaxSpec FBS is Thermo Fisher Scientific’s highest quality FBS, delivering our best testing levels and quality specifications.
Features of MaxSpec FBS
  • Most characterized Gibco sera, with the lowest levels of endotoxin and hemoglobin
  • Endotoxin level: ≤1 EU/mL
  • Hemoglobin level: ≤15 mg/dL
  • Meets USP/EP guidelines with up to 76 quality specification tests
  • Lowest BSE risk and lower viral risk than other Gibco FBS
Order MaxSpec FBS
N.° de Catálogo Nombre Tamaño Precio (USD) Cantidad
A4766801 Fetal Bovine Serum, MaxSpec Each -
Ultra-low IgG FBS
Artistic rendering of multiple red viruses
Ultra-low IgG fetal bovine serum has been chromatographically depleted of IgG, an important factor in isolating organisms that may be inhibited by pre-existing levels of IgG or for quantitative IgG immunoassays. Despite IgG depletion, the serum retains full biological activity.
Features of ultra-low IgG FBS
  • Lowers Ig levels (<5 μg/ml) in fetal bovine serum while preserving comparable performance to regular FBS
  • Low or undetectable BVD antibody titer levels
  • Available in the innovatively designed, aliquot-free Gibco One Shot FBS 50 mL bottle
Research areas using ultra-low IgG FBS*
  • Antibodies
  • Viruses and viral response
  • Cell-surface epitopes
  • IgG-dependent cell cytotoxicity
  • Ligand-receptor studies
* These results are based on a review of approximately 10,000 publications using query terms based on six specialty FBS products offered by Thermo Fisher Scientific. These terms were generated by the MeSH (medical subject headings) taxonomy based on the full text of the paper.
Order ultra-low IgG FBS
Publications using Gibco Ultra-Low IgG FBS
Exosome-depleted FBS (exosome-free FBS)
Artistic rendering showing multiple cells and exosomes
Exosome-depleted FBS, also known as exosome-free FBS, is an ultrapure FBS that provides high levels of exosome depletion (≥90%) while maintaining cell culture performance. This specialty FBS is relied upon for exosome isolation assays and research involving exosome contents, such as miRNAs. By choosing exosome-depleted FBS, the inefficiencies that come with ultracentrifugation of serum are removed.
Features of exosome-depleted FBS (exosome-free FBS)
  • ≥90% of exosomes depleted
  • Sophisticated manufacturing process retains the nutrients your cells need
  • Avoids need for time-consuming ultracentrifugation
  • Cell culture testing of every lot
  • Specially developed for exosome research
  • Full quality testing for sterility, mycoplasmas, performance, and endotoxins
  • Available in aliquot-free Gibco One Shot FBS 50 mL bottle
Research areas using exosome-depleted FBS*
  • Exosomes and extracellular vesicles
  • MicroRNA
  • Cell-cell communication
* These results are based on a review of approximately 10,000 publications using query terms based on six specialty FBS products offered by Thermo Fisher Scientific. These terms were generated by the MeSH (medical subject headings) taxonomy based on the full text of the paper.
Why researchers choose exosome-depleted FBS
Hear how Gibco Exosome-Depleted FBS helps save time, reduce batch-to-batch variation, and support excellent growth of cells.
Video Player is loading.
Current Time 0:00
Duration 3:30
Loaded: 0%
Stream Type LIVE
Remaining Time 3:30
1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
    • en (Main), selected
    “I was able to culture my cells and get very good growth in only 2.5% serum, compared to 10%, due to Gibco Exosome-Depleted FBS, maintaining a lot of the important nutrients ("the good things") that were lost during ultracentrifugation.”
    Dr. Jonathan Gilthorpe, Pharmacology and Clinical Neuroscience University, Sweden.
    Exosome-depleted FBS supporting data
    72-hour cell growth of rat oligodendrocyte cells with exosome-depleted FBS
    Figure 1. Growth rate of rat oligodendrocyte cells in exosome-depleted FBS. Rat oligodendrocyte cells were seeded at 1,000 cells per well in 96-well plates and grown in medium containing 2% or 10% by volume of one of the following supplements: Gibco Exosome-Depleted FBS, Gibco FBS, the source of the exosome-depleted version), or ultracentrifuged FBS. After 72 hours in culture, the live cells were stained with Invitrogen Hoechst 33342, and the plate was imaged and analyzed on a 96-well plate imaging instrument (Trophos Plate RUNNER HD). Results are presented as the total cell count as reported by the 96-well plate analysis. (The results were obtained from the laboratory of Dr. Jonathan Gilthorpe in the department of Pharmacology and Clinical Neuroscience at Umeå University, Sweden.)
    Cell culture performance: 2-day viability and viable cell density as a percentage of source FBS
    Bar graph showing viability and viable cell density for different cell lines grown in exosome-depleted FBS. Viability and viable cell density results for each cell line were similar in both source (standard) FBS and exosome-depleted FBS
     Click image to enlarge
    Figure 2. Viability and viable cell density of cells grown in exosome-depleted FBS. Several cell lines were grown in basal medium (DMEM, high glucose, GlutaMAX Supplement) containing 10% exosome-depleted FBS or 10% source FBS and assayed for viable cell density (VCD) and cell viability by Vi-CELL instrument analysis. Results are presented as the viability or viable cell density that was achieved in exosome-depleted FBS as a percentage of that achieved in the source FBS (used to make the exosome-depleted FBS).
    Comparison of exosome depletion methods for FBS
    Bar graph comparing exosome depletion methods
     Click image to enlarge
    Figure 3. Comparison of exosome depletion methods for FBS. Three different lots of FBS were subjected to 3 different methods of exosome depletion. The first used ultracentrifugation at 110,000 X g for 3 hours (labeled as Ultracentrifuge 1). The second method used ultracentrifugation at 150,000 X g for 18 hours (labeled as Ultracentrifuge 2). The third method used our proprietary process for the depletion of exosomes from FBS (labeled as exosome-depleted FBS). Source FBS refers to the FBS prior to any exosome depletion process. After the exosome depletions, relative levels of remaining exosomes were measured by extracting exosomes from depleted sera using the Total Exosome Isolation Reagent (from serum), then staining the isolated exosomes with a lipophilic dye. The fluorescent signal is displayed as Relative Fluorescence Units (RFU). The data demonstrate the improved efficiency of exosome depletion using our proprietary method compared to ultracentrifugation.
    Exosome depletion of FBS verified by NanoSight instrument measurement and fluorescence exosomes staining assay
    Bar graph comparing exosome depletion in source FBS, ultracentrifuged FBS, and exosome-depleted FBS. Significantly lower fluorescent signals in exosome-depleted FBS indicate an improvement in exosome depletion compared to ultracentrifuged and source FBS
     Click image to enlarge
    Figure 4. Verification of FBS exosome depletion. The exosome depletion from FBS samples was verified by analysis on a NanoSight instrument (comparing the 30–150 nm count before and after exosome depletion) as well as via fluorescence-based assay. Briefly, this assay involves extracting exosomes from serum using the Total Exosome Isolation Reagent, and then staining the isolated exosomes with Invitrogen BODIPY TR Ceramide. The percent depletion is derived from comparing the fluorescent signal of the exosome-depleted FBS with the source FBS. The first two exosome-depleted lots shown were produced by our proprietary manufacturing method. Included in the same analysis was a sample of FBS that was ultracentrifuged to deplete exosomes and an exosome-depleted FBS product from a competitor.
    Analysis of exosome-depleted FBS by SDS-PAGE and CD63 content by western blot
    Figure 5. Western blot analysis of exosome-depleted FBS. (A) Analysis of FBS by SDS-PAGE: Equal masses (10 µg) of FBS proteins were analyzed by SDS-PAGE. Exosome-depleted FBS looks very similar to the source FBS used in production. (B) CD63 content by western blot analysis: Exosomes were precipitated from FBS samples, run by SDS-PAGE, and probed by western blotting for CD63. Compared to all other FBS samples tested, exosome-depleted FBS shows no observable CD63, indicating excellent exosome depletion. MW = molecular weight marker.
    miRNA detection by qRT-PCR in FBS sample-derived exosomes
    Figure 6. miRNA detection in FBS sample-derived exosomes. Exosomes were isolated from FBS samples, and total RNA was isolated from the exosomes (using the Total Exosome RNA & Protein Isolation Kit). RNA was then analyzed by qRT-PCR for Let7e, miR16, miR21, miR23a, miR24, and miR122 using the Applied Biosystems TaqMan MicroRNA Reverse Transcription Kit.
    In addition to exosome-depleted FBS, explore additional exosome products
    Order exosome-depleted FBS
    Publications using Gibco exosome-depleted FBS
    MSC-qualified FBS
    Microscopic stained mesenchymal stem cells (MSCs)
    MSC-qualified FBS is specially tested to support the expansion and clonal enumeration (by colony forming unit-fibroblast assay) of mesenchymal stem cells (MSCs). MSC-qualified FBS has been carefully designed to eliminate the need for testing multiple FBS lots to identify the optimal one for MSC research.
    Features of MSC-qualified FBS
    • Performance-tested using standard 14-day MSC CFU-F assay
    • Each lot is tested against an in-house FBS reference standard using cells from a master cell bank of MSCs from normal bone marrow donors, which helps ensure lot-to-lot consistency
    Research areas using MSC-qualified FBS*
    • Mesenchymal stem cells
    • Mesenchymal stromal cells
    • Osteogenesis
    • Chondrogenesis and cartilage
    • Collagen and other extracellular matrices (ECM)
    • Adipose tissue and adipogenesis
    * These results are based on a review of approximately 10,000 publications using query terms based on six specialty FBS products offered by Thermo Fisher Scientific. These terms were generated by the MeSH (medical subject headings) taxonomy based on the full text of the paper.
    MSC-qualified FBS supporting data
    Effect of MSC-qualified FBS on MSC clonal efficiency and expansion
    Bar chart showing higher MSC clonal efficiency for Gibco MSC-qualified FBS than for competitor FBS
    Figure 7. Effect of FBS source on MSC clonal efficiency. Results from Gibco FBS (2 lots tested are indicated by red and blue bars) are shown along with the results from a competitor's FBS product (yellow bar). P<0.05 via student’s t-test.
    Bar chart showing higher MSC expansion (cells/mL) for Gibco MSC-qualified FBS than for competitor FBS
    Figure 8. Effect of FBS source on MSC expansion. Results from Gibco FBS (2 lots tested are indicated by red and blue bars) are shown along with the results from a competitor's FBS product (yellow bar). P<0.05 via student’s t-test.
    Order MSC-qualified FBS
    Publications using Gibco MSC-qualified FBS
    ESC-qualified FBS
    Artistic rendering showing multiple embryonic stem cells
    ESC-qualified FBS, produced using an industry-leading qualification assay, is specifically designed for achieving high plating efficiencies and maintaining pluripotency of embryonic stem cells (ES cells). ES cells are derived from the inner cell mass of blastocysts and are used in a variety of studies regarding biological functioning. When culturing ES cells, it is critical that they maintain the ability to differentiate into the cell type of interest in order to test expression and other characteristics. Therefore, ESC-qualified FBS helps maintain proper growth conditions for ES cells.
    Features of ES cell FBS
    • Tested for the ability to sustain undifferentiated ES cells (Figure 9A) while maintaining karyotype integrity (Figure 11), LIF responsiveness (Figure 12), and pluripotency markers (Figure 9B)
    • Improved screening with germ line-competent PRX129/X1 mESC line using a predictive assay that measures plating efficiency and pluripotency maintenance
    • High consistency between lots with proven applications in iPSC generation and PSC culture
    • Available in the innovatively designed, aliquot-free Gibco One Shot FBS 50 mL bottle
    Research areas using ES cell FBS*
    • Hormones or hormone receptors (androgens, estrogens, progesterone)
    • Induced pluripotent stem cells (iPSCs)
    • Cellular reprogramming
    • Embryonic stem cells (ESCs)
    • Embryonic development
    Using ES cells to study disease, cancer, or cellular mutation can help pave the way for new treatment options and potential biological advancements.
    * These results are based on a review of approximately 10,000 publications using query terms based on six specialty FBS products offered by Thermo Fisher Scientific. These terms were generated by the MeSH (medical subject headings) taxonomy based on the full text of the paper.
    ES cell FBS supporting data
    mESCs grown in ES cell FBS exhibit pluripotency traits and maintain normal karyotypes
    Phase-contrast micrograph images showing mESC colonies grown using ES-qualified FBS; morphological traits shown in panel A and fluorescent AP staining shown in the panel B indicate that the ES cells are pluripotent
    Figure 9. Image analysis of mESC colonies grown in media containing Gibco ES Cell FBS Qualified. (A) Phase-contrast image of mESC colonies grown in media containing ES cell FBS showing colonies with morphological traits consistent with pluripotency. (B) An overlay of the fluorescence image (obtained after staining the sample with Alkaline Phosphatase (AP) Live Stain) with the phase-contrast image of mESC colonies, showing uniform AP staining. Colonies that appear green are expressing AP and are therefore pluripotent.
    Phase-contrast and fluorescent micrograph images showing mESC colonies grown using ES-qualified FBS. Morphological traits, AP staining, and Oct4 and Sox2 expression indicate that these cells are pluripotent.
    Figure 10. Phase contrast and fluorescent images of mESC colonies. mESCs cultured in media supplemented with ESC-qualified FBS exhibit the morphological traits of well-defined refractive edges, tight colonies with small cells, and few differentiated cells. These cells also exhibit pluripotency via Alkaline Phosphatase (AP) Live Stain and the expression of Oct4 and Sox2.
    Figure 11. mESCs grown in media supplemented with ESC-qualified FBS exhibit normal karyotypes. Normal karyotype report for PRX129/X1 mESC line. 18 of 20 cells tested exhibited a normal karyotype (Cell Line Genetics).
    Graph showing that mESCs grown with LIF in ES-qualified FBS maintain higher levels of pluripotency whereas those grown in the absence of LIF differentiate
    Figure 12. mESCs cultured in media supplemented with ESC-qualified FBS maintain LIF responsiveness. When LIF is removed from mESC media, normal mESC cultures respond with widespread differentiation. PRX129/X1 murine ES cells grown with and without LIF demonstrate this response.
    ESC-qualified FBS can support reprogramming workflows
    Timeline and images showing that proper supplementation of growth media with ES-qualified FBS supports workflows to generate iPSCs from mouse and human fibroblasts
    Figure 13. When used to supplement growth media, ESC-qualified FBS helps support generation of iPSCs from both mouse and human fibroblasts. In a standard workflow using Sendai virus–based reprogramming (CytoTune iPS 2.0 Sendai Reprogramming Kit), both murine and human fibroblasts successfully generated iPSCs.
    Rigorous screening methodology helps ensure performance of ESC-qualified FBS
    Figure 14. Plating efficiency and colony morphology assessed in murine embryonic stem cells grown in media containing ESC-qualified FBS. (A) Undifferentiated colonies (top row) appear bright and sharp, and differentiated colonies (bottom row) appear dim, fuzzy and flattened when stained with alkaline phosphatase. (B) Methylene blue stains all cells and is used to count the total colony number. (C) Results plotted as % AP+/MB+ demonstrates assay consistency among three users.
    Order ESC-qualified FBS
    Publications using Gibco ES Cell FBS Qualified
    Resources
    Fetal Bovine Serum Basics
    Learn the basics of FBS for cell culture, including information on the FBS uses, components, and the market dynamics driving this industry.
    Cell Culture Basics
    Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
    Gibco iMatch Sera Lot Matching Tool 
    Provide your previous Gibco FBS lot number or answer a few questions to find your ideal serum match.
    Related products
    Support
    Find your regional sales representative
    Connect with a regional FBS sales representative.
    Cell Culture & Transfection Support Center
    Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
    Need technical support?
    Contact our expert team for technical and application support of laboratory products.
    For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.
    Stylesheet for Classic Wide Template adjustments