Cell isolation helps researchers characterize specific, pure cell populations. Cell depletion indirectly helps this characterization by depleting the cells that are not of interest. Cell isolation, also commonly referred to as cell separation or cell sorting, can be achieved using magnetic beads to target cells for selection or depletion by using ligands or antibodies directed against specific cell surface antigens. Labeled cells that are cross-linked to magnetic beads are immobilized using magnets or magnetic racks. This method is simpler and faster compared to other commonly used cell isolation methods to isolate highly purified populations of specific cell subsets.
Magnetic bead cell separation methods offer the capability to perform positive and negative cell selection, as well as cell depletion. For positive selection, the magnetic-bound cells of interest are immobilized until removed from the magnet, while the supernatant is discarded. For negative selection, the cells of interest are in the supernatant. For cell depletion, cell-specific magnetic beads are used to remove specific unwanted cell types from a mixed cell sample with >99% efficiency.
Read more about positive and negative selection and depletion, as well as view our variety of kits and products to help improve your cell isolation protocols.
Cell depletion with Dynabeads magnetic beads
Cell depletion is aided by cell separation steps using magnetic beads. Cell depletion using magnetic beads involves selectively removing specific unwanted cells from a mixed population by using magnetic beads coupled with antibodies or ligands targeting specific cell surface markers. Through exposure to a magnetic field, the bead-bound unwanted cells are separated and removed from sample, allowing the isolation of desired cell populations for downstream characterization and analysis.
Human cell depletion kits
Human icon Cell depletion kits are available to target specific human cell surface markers to deplete certain cell populations, which can be found in the table below.
Cell type to depleteCell surface marker(s) to target for depletion
T cells and natural killer (NK) cellsCD2
Mature T cellsCD3
T helper cellsCD4
Cytotoxic T cellsCD8
MonocytesCD14
Neutrophil and eosinophil granulocytesCD15
Pan B cellsCD19
Regulatory T cellsCD25
Endothelial cellsCD31
LeukocytesCD45
Epithelial cellsEPCAM
Cell depletion or positive isolation automation scripts
Automated protocol descriptionSupporting productsDownload automation scripts
Cell depletion or positive cell isolation for downstream nucleic acid, protein analysis, or cell depletion             
Catalog # Name Size Price (USD) Qty
11159D Dynabeads™ CD2 Each
1,201.65
Online Exclusive
1,336.00
Save 134.35 (10%)
11151D Dynabeads™ CD3, 5 mL Each
1,336.00
11145D Dynabeads™ CD4, 5 mL Each
1,267.65
Online Exclusive
1,336.00
Save 68.35 (5%)
11147D Dynabeads™ CD8, 5 mL Each
1,270.65
Online Exclusive
1,336.00
Save 65.35 (5%)
11149D Dynabeads™ CD14, 5 mL Each
1,262.65
Online Exclusive
1,336.00
Save 73.35 (5%)
11137D Dynabeads™ CD15, 5 mL Each
1,226.65
Online Exclusive
1,336.00
Save 109.35 (8%)
11143D Dynabeads™ CD19 Pan B, 5 mL Each
1,211.65
Online Exclusive
1,336.00
Save 124.35 (9%)
11157D Dynabeads™ CD25 Each
1,336.00
11155D Dynabeads™ CD31 Endothelial Cell Each
1,416.65
Online Exclusive
1,460.00
Save 43.35 (3%)
11153D Dynabeads™ CD45 Each
1,336.00
16102 Dynabeads™ Epithelial Enrich, 5 mL Each
1,898.00
11170D Dynabeads™ MyOne™ CD45 Leukocyte Depletion, 2 mL Each
758.00
11152D Dynabeads™ CD3, 2 mL Each
656.00
11148D Dynabeads™ CD8, 2 mL Each
623.65
Online Exclusive
656.00
Save 32.35 (5%)
11146D Dynabeads™ CD4, 2 mL Each
622.65
Online Exclusive
656.00
Save 33.35 (5%)
11144D Dynabeads™ CD19 Pan B, 2 mL Each
594.65
Online Exclusive
656.00
Save 61.35 (9%)
11150D Dynabeads™ CD14, 2 mL Each
619.65
Online Exclusive
656.00
Save 36.35 (6%)
11138D Dynabeads™ CD15, 2 mL Each
602.65
Online Exclusive
656.00
Save 53.35 (8%)
16103D Dynabeads™ Epithelial Enrich, 2 mL Each
1,060.00
Mouse cell depletion kits
Mouse icon Cell depletion kits are available to target specific mouse cell surface markers to deplete certain cell populations, which can be found in the table below.
Cell type to depleteCell surface marker(s) to target for depletion
CD4+ T cellsCD4
CD8+ T cellsCD8
Pan B cellsB220
Pan T cellsThy1.2
Catalog # Name Size Price (USD) Qty
11445D Dynabeads™ Mouse CD4 Each
1,232.00
11447D Dynabeads™ Mouse CD8 (Lyt 2) Each
1,147.65
Online Exclusive
1,232.00
Save 84.35 (7%)
11441D Dynabeads™ Mouse Pan B (B220) Each
1,232.00
11443D Dynabeads™ Mouse Pan T (Thy1.2) Each
1,146.65
Online Exclusive
1,232.00
Save 85.35 (7%)
Build-your-own negative cell depletion kit
Build your own kit and attach your own antibody for custom cell targeting.
Target cell speciesRecommended antibodySuggested product
Any species (except mouse)Mouse IgGDynabeads Pan Mouse IgG
Mouse IgG1, IgG2a, and IgG2bDynabeads Sheep Anti-Mouse IgG
Mouse IgGDynabeads Goat Anti-Mouse IgG
Mouse IgMDynabeads Rat Anti-Mouse IgM
Any species (except rat)Rat IgGDynabeads Sheep Anti-Rat IgG
Any species (except rabbit)Rabbit IgGDynabeads M-280 Sheep Anti–Rabbit IgG
Any speciesBiotinylated antibodyDynabeads Biotin Binder
Dynabeads MyOne Streptavidin T1
Any purified antibody/ligandDynabeads M-450 Epoxy
Dynabeads M-450 Tosylactivated
Dynabeads Antibody Coupling Kit*
* The Dynabeads Antibody Coupling Kits contain buffers for optimal coupling conditions.
Catalog # Name Size Price (USD) Qty
11035 Dynabeads™ Sheep Anti-Rat IgG, 5 mL Each
876.00
11041 Dynabeads™ Pan Mouse IgG, 5 mL Each
793.65
Online Exclusive
820.00
Save 26.35 (3%)
11047 Dynabeads™ Biotin Binder, 5 mL Each
856.00
11204D Dynabeads™ M-280 Sheep Anti-Rabbit IgG, 10 mL Each
1,680.00
11201D Dynabeads™ M-280 Sheep Anti-Mouse IgG, 2 mL Each
540.00
11048D Dynabeads™ Biotin Binder, 2 mL Each
409.00
11036D Dynabeads™ Sheep anti-Rat IgG, 2 mL Each
409.00
11040D Dynabeads™ Pan Mouse IgG, 2 mL Each
395.65
Online Exclusive
409.00
Save 13.35 (3%)
11031 Dynabeads™ Sheep-Anti Mouse IgG Each
878.65
Online Exclusive
906.00
Save 27.35 (3%)
11033 Dynabeads™ Goat Anti-Mouse IgG Each
906.00
11039D Dynabeads™ Rat Anti-Mouse IgM Each
858.00
11203D Dynabeads™ M-280 Sheep Anti-Rabbit IgG, 2 mL Each
540.00
65601 Dynabeads™ MyOne™ Streptavidin T1, 2 mL Each
596.70
Special offer
Online exclusive
Ends: 01-Jun-2025
702.00
Save 105.30 (15%)
65602 Dynabeads™ MyOne™ Streptavidin T1, 10 mL Each
1,976.25
Special offer
Online exclusive
Ends: 01-Jun-2025
2,325.00
Save 348.75 (15%)
65604D Dynabeads™ MyOne™ Streptavidin T1, 50 mL Each
8,092.00
Special offer
Online exclusive
Ends: 01-Jun-2025
9,520.00
Save 1,428.00 (15%)
14011 Dynabeads™ M-450 Epoxy Each
960.00
14013 Dynabeads™ M-450 Tosylactivated Each
1,082.00
14311D Dynabeads™ Antibody Coupling Kit Each
635.65
Online Exclusive
666.00
Save 30.35 (5%)
Positive cell isolation
Positive cell isolation involves directly labeling desired cells for selection using an antibody or ligand that targets a specific cell surface protein.
Positive cell isolation with Dynabeads magnetic beads
In positive cell isolation using Dynabeads magnetic beads, the beads are coated with the specific antibody or ligand to bind to the target cells and the bead/cell complexes are pulled down using a magnet. The supernatant contains non-specific cells and debris which can be discarded, leaving only your pure target cells attached to the bead.
 Cell separation using magnetic beads:
  • Is a gentle, tube-based method
  • Isolates high yields of pure, viable, and functional cells
  • Preserves the cells’ native characteristics
Human positive cell isolation kits
Human icon 
Positive cell isolation kits available for human cells target the following cell surface markers:
Cell surface marker to target for isolationCell type targetedFormat(s) available
CD3Cytotoxic T cells and T helper cellsFlowComp kit
CD4T helper cells, regulatory T cells, and natural killer T cellsFlowComp kit
CD8Cytotoxic T cells FlowComp kit
DETACHaBEAD kit
CD14CD14+ monocytesFlowComp kit
CD19Pan B cellsDETACHaBEAD kit
CD34Progenitor stem cellsDETACHaBEAD kit
CD4/CD25Regulatory CD4+ T cellsDETACHaBEAD kit
EPCAMEpithelial cellsCELLection kit
Learn more about flow compatible Dynabeads kits
Positive cell isolation automation scripts
Automated protocol descriptionSupporting productsDownload automation scripts
Positive cell isolation for downstream cellular applications, such as T cell activation
Catalog # Name Size Price (USD) Qty
11365D Dynabeads™ FlowComp™ Human CD3 Kit Each
1,319.65
Online Exclusive
1,484.00
Save 164.35 (11%)
11361D Dynabeads™ FlowComp™ Human CD4 Kit Each
1,484.00
11362D Dynabeads™ FlowComp™ Human CD8 Kit Each
1,484.00
11333D Dynabeads™ CD8 Positive Isolation Kit Each
1,600.00
11367D Dynabeads™ FlowComp™ Human CD14 Kit Each
1,484.00
12506D DETACHaBEAD™ CD19 Kit Each
770.00
11301D Dynabeads™ CD34 Positive Isolation Kit Each
1,516.00
11363D Dynabeads™ Regulatory CD4+/CD25+ T Cell Kit Each
1,180.65
Online Exclusive
1,252.00
Save 71.35 (6%)
16203 CELLection™ Epithelial Enrich Dynabeads™ Each
1,972.00
Mouse positive cell isolation kits
Mouse icon 
Positive cell isolation kits available for mouse cells target the following cell surface markers:
Cell surface marker to target for isolationCell type targetedFormat(s) available
CD4Naive CD4+ T cellsFlowComp kit
CD8Naive CD8+ T cellsFlowComp kit
CD4/CD25Regulatory CD4+ T cellsFlowComp kit
CD90.2Pan T cellsFlowComp kit
Pan IgGDepends on your antibodyCELLection kit
Dynabeads FlowComp positive cell isolation kits for your flow cytometry cell sorting experiments
In contrast to column-based systems, FlowComp technology lets you isolate bead-free cells directly from human whole blood, buffy coat, or from mouse/human mononuclear cells (MNCs). Following separation, the beads are immediately released and removed, helping ensure the least possible interaction with your cells. Isolated bead-free cells are then ready for use in any downstream application, including flow cytometry. Get high cell recovery, viability, and purity using FlowComp kits and markers.
Explore Dynabeads FlowComp kits and markers
Catalog # Name Size Price (USD) Qty
11461D Dynabeads™ FlowComp™ Mouse CD4 Kit Each
940.00
11462D Dynabeads™ FlowComp™ Mouse CD8 Kit Each
861.65
Online Exclusive
940.00
Save 78.35 (8%)
11465D Dynabeads™ FlowComp™ Mouse Pan T (CD90.2) Kit Each
868.65
Online Exclusive
940.00
Save 71.35 (8%)
11531D CELLection™ Pan Mouse IgG Kit Each
988.00
Build-your-own positive cell isolation kit
Additional positive cell isolation kits are available to use for a wide range of species, with any antibody of your choice.
Use withFormat available
Any antibody targeting any cell type in any speciesFlowComp kit
Any biotinylated antibody targeting any cell type in any speciesCELLection kit
Catalog # Name Size Price (USD) Qty
11061D Dynabeads™ FlowComp™ Flexi Kit Each
976.65
Online Exclusive
1,022.00
Save 45.35 (4%)
11533D CELLection™ Biotin Binder Kit Each
988.00
Negative cell isolation
Negative cell involves labeling unwanted cell types for removal. The unwanted cell types can be bound using magnetic beads coupled to antibodies or ligands that target specific cell surface proteins. The bound, unwanted cells can be depleted from the final isolated fraction by incubating the sample in a magnetic field, leaving desired unbound cells free floating in solution.
Negative cell isolation with Dynabeads magnetic beads
In negative cell isolation, Dynabeads magnetic beads are used to deplete all the unwanted cells using the supplied antibody mix. The target cells are left in the sample untouched and free of beads. The isolated target cells can be directly analyzed in a flow cytometer and used in any functional assay.
Negative cell separation using magnetic beads:
  • Is a gentle, tube-based method
  • Isolates high yields of pure, viable, and functional cells
  • Preserves the cells’ native characteristics
Human negative cell isolation kits
Human icon 
Negative cell isolation kits are available for human cells to target the following cell types:
Enriched untouched cell typeSurface markers on depleted cellsFormat available
CD4+ T cells: T helper cells, regulatory T cells, and natural killer T cellsCD8, CD14, CD16 (a and b), CD19, CD36, CD56, CDw123 and CD235aUntouched kit
CD8+ T cells: cytotoxic T cellsCD4, CD14, CD16 (a and b), CD19, CD36, CD56, CDw123 and CD235aUntouched kit
Dendritic cells (DCs)CD3, CD14, CD16, CD19, CD56, and CD235aUntouched kit
MonocytesCD3, CD7, CD16 (a and b), CD19, CD56, CD123, and CD235aUntouched kit
Pan B cellsCD2, CD14, CD16 (a and b), CD36, CD43, and CD235aUntouched kit
Pan T cellsCD14, CD16 (a and b), CD19, CD36, CD56, CD123, and CD235aUntouched kit
Natural killer (NK) cellsCD3, CD14, CD36, HLA Class II, CD123, and CD235aUntouched kit
Mouse negative cell isolation kits
Mouse icon 
Negative cell isolation kits are available for mouse cells to target the following cell types:
Enriched untouched cell typeSurface markers on depleted cellsFormat available
Naive CD4+ T cellsCD8, CD45R (B220), CD11b (Mac1), Ter-119, and CD16/CD32Untouched kit
Naive CD8+ T cellsCD4, CD45R (B220), CD11b (Mac1), Ter-119, and CD16/CD32Untouched kit
Dendritic cells (DCs)CD2, CD3ε, CD49b, mIgM, and Ter-119Dynabeads kit
Pan B cellsCD43Dynabeads kit
Pan T cellsCD11b, CD16/32, CD45R, and Ter-119Untouched kit
Catalog # Name Size Price (USD) Qty
11415D Dynabeads™ Untouched™ Mouse CD4 Cells Kit, 2 x 10 mL Each
805.65
Online Exclusive
888.00
Save 82.35 (9%)
11417D Dynabeads™ Untouched™ Mouse CD8 Cells Kit Each
805.65
Online Exclusive
888.00
Save 82.35 (9%)
11429D Dynabeads™ Mouse DC (Dendritic Cell) Enrichment Each
1,252.00
11422D Dynabeads™ Mouse CD43 (Untouched™ B Cells) Each
851.65
Online Exclusive
918.00
Save 66.35 (7%)
11413D Dynabeads™ Untouched™ Mouse T Cells Kit Each
809.65
Online Exclusive
888.00
Save 78.35 (9%)
Cell depletion with Dynabeads magnetic beads
Cell depletion is aided by cell separation steps using magnetic beads. Cell depletion using magnetic beads involves selectively removing specific unwanted cells from a mixed population by using magnetic beads coupled with antibodies or ligands targeting specific cell surface markers. Through exposure to a magnetic field, the bead-bound unwanted cells are separated and removed from sample, allowing the isolation of desired cell populations for downstream characterization and analysis.
Human cell depletion kits
Human icon Cell depletion kits are available to target specific human cell surface markers to deplete certain cell populations, which can be found in the table below.
Cell type to depleteCell surface marker(s) to target for depletion
T cells and natural killer (NK) cellsCD2
Mature T cellsCD3
T helper cellsCD4
Cytotoxic T cellsCD8
MonocytesCD14
Neutrophil and eosinophil granulocytesCD15
Pan B cellsCD19
Regulatory T cellsCD25
Endothelial cellsCD31
LeukocytesCD45
Epithelial cellsEPCAM
Cell depletion or positive isolation automation scripts
Automated protocol descriptionSupporting productsDownload automation scripts
Cell depletion or positive cell isolation for downstream nucleic acid, protein analysis, or cell depletion             
Catalog # Name Size Price (USD) Qty
11159D Dynabeads™ CD2 Each
1,201.65
Online Exclusive
1,336.00
Save 134.35 (10%)
11151D Dynabeads™ CD3, 5 mL Each
1,336.00
11145D Dynabeads™ CD4, 5 mL Each
1,267.65
Online Exclusive
1,336.00
Save 68.35 (5%)
11147D Dynabeads™ CD8, 5 mL Each
1,270.65
Online Exclusive
1,336.00
Save 65.35 (5%)
11149D Dynabeads™ CD14, 5 mL Each
1,262.65
Online Exclusive
1,336.00
Save 73.35 (5%)
11137D Dynabeads™ CD15, 5 mL Each
1,226.65
Online Exclusive
1,336.00
Save 109.35 (8%)
11143D Dynabeads™ CD19 Pan B, 5 mL Each
1,211.65
Online Exclusive
1,336.00
Save 124.35 (9%)
11157D Dynabeads™ CD25 Each
1,336.00
11155D Dynabeads™ CD31 Endothelial Cell Each
1,416.65
Online Exclusive
1,460.00
Save 43.35 (3%)
11153D Dynabeads™ CD45 Each
1,336.00
16102 Dynabeads™ Epithelial Enrich, 5 mL Each
1,898.00
11170D Dynabeads™ MyOne™ CD45 Leukocyte Depletion, 2 mL Each
758.00
11152D Dynabeads™ CD3, 2 mL Each
656.00
11148D Dynabeads™ CD8, 2 mL Each
623.65
Online Exclusive
656.00
Save 32.35 (5%)
11146D Dynabeads™ CD4, 2 mL Each
622.65
Online Exclusive
656.00
Save 33.35 (5%)
11144D Dynabeads™ CD19 Pan B, 2 mL Each
594.65
Online Exclusive
656.00
Save 61.35 (9%)
11150D Dynabeads™ CD14, 2 mL Each
619.65
Online Exclusive
656.00
Save 36.35 (6%)
11138D Dynabeads™ CD15, 2 mL Each
602.65
Online Exclusive
656.00
Save 53.35 (8%)
16103D Dynabeads™ Epithelial Enrich, 2 mL Each
1,060.00
Mouse cell depletion kits
Mouse icon Cell depletion kits are available to target specific mouse cell surface markers to deplete certain cell populations, which can be found in the table below.
Cell type to depleteCell surface marker(s) to target for depletion
CD4+ T cellsCD4
CD8+ T cellsCD8
Pan B cellsB220
Pan T cellsThy1.2
Catalog # Name Size Price (USD) Qty
11445D Dynabeads™ Mouse CD4 Each
1,232.00
11447D Dynabeads™ Mouse CD8 (Lyt 2) Each
1,147.65
Online Exclusive
1,232.00
Save 84.35 (7%)
11441D Dynabeads™ Mouse Pan B (B220) Each
1,232.00
11443D Dynabeads™ Mouse Pan T (Thy1.2) Each
1,146.65
Online Exclusive
1,232.00
Save 85.35 (7%)
Build-your-own negative cell depletion kit
Build your own kit and attach your own antibody for custom cell targeting.
Target cell speciesRecommended antibodySuggested product
Any species (except mouse)Mouse IgGDynabeads Pan Mouse IgG
Mouse IgG1, IgG2a, and IgG2bDynabeads Sheep Anti-Mouse IgG
Mouse IgGDynabeads Goat Anti-Mouse IgG
Mouse IgMDynabeads Rat Anti-Mouse IgM
Any species (except rat)Rat IgGDynabeads Sheep Anti-Rat IgG
Any species (except rabbit)Rabbit IgGDynabeads M-280 Sheep Anti–Rabbit IgG
Any speciesBiotinylated antibodyDynabeads Biotin Binder
Dynabeads MyOne Streptavidin T1
Any purified antibody/ligandDynabeads M-450 Epoxy
Dynabeads M-450 Tosylactivated
Dynabeads Antibody Coupling Kit*
* The Dynabeads Antibody Coupling Kits contain buffers for optimal coupling conditions.
Catalog # Name Size Price (USD) Qty
11035 Dynabeads™ Sheep Anti-Rat IgG, 5 mL Each
876.00
11041 Dynabeads™ Pan Mouse IgG, 5 mL Each
793.65
Online Exclusive
820.00
Save 26.35 (3%)
11047 Dynabeads™ Biotin Binder, 5 mL Each
856.00
11204D Dynabeads™ M-280 Sheep Anti-Rabbit IgG, 10 mL Each
1,680.00
11201D Dynabeads™ M-280 Sheep Anti-Mouse IgG, 2 mL Each
540.00
11048D Dynabeads™ Biotin Binder, 2 mL Each
409.00
11036D Dynabeads™ Sheep anti-Rat IgG, 2 mL Each
409.00
11040D Dynabeads™ Pan Mouse IgG, 2 mL Each
395.65
Online Exclusive
409.00
Save 13.35 (3%)
11031 Dynabeads™ Sheep-Anti Mouse IgG Each
878.65
Online Exclusive
906.00
Save 27.35 (3%)
11033 Dynabeads™ Goat Anti-Mouse IgG Each
906.00
11039D Dynabeads™ Rat Anti-Mouse IgM Each
858.00
11203D Dynabeads™ M-280 Sheep Anti-Rabbit IgG, 2 mL Each
540.00
65601 Dynabeads™ MyOne™ Streptavidin T1, 2 mL Each
596.70
Special offer
Online exclusive
Ends: 01-Jun-2025
702.00
Save 105.30 (15%)
65602 Dynabeads™ MyOne™ Streptavidin T1, 10 mL Each
1,976.25
Special offer
Online exclusive
Ends: 01-Jun-2025
2,325.00
Save 348.75 (15%)
65604D Dynabeads™ MyOne™ Streptavidin T1, 50 mL Each
8,092.00
Special offer
Online exclusive
Ends: 01-Jun-2025
9,520.00
Save 1,428.00 (15%)
14011 Dynabeads™ M-450 Epoxy Each
960.00
14013 Dynabeads™ M-450 Tosylactivated Each
1,082.00
14311D Dynabeads™ Antibody Coupling Kit Each
635.65
Online Exclusive
666.00
Save 30.35 (5%)
Positive cell isolation
Positive cell isolation involves directly labeling desired cells for selection using an antibody or ligand that targets a specific cell surface protein.
Positive cell isolation with Dynabeads magnetic beads
In positive cell isolation using Dynabeads magnetic beads, the beads are coated with the specific antibody or ligand to bind to the target cells and the bead/cell complexes are pulled down using a magnet. The supernatant contains non-specific cells and debris which can be discarded, leaving only your pure target cells attached to the bead.
 Cell separation using magnetic beads:
  • Is a gentle, tube-based method
  • Isolates high yields of pure, viable, and functional cells
  • Preserves the cells’ native characteristics
Human positive cell isolation kits
Human icon 
Positive cell isolation kits available for human cells target the following cell surface markers:
Cell surface marker to target for isolationCell type targetedFormat(s) available
CD3Cytotoxic T cells and T helper cellsFlowComp kit
CD4T helper cells, regulatory T cells, and natural killer T cellsFlowComp kit
CD8Cytotoxic T cells FlowComp kit
DETACHaBEAD kit
CD14CD14+ monocytesFlowComp kit
CD19Pan B cellsDETACHaBEAD kit
CD34Progenitor stem cellsDETACHaBEAD kit
CD4/CD25Regulatory CD4+ T cellsDETACHaBEAD kit
EPCAMEpithelial cellsCELLection kit
Learn more about flow compatible Dynabeads kits
Positive cell isolation automation scripts
Automated protocol descriptionSupporting productsDownload automation scripts
Positive cell isolation for downstream cellular applications, such as T cell activation
Catalog # Name Size Price (USD) Qty
11365D Dynabeads™ FlowComp™ Human CD3 Kit Each
1,319.65
Online Exclusive
1,484.00
Save 164.35 (11%)
11361D Dynabeads™ FlowComp™ Human CD4 Kit Each
1,484.00
11362D Dynabeads™ FlowComp™ Human CD8 Kit Each
1,484.00
11333D Dynabeads™ CD8 Positive Isolation Kit Each
1,600.00
11367D Dynabeads™ FlowComp™ Human CD14 Kit Each
1,484.00
12506D DETACHaBEAD™ CD19 Kit Each
770.00
11301D Dynabeads™ CD34 Positive Isolation Kit Each
1,516.00
11363D Dynabeads™ Regulatory CD4+/CD25+ T Cell Kit Each
1,180.65
Online Exclusive
1,252.00
Save 71.35 (6%)
16203 CELLection™ Epithelial Enrich Dynabeads™ Each
1,972.00
Mouse positive cell isolation kits
Mouse icon 
Positive cell isolation kits available for mouse cells target the following cell surface markers:
Cell surface marker to target for isolationCell type targetedFormat(s) available
CD4Naive CD4+ T cellsFlowComp kit
CD8Naive CD8+ T cellsFlowComp kit
CD4/CD25Regulatory CD4+ T cellsFlowComp kit
CD90.2Pan T cellsFlowComp kit
Pan IgGDepends on your antibodyCELLection kit
Dynabeads FlowComp positive cell isolation kits for your flow cytometry cell sorting experiments
In contrast to column-based systems, FlowComp technology lets you isolate bead-free cells directly from human whole blood, buffy coat, or from mouse/human mononuclear cells (MNCs). Following separation, the beads are immediately released and removed, helping ensure the least possible interaction with your cells. Isolated bead-free cells are then ready for use in any downstream application, including flow cytometry. Get high cell recovery, viability, and purity using FlowComp kits and markers.
Explore Dynabeads FlowComp kits and markers
Catalog # Name Size Price (USD) Qty
11461D Dynabeads™ FlowComp™ Mouse CD4 Kit Each
940.00
11462D Dynabeads™ FlowComp™ Mouse CD8 Kit Each
861.65
Online Exclusive
940.00
Save 78.35 (8%)
11465D Dynabeads™ FlowComp™ Mouse Pan T (CD90.2) Kit Each
868.65
Online Exclusive
940.00
Save 71.35 (8%)
11531D CELLection™ Pan Mouse IgG Kit Each
988.00
Build-your-own positive cell isolation kit
Additional positive cell isolation kits are available to use for a wide range of species, with any antibody of your choice.
Use withFormat available
Any antibody targeting any cell type in any speciesFlowComp kit
Any biotinylated antibody targeting any cell type in any speciesCELLection kit
Catalog # Name Size Price (USD) Qty
11061D Dynabeads™ FlowComp™ Flexi Kit Each
976.65
Online Exclusive
1,022.00
Save 45.35 (4%)
11533D CELLection™ Biotin Binder Kit Each
988.00
Negative cell isolation
Negative cell involves labeling unwanted cell types for removal. The unwanted cell types can be bound using magnetic beads coupled to antibodies or ligands that target specific cell surface proteins. The bound, unwanted cells can be depleted from the final isolated fraction by incubating the sample in a magnetic field, leaving desired unbound cells free floating in solution.
Negative cell isolation with Dynabeads magnetic beads
In negative cell isolation, Dynabeads magnetic beads are used to deplete all the unwanted cells using the supplied antibody mix. The target cells are left in the sample untouched and free of beads. The isolated target cells can be directly analyzed in a flow cytometer and used in any functional assay.
Negative cell separation using magnetic beads:
  • Is a gentle, tube-based method
  • Isolates high yields of pure, viable, and functional cells
  • Preserves the cells’ native characteristics
Human negative cell isolation kits
Human icon 
Negative cell isolation kits are available for human cells to target the following cell types:
Enriched untouched cell typeSurface markers on depleted cellsFormat available
CD4+ T cells: T helper cells, regulatory T cells, and natural killer T cellsCD8, CD14, CD16 (a and b), CD19, CD36, CD56, CDw123 and CD235aUntouched kit
CD8+ T cells: cytotoxic T cellsCD4, CD14, CD16 (a and b), CD19, CD36, CD56, CDw123 and CD235aUntouched kit
Dendritic cells (DCs)CD3, CD14, CD16, CD19, CD56, and CD235aUntouched kit
MonocytesCD3, CD7, CD16 (a and b), CD19, CD56, CD123, and CD235aUntouched kit
Pan B cellsCD2, CD14, CD16 (a and b), CD36, CD43, and CD235aUntouched kit
Pan T cellsCD14, CD16 (a and b), CD19, CD36, CD56, CD123, and CD235aUntouched kit
Natural killer (NK) cellsCD3, CD14, CD36, HLA Class II, CD123, and CD235aUntouched kit
Mouse negative cell isolation kits
Mouse icon 
Negative cell isolation kits are available for mouse cells to target the following cell types:
Enriched untouched cell typeSurface markers on depleted cellsFormat available
Naive CD4+ T cellsCD8, CD45R (B220), CD11b (Mac1), Ter-119, and CD16/CD32Untouched kit
Naive CD8+ T cellsCD4, CD45R (B220), CD11b (Mac1), Ter-119, and CD16/CD32Untouched kit
Dendritic cells (DCs)CD2, CD3ε, CD49b, mIgM, and Ter-119Dynabeads kit
Pan B cellsCD43Dynabeads kit
Pan T cellsCD11b, CD16/32, CD45R, and Ter-119Untouched kit
Catalog # Name Size Price (USD) Qty
11415D Dynabeads™ Untouched™ Mouse CD4 Cells Kit, 2 x 10 mL Each
805.65
Online Exclusive
888.00
Save 82.35 (9%)
11417D Dynabeads™ Untouched™ Mouse CD8 Cells Kit Each
805.65
Online Exclusive
888.00
Save 82.35 (9%)
11429D Dynabeads™ Mouse DC (Dendritic Cell) Enrichment Each
1,252.00
11422D Dynabeads™ Mouse CD43 (Untouched™ B Cells) Each
851.65
Online Exclusive
918.00
Save 66.35 (7%)
11413D Dynabeads™ Untouched™ Mouse T Cells Kit Each
809.65
Online Exclusive
888.00
Save 78.35 (9%)
Automate your sample preparation using Dynabeads magnetic beads with protocols for Kingfisher instruments
KingFisher instruments automate the isolation of DNA, RNA, proteins, exosomes, viruses, and cells. KingFisher instruments help eliminate cross contamination and produce reproducible results. Protocol time will vary depending on the target and type of beads.
Learn more about KingFisher instruments for automated protocols
Cell isolation related content
Videos on cell isolation
Video: The two worlds of cell isolation
Video Player is loading.
Current Time 0:00
Duration 7:22
Loaded: 0%
Stream Type LIVE
Remaining Time 7:22
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Video: Uptake and intracellular accumulation of Miltenyi MicroBeads in human CD3+ T cells
    Video Player is loading.
    Current Time 0:00
    Duration 5:04
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 5:04
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      MagniSort cell isolation products
      Conceptual art of a large magnet attracting a magnet-bound surface
      For simpler, faster, and significant cost savings compared to traditional column-based separation methods, use MagniSort technology to obtain enriched T cells, B cells, and NK cells.
      Learn more about MagniSort magnetic cell separation technology
      For Research Use Only. Not for use in diagnostic procedures.
      Hero banner dark gradient overlay
      Stylesheet for Classic Wide Template adjustments