Imaging applications
Imaging reagents detecting cell structures
Using fluorescent dyes, probes, and reagents allows researchers to gain insights into various aspects of cellular biology, such as cellular function, organelle structure, and detect proteins, nucleic acids, lipids, and carbohydrates. These fluorescent reagents can be combined with other dyes or antibodies, enabling multiplex analysis. This multiplexing is achieved by selecting appropriate fluorophores that can be excited at distinct wavelengths and using microscopes equipped with suitable filters to separate the emitted fluorescence signals. By leveraging these reagents, researchers can obtain complex and comprehensive information about cellular processes and structures.
Explore fluorescence microscopy reagents and dyes identifying cellular function and organelle structures:
Cell painting assay reagents
Imaging reagents for apoptosis analysis
Various approaches can used to study apoptosis due to the complex and dynamic nature of programmed cell death cascades. There are an array of fluorescent assays and reagents that can be used to detect these apoptotic process. These assays and reagents can be combined with other dyes or antibodies, each excited and emitted at distinct wavelengths. The fluorescent signals can be visualized using microscopes equipped with suitable filters. This multi-parametric approach allows for a comprehensive understanding of the mechanism involved in cell death.
Explore fluorescence microscopy assays and reagents identifying cells undergoing apoptosis:
illustration of the differences in cellular process of a cell undergoing apoptosis versus necrosis
Interested in the non-apoptotic form of cell death? Learn more about Ferroptosis Research Solutions
Fluorophore technology for microscopy experiments
Learn about fluorophore selection and understand how to pick the correct fluorophore technology for your imaging experiments.
Fluorescence spectra of Alexa Fluor dyes
Overview of Alexa Fluor Dyes
Choose Alexa Fluor fluorescent dyes for your imaging experiments by viewing a selection guide and examining spectral information.
Learn about Alexa Fluor Dyes
Diagram of tyramide signal amplification mechanism of action
Overview of Tyramide Amplification Kits and Reagents
Having issues detecting your low abundant proteins? Understand how to amplify the signal of fluorescent proteins using the tyramide amplification method. 
Learn about TSA
Diagram of Click-iT EdU reaction
Click-iT EdU Assays for cellular proliferation 
Discover a faster and gentler BrdU alternative to measure cell proliferation with Click-iT EdU assays. Read the selection guide and read the protocol.
Learn about Click-iT EdU Assays
ViewRNA Assays for RNA detection
Looking to image your RNA transcripts? Use our commercially available in-situ hybridization assay to visualize RNA in formalin-fixed paraffin-embedded tissue.
Learn about ViewRNA Assays
Curing Antifade Mountants
Want to prevent fluorescence bleaching? Looking for a more permanent mountant? Read about Prolong hard set antifade mountants.
Learn about Prolong Mountants
Non-Setting Antifade Mountants
Want to wash your mountant and re-probe your sample? Understand the properties of SlowFade antifade mountants.
Learn about SlowFade Mountants
Microscopy protocols
Using fluorescent reagents is often less complex compared to other techniques and are more easily detected. Typical labeling times span between 5 and 60 mins. Certain fluorophore signal can be retained with fixation, and a number can be used for live-cell imaging. The protocols and reagents described in imaging protocol handbook and linked below are to be used as a guide only, and may require optimization for your experiment.
Popular microscopy protocols include:
Enhance your imaging experiments with optimized product pairings
Save time with carefully matched product pairings designed to deliver exceptional imaging results.
Explore now
Imaging tools
Find helpful tools below to help you plan your imaging experiments. 
Resources
  • Cell Imaging Resource Center 
    Access articles, webinars and videos, experimental design, and more for imaging microscopy and high-content analysis
  • Molecular Probes School of Fluorescence
    Learn about fluorescence microscopy, find protocols, and get troubleshooting help for your experiments
  • Imaging Insights
    Read or listen to articles on the latest product innovations, novel techniques, and trending news that educate, inform, and inspire
Posters and guides
Related instruments and brochures
Support
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.