Qdot 565 spectra excitation shown as dotted line and emission shown as solid green-yellow histogram
5350, 488, 525574/26300564(in buffer) 5microscopy, flow cytometry
Invitrogen Qdot 565 probe is an excellent orange-fluorescent label with outstanding brightness and photostability for imaging and flow cytometry applications. With an emission maximum of ~565 nm, Qdot 565 probe can be excited using 350, 488, or 525 nm laser lines or a standard TRITC filter. Optimum imaging results with brighter signals are achieved with a Qdot 565 filter or light cube.
Organic fluorophores and Qdot probes can be readily combined in multiplexed flow cytometry or imaging studies. Longer-wavelength Qdot probes are also well-suited for multiplexed western blots.
We offer Qdot 565 probe conjugated to a variety of antibodies, peptides, and proteins, optimized for cellular labeling and detection. In addition, reactive dye forms and antibody labeling kits are available to allow you to generate your own antibody conjugates or probes.
Qdot Label Conjugates for Cell and Tissue Staining    Search Qdot 565 secondary antibodies
Immunofluorescence analysis of Qdot 565 secondary antibody. A549 cells were stained with alpha Tubulin Rat Monoclonal Antibody (YL1/2). The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, blocked with 1% BSA for 1 hour and labeled with 2 µg/mL of Rat primary antibody for 3 hours at room temperature. F(ab')2-Goat anti-Rat IgG (H+L) Secondary Antibody, Qdot 565 was used at 1:1000 dilution in PBS containing 0.2% BSA for 45 minutes at room temperature, for detection of alpha Tubulin in the cytoplasm (Panel a: red). Nuclei (Panel b: blue) were stained with SlowFade Gold Antifade Mountant with DAPI. F-actin was stained with Alexa Fluor 488 Phalloidin, 1:300 (Panel c: green). Panel d represents the composite image. No non-specific staining was observed with the secondary antibody alone (panel f), or with an isotype control (panel e). The images were captured at 60X magnification.
Qdot Probes
Qdot probes are made from nanocrystals of semiconductor material and are broadly compatible with standard fluorescence techniques, but there are some novel aspects to their use that can be optimized to obtain the best possible results. The exceptional brightness and photostability of Qdot probes make it easier to detect low-abundance targets. With narrow and symmetric emission spectra, Qdot probes are also well-suited for multiplexed fluorescence detection using a combination of Qdot probes and organic dyes.
Additional resources
Flow cytometry protocols handbook
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
Request the handbook
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.