BenchStable Media are engineered for flexibility and convenience
Save your lab’s valuable cold storage space with Gibco BenchStable Media, engineered for flexibility and convenience, this media enables storage at room temperature. BenchStable Media is available in the most commonly used basal media formulations: DMEM, DMEM/F-12, MEM, and RPMI 1640. These media have been optimized for routine cell culture, maintaining expected morphology and function of many common cell lines.
Explore the benefits of BenchStable Media:
  • Stable at room temperature—no need to refrigerate means the media temperature is just right when you are ready to use it
  • Flexibility—store it anywhere: on your lab bench or in the refrigerator
  • Easy to use—direct replacement for your current media when supplemented with ≥10% FBS
  • Light protection—bottles provided in protective packaging to help mitigate the risk of light exposure; no more wrapping bottles in foil
BenchStable Media offer a sustainable solution
BenchStable Media have more sustainable packaging and help reduce energy consumption by promoting the efficient use of cold storage when compared to traditional media.
Energy efficient
Globally, a switch by scientists to BenchStable Media from current refrigerated media would save 34 GWh of energy each year, equivalent to the yearly greenhouse gas (GHG) emissions from more than 5,000 passenger cars.
Recyclable materials
BenchStable Media is packaged in a Gibco bottle made from PET with HDPE lid—two of the most highly recycled plastics. The media bottle comes in protective packaging, a paperboard box made from 100% recyclable material, to prevent degradation from light exposure.
Protective packaging
While the paperboard box is added packaging, it contributes 0.05 lb of CO2 equivalents compared with the 6.5 lb of CO2 equivalents saved by enabling ambient media storage, resulting in a 6.45 lb of CO2 equivalents benefit with a switch to BenchStable Media.
Reliable results for routine cell culture with BenchStable Media
Figures 1 and 2 show that BenchStable Media support cellular growth rates equivalent to those observed in conventional media. Figures 3, 4, and 5 show that cell functionality, including morphology and health, is maintained in BenchStable Media.
BenchStable Media maintain expected growth rates
Graph of population doubling times for 4 human cell lines when grown in standard and BenchStable media
Figure 1. BenchStable Media support equivalent growth rates in long-term cultures. HeLa (MEM; pink), HEK293 (DMEM; blue), Jurkat (RPMI; green), and SH-SY5Y (DMEM/F-12; yellow) cells were grown in standard (closed points) and BenchStable (open points) media supplemented with 10% FBS for 15 passages. Cell counts at each passage were used to calculate population doubling times.
Graph of population doubling times for 4 human cell lines when grown in standard and BenchStable media
Figure 2. BenchStable Media support equivalent growth rates in short-term cultures. A549 (MEM; pink), MCF7 (DMEM; blue), THP-1 (RPMI; green), and CHO-K1 (DMEM/F-12; yellow) cells were grown in standard (closed points) and BenchStable (open points) media supplemented with 10% FBS for 5 passages. Cell counts at each passage were used to calculate population doubling times.
BenchStable Media maintain cell morphology
Brightfield microscope views of HEK293 cells cultured in conventional DMEM or BenchStable DMEM
Figure 3. BenchStable DMEM supports HEK293 cell culture. HEK293 human embryonic kidney cells were cultured for 15 passages in DMEM (leftCat. No. 10566016) or BenchStable DMEM (rightCat. No. A4192101) supplemented with 10% FBS. Images were captured at passage 15 using 10x magnification on an EVOS FL Auto Imaging System.
Brightfield microscope views of HeLa cells cultured in conventional MEM or BenchStable MEM
Figure 4. BenchStable DMEM supports HeLa cell culture. HeLa human cervical adenocarcinoma cells were cultured for 15 passages in MEM (leftCat. No. 41090036) or BenchStable MEM (rightCat. No. A4192201) supplemented with 10% FBS. Images were captured at passage 15 using 10x magnification on an EVOS FL Auto Imaging System.
BenchStable Media maintain cell health
benchstable-mitochondrial-health-figure5
Figure 5. Cells cultured in BenchStable Media maintain healthy mitochondria. HeLa cells cultured in DMEM (left) and BenchStable DMEM (right) for eight passages were stained with tetramethylrhodamine methyl ester (TMRM; Cat. No. T668) and Hoechst 33342 Solution (Cat. No. 62249). TMRM is a cell-permeant dye that accumulates in active mitochondria, resulting in a bright signal in healthy cells.
BenchStable Media videos
Watch these videos to learn how you can break through your lab's limitations in cold-storage space with BenchStable Media.
BenchStable Media literature
The coolest media are totally uncool.
Get out of the cold and warm up to room temperature stable cell culture media.
Order BenchStable Media
Catalog # Name Size Price (USD) Qty
A4192001 Each
66.65
Online Exclusive
71.75
Save 5.10 (7%)
A4192101 Each
44.65
Online Exclusive
48.37
Save 3.72 (8%)
A4192002 Each
550.65
Online Exclusive
596.00
Save 45.35 (8%)
A4192102 Each
399.65
Online Exclusive
431.00
Save 31.35 (7%)
A4192201 Each
49.91
A4192202 Each
434.00
A4192301 Each
45.65
Online Exclusive
49.31
Save 3.66 (7%)
A4192302 Each
397.65
Online Exclusive
432.00
Save 34.35 (8%)
Other room temperature-stable products
These highly purified, recombinant enzymes provide fast yet gentle cell dissociation. Ideal for dissociating attachment-dependent cell lines, TrypLE reagents can be directly substituted for trypsin without protocol changes.
Extend the life of your cells with GlutaMAX supplement. GlutaMAX supplement is stable in aqueous solutions and does not spontaneously degrade. This improved cell culture supplement can be used as a direct substitute for L-glutamine in your cell culture media.
Already using Gibco media? Here is a quick cross-reference to help you make the switch. BenchStable Media contain GlutaMAX supplement and phenol red.
Try Gibco One Shot FBS for quick and easy 10% FBS supplementation
Frequently asked questions (FAQs) about BenchStable Media
Do you need more information about using BenchStable Media in your cell culture experiments? Get your answers by visiting our FAQs for BenchStable Media.
Cell Culture Samples and Request a Quote Program
Simply complete the form and consult with a specialist to receive your sample(s) and/or quote.
GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Resources and support
Resources
Gibco Media Selection Tool
Find the right Gibco media for your application.  Use this interactive tool to select media based upon culture type, components, additives, and format. 
Cell Culture Select Tool
Search your cell line of interest to find product recommendations and resources to help enable a successful cell culture.
Gibco Cell Culture Basics
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Cell Culture Videos
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and the latest advancements in the field.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.