Culture your cells with Gibco DMEM
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12.
Gibco DMEM is formulated for superior quality, consistency, and control for your mammalian cell culture.
DMEM composition
Dulbecco's Modified Eagle Medium (DMEM) is modified to contain four times the amino acid and vitamin concentration of original Eagle's Minimal Essential Medium. The original DMEM formulation contained a low concentration of glucose (1 g/L) and sodium pyruvate. However, DMEM is now available in a variety of formulations, including those with higher glucose levels and with or without sodium pyruvate. DMEM also uses a sodium bicarbonate buffer system, allowing for maintenance of physiological pH in a 5–10% CO2 environment. Importantly, DMEM commonly requires fetal bovine serum (FBS) supplementation.
Thermo Fisher Scientific offers a variety of Gibco DMEM formulations to support a wide range of cell culture applications—view an example of a detailed DMEM formulation here. Learn about additional DMEM formulation options in the Gibco Media selection tool.
Choose the right DMEM
There are several factors to consider when selecting the right Gibco DMEM composition
for your cell cultures, including format, supplementation, and additives. Our interactive
media selection tool helps enable easy selection of the DMEM you need.
Innovative DMEM formulations
Take your cell culture to the next level with enhanced DMEM formulations. Explore the following formulations that can help improve your cell viability, serum supplementation, cell imaging, and cold storage constraints.
DMEM Advanced
Advanced DMEM
Gibco Advanced DMEM is enriched with normal-serum constituents and therefore requires 50–90% less FBS supplementation, helping reduce variability in your cell culture and lower your cost. 
FluoroBrite DMEM
FluoroBrite DMEM
Gibco FluoroBrite DMEM is a DMEM-based media formulation with background fluorescence that is 90% lower than that emitted by standard phenol red-free DMEM, enhancing the signal-to-noise ratio of fluorophores to visualize even the weakest fluorescent events. 
BenchStable DMEM
BenchStable DMEM
Gibco BenchStable DMEM is formulated for storage at room temperature, supporting cell proliferation and densities comparable to conventional DMEM while freeing up cold storage space.
DMEM with GlutaMAX supplement
DMEM with GlutaMAX supplement
Gibco DMEM with GlutaMAX supplement provides your cells with a stable source of L-glutamine that does not spontaneously degrade in cell culture, improving the health and viability of your cells by helping reduce the accumulation of toxic ammonia.
Custom DMEM
If you require a custom DMEM formulation for your cell culture experiments, our media customization services can provide you with the media you need, including custom components, packaging, and QC testing.
GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Resources and support
Resources
Gibco Media Selection Tool
Find the right Gibco media for your application.  Use this interactive tool to select media based upon culture type, components, additives, and format. 
Cell Culture Select Tool
Search your cell line of interest to find product recommendations and resources to help enable a successful cell culture.
Gibco Cell Culture Basics
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Cell Culture Videos
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and the latest advancements in the field.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.