Chromatin immunoprecipitation (ChIP) assays are performed to identify regions of the genome with which DNA-binding proteins, such as transcription factors and histones, associate. In ChIP assays, proteins bound to DNA are temporarily crosslinked and the DNA is sheared prior to cell lysis. The target proteins are immunoprecipitated along with the crosslinked nucleotide sequences, and the DNA is then removed and identified by PCR, sequenced, applied to microarrays, or analyzed in some other way.
RNA immunoprecipitation (RIP) uses an approach similar to ChIP, except that RNA-binding proteins are immunoprecipitated instead of DNA-binding proteins. Immunoprecipitated RNAs can then be identified by RT-PCR and cDNA sequencing.
See information on Co-IP Products
Protein-Nucleic Acid Pull-Downs, in particular, Protein-RNA Pull-Downs are performed by enriching RNA binding proteins using end-labeled RNA with desthiobiotin as the bait. Ribonucleoprotein complexes are captured using streptavidin magnetic beads, and the complexes can be recovered using biotin elution buffer. Downstream applications include western blotting and mass spectrometry (MS).
View products
ChIP assay procedure
Chromatin Immunoprecipitation assays (ChIP assays) identify links between the genome and the proteome by monitoring transcription regulation through histone modification (epigenetics) or transcription factor:DNA binding interactions. The strength of ChIP assays is their ability to capture a snapshot of specific protein:DNA interactions occurring in a system and to quantify the interactions using quantitative polymerase chain reaction (qPCR).
The chromatin immunoprecipitation (ChIP) assay is a powerful method for analyzing epigenetic modifications and genomic DNA sequences bound to specific regulatory proteins.
Chromatin IP experiments require a variety of proteomics and molecular biology methods including crosslinking, cell lysis (protein-DNA extraction), nucleic acid shearing, antibody-based immunoprecipitation, DNA sample clean-up and PCR. Additional techniques such as gel electrophoresis are usually used during optimization experiments to validate specific steps.
Crosslinking
Chromatin immunoprecipitation (ChIP) assays begin with covalent stabilization of protein-DNA complexes. Many protein-DNA interactions are transient, and involve multi-protein complexes to orchestrate biological function. In vivo crosslinking covalently stabilizes protein-DNA complexes.
In vivo crosslinking is traditionally achieved with formaldehyde but can be combined with other crosslinkers such as EGS and DSG. Formaldehyde crosslinking is ideal for two molecules which interact directly. However, formaldehyde is a zero-length crosslinker, limiting its functionality. For higher order interactions, longer crosslinkers such as EGS (16.1 Angstroms) or DSG (7.7 Angstroms) can trap larger protein complexes with complex quaternary structure.
Researchers often use a combination of crosslinkers to trap protein-DNA interacting partners. These crosslinkers permeate directly into intact cells to effectively lock protein-DNA complexes together, allowing even transient interaction complexes to be trapped and stabilized for analysis.
Cell lysis
The lysis stage extracts the crosslinked protein-DNA complexes from cells or tissue and brings them into solution. At this stage, cellular components are liberated by dissolving the cell membrane with detergent based solutions. Because protein-DNA interactions occur primarily in the nuclear compartment, removing cytosolic protein can help reduce background and increase sensitivity. The presence of detergents or salts will not affect the protein-DNA complex, as the covalent crosslinking achieved in step one will keep the complex stable throughout the ChIP procedure.
Mechanical lysis of cells is not recommended, as it can result in inefficient nuclear lysis. Reagents such as the Thermo Scientific Pierce Chromatin Prep Module, which isolate the nuclear fraction from other cellular components, are used to eliminate background signal and enhance sensitivity.
Chromatin preparation
The extraction step yields all nuclear material, which includes unbound nuclear protein, full length chromatin and the crosslinked protein-DNA complexes. In order to analyze protein binding sequences, the extracted genomic DNA must be sheared into smaller, workable pieces. DNA fragmentation is usually achieved either mechanically by sonication or enzymatically by digestion with micrococcal nuclease (MNase).
Ideal chromatin fragments can range from 200 to >1000 bp, however DNA shearing is one of the most difficult steps to control. Sonication provides truly randomized fragments, but limitations include the requirement of dedicated machinery which may need tuning, difficulty in maintaining temperature during sonication and extended hands-on time and extensive optimization steps. Enzymatic digestion with micrococcal nuclease is highly reproducible and more amendable to processing multiple samples, but can lead to variability due to changes in enzyme activity and the enzyme has higher affinity for inter-nucleosome regions.
Immunoprecipitation
To isolate a specific modified histone, transcription factor or co-factor of interest, ChIP validated antibodies are used to immunoprecipitate and isolate the target from other nuclear components. This step selectively enriches for the protein-DNA complex of interest and eliminates all other unrelated cellular material.
Selection of the appropriate antibody is a critical parameter to successful ChIP assays. For mammalian samples, numerous ChIP-grade antibodies are available that are validated for this procedure. For target proteins in which qualified antibodies are unavailable, fusion proteins such as HA, myc or GST can be expressed in the biological sample and then antibodies against the affinity tags can be used to immunoprecipitate the target.
The antibody-protein-DNA complex is affinity purified using an antibody binding resin such immobilized protein A, protein G or protein A/G. For biotinylated antibodies, immobilized streptavidin or immobilized Thermo Scientific NeutrAvidin Protein can also be used. For reduced background, it is necessary to block antibody binding beads with a combination of nucleic acid and protein blocking buffers such as salmon sperm DNA and a generic protein source. The volume of beads used in each ChIP sample can also influence background, as the increase in bead volume increases non-specific binding.
Crosslinking Reversal and DNA Clean-up
Enrichment of DNA bound to the protein of interest is the goal for chromatin immunoprecipitation. DNA levels can be determined by agarose gel electrophoresis or more commonly by quantitative polymerase chain reaction (qPCR). Before the specific DNA products of a chromatin IP can be amplified and measured, the crosslinks between protein and DNA must to be reversed. This is typically done through extensive heat incubations or through digestion of the protein component with proteinase K.
Proteinase K cleaves at the carboxy-side of aliphatic, aromatic or hydrophobic residues. Because of its broad specificity, proteinase K is often used for removing proteins from DNA or RNA preparations. Additionally, proteinase K digestion eliminates nucleases from the purified DNA, which prevents degradation. To separate DNA from protein fragments, phenol-chloroform is used in conjunction with a standard DNA purification method. Alternatively, spin columns designed to purify nucleic acid material from complex biological samples may be used.
DNA quantitation
A hallmark of ChIP is the ability to quantitate the purified DNA products with quantitative PCR. Despite being amplification techniques, qPCR procedures are sufficiently accurate to enable measurement of target protein-DNA levels in different experimental conditions. There is a direct correlation between the amounts of immunoprecipitated complex and bound DNA.
Crosslinking
Chromatin immunoprecipitation (ChIP) assays begin with covalent stabilization of protein-DNA complexes. Many protein-DNA interactions are transient, and involve multi-protein complexes to orchestrate biological function. In vivo crosslinking covalently stabilizes protein-DNA complexes.
In vivo crosslinking is traditionally achieved with formaldehyde but can be combined with other crosslinkers such as EGS and DSG. Formaldehyde crosslinking is ideal for two molecules which interact directly. However, formaldehyde is a zero-length crosslinker, limiting its functionality. For higher order interactions, longer crosslinkers such as EGS (16.1 Angstroms) or DSG (7.7 Angstroms) can trap larger protein complexes with complex quaternary structure.
Researchers often use a combination of crosslinkers to trap protein-DNA interacting partners. These crosslinkers permeate directly into intact cells to effectively lock protein-DNA complexes together, allowing even transient interaction complexes to be trapped and stabilized for analysis.
Cell lysis
The lysis stage extracts the crosslinked protein-DNA complexes from cells or tissue and brings them into solution. At this stage, cellular components are liberated by dissolving the cell membrane with detergent based solutions. Because protein-DNA interactions occur primarily in the nuclear compartment, removing cytosolic protein can help reduce background and increase sensitivity. The presence of detergents or salts will not affect the protein-DNA complex, as the covalent crosslinking achieved in step one will keep the complex stable throughout the ChIP procedure.
Mechanical lysis of cells is not recommended, as it can result in inefficient nuclear lysis. Reagents such as the Thermo Scientific Pierce Chromatin Prep Module, which isolate the nuclear fraction from other cellular components, are used to eliminate background signal and enhance sensitivity.
Chromatin preparation
The extraction step yields all nuclear material, which includes unbound nuclear protein, full length chromatin and the crosslinked protein-DNA complexes. In order to analyze protein binding sequences, the extracted genomic DNA must be sheared into smaller, workable pieces. DNA fragmentation is usually achieved either mechanically by sonication or enzymatically by digestion with micrococcal nuclease (MNase).
Ideal chromatin fragments can range from 200 to >1000 bp, however DNA shearing is one of the most difficult steps to control. Sonication provides truly randomized fragments, but limitations include the requirement of dedicated machinery which may need tuning, difficulty in maintaining temperature during sonication and extended hands-on time and extensive optimization steps. Enzymatic digestion with micrococcal nuclease is highly reproducible and more amendable to processing multiple samples, but can lead to variability due to changes in enzyme activity and the enzyme has higher affinity for inter-nucleosome regions.
Immunoprecipitation
To isolate a specific modified histone, transcription factor or co-factor of interest, ChIP validated antibodies are used to immunoprecipitate and isolate the target from other nuclear components. This step selectively enriches for the protein-DNA complex of interest and eliminates all other unrelated cellular material.
Selection of the appropriate antibody is a critical parameter to successful ChIP assays. For mammalian samples, numerous ChIP-grade antibodies are available that are validated for this procedure. For target proteins in which qualified antibodies are unavailable, fusion proteins such as HA, myc or GST can be expressed in the biological sample and then antibodies against the affinity tags can be used to immunoprecipitate the target.
The antibody-protein-DNA complex is affinity purified using an antibody binding resin such immobilized protein A, protein G or protein A/G. For biotinylated antibodies, immobilized streptavidin or immobilized Thermo Scientific NeutrAvidin Protein can also be used. For reduced background, it is necessary to block antibody binding beads with a combination of nucleic acid and protein blocking buffers such as salmon sperm DNA and a generic protein source. The volume of beads used in each ChIP sample can also influence background, as the increase in bead volume increases non-specific binding.
Crosslinking Reversal and DNA Clean-up
Enrichment of DNA bound to the protein of interest is the goal for chromatin immunoprecipitation. DNA levels can be determined by agarose gel electrophoresis or more commonly by quantitative polymerase chain reaction (qPCR). Before the specific DNA products of a chromatin IP can be amplified and measured, the crosslinks between protein and DNA must to be reversed. This is typically done through extensive heat incubations or through digestion of the protein component with proteinase K.
Proteinase K cleaves at the carboxy-side of aliphatic, aromatic or hydrophobic residues. Because of its broad specificity, proteinase K is often used for removing proteins from DNA or RNA preparations. Additionally, proteinase K digestion eliminates nucleases from the purified DNA, which prevents degradation. To separate DNA from protein fragments, phenol-chloroform is used in conjunction with a standard DNA purification method. Alternatively, spin columns designed to purify nucleic acid material from complex biological samples may be used.
DNA quantitation
A hallmark of ChIP is the ability to quantitate the purified DNA products with quantitative PCR. Despite being amplification techniques, qPCR procedures are sufficiently accurate to enable measurement of target protein-DNA levels in different experimental conditions. There is a direct correlation between the amounts of immunoprecipitated complex and bound DNA.
ChIP and RIP product ordering table
Order Chromatin Immunoprecipitation (ChIP) products
Catalog # Name Size Price (USD) Qty
492024 MAGnify™ Chromatin Immunoprecipitation System Each
884.00
26157 Pierce™ Magnetic ChIP Kit Each
764.65
Online Exclusive
850.00
Save 85.35 (10%)
26162 Pierce™ ChIP-grade Protein A/G Magnetic Beads Each
975.65
Online Exclusive
1,094.00
Save 118.35 (11%)
26160 Proteinase K Solution, ChIP grade Each
144.00
14311D Dynabeads™ Antibody Coupling Kit Each
673.65
Online Exclusive
706.00
Save 32.35 (5%)
10001D Dynabeads™ Protein A for Immunoprecipitation, 1 mL Each
247.65
Online Exclusive
265.00
Save 17.35 (7%)
10003D Dynabeads™ Protein G for Immunoprecipitation, 1 mL Each
253.65
Online Exclusive
265.00
Save 11.35 (4%)
Order Protein-Nucleic Acid Pull-Down and RNA Immunoprecipitation (RIP) products
Catalog # Name Size Price (USD) Qty
20164 Pierce™ Magnetic RNA-Protein Pull-Down Kit Each
1,012.00
20163 Pierce™ RNA 3' End Desthiobiotinylation Kit Each
673.65
Online Exclusive
828.00
Save 154.35 (19%)
88816 Pierce™ Streptavidin Magnetic Beads, 1 mL Each
359.55
Special offer
Online exclusive
Ends: 01-Jun-2025
423.00
Save 63.45 (15%)
10001D Dynabeads™ Protein A for Immunoprecipitation, 1 mL Each
247.65
Online Exclusive
265.00
Save 17.35 (7%)
10003D Dynabeads™ Protein G for Immunoprecipitation, 1 mL Each
253.65
Online Exclusive
265.00
Save 11.35 (4%)
14311D Dynabeads™ Antibody Coupling Kit Each
673.65
Online Exclusive
706.00
Save 32.35 (5%)
65601 Dynabeads™ MyOne™ Streptavidin T1, 2 mL Each
632.40
Special offer
Online exclusive
Ends: 01-Jun-2025
744.00
Save 111.60 (15%)
11205D Dynabeads™ M-280 Streptavidin, 2 mL Each
652.80
Special offer
Online exclusive
Ends: 01-Jun-2025
768.00
Save 115.20 (15%)
65801D Dynabeads™ Streptavidin Trial Kit Each
302.40
Special offer
Online exclusive
Ends: 01-Jun-2025
378.00
Save 75.60 (20%)
The publication trend for immunoprecipitation says it all. The number of publications citing the use of Dynabeads magnetic beads vs. other technologies and products for immunoprecipitation procedures is skyrocketing.
Dynabeads IP citations
Dynabeads IP citations.
Dynabeads IP citations
Dynabeads ChIP citations.
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.