1x1 image pixel for data collection
Pierce™ Streptavidin Magnetic Beads
New Protein Purification and Isolation Support Center—Find technical tips and troubleshooting recommendations from our scientists.
Pierce™ Streptavidin Magnetic Beads
Thermo Scientific™

Pierce™ Streptavidin Magnetic Beads

Purify or immunoprecipitate biotinylated proteins or affinity ligands using streptavidin-coated magnetic particles for magnet-based separation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
888161 mL
888175 mL
Catalog number 88816
Price (USD)
374.65
Online Exclusive
423.00
Save 48.35 (11%)
Each
In stock
Add to cart
Quantity:
1 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
374.65
Online Exclusive
423.00
Save 48.35 (11%)
Each
Add to cart
Ask our AI about this Product

Thermo Scientific Pierce Streptavidin Magnetic Beads accelerate throughput for automated magnetic purification of biotinylated molecules.

Features of Streptavidin Magnetic Beads:

  • High-performance beads—non-aggregating, pre-blocked, iron oxide, superparamagnetic microparticles provide exceptional uniformity for automated HTS and manual applications alike
  • Stable immobilization chemistry—streptavidin is immobilized using leach-resistant chemistry
  • High capacity—superior quality beads with high binding capacity provide rapid and efficient biomolecule purification from complex samples
  • Low non-specific binding—stable, pre-blocked beads provide clean purification products (e.g., antigen eluted in IP with biotinylated antibody) that are compatible with mass spectrometry analysis and other downstream analyses
  • Superior performance—nearly three times higher binding capacity than typical beads from other suppliers, allowing the use of smaller amounts per experiment

Applications:

  • Immunoprecipitate antigens (using biotinylated antibodies) from a wide variety of sources
  • Co-immunoprecipitate interaction complexes using biotinylated antibodies
  • Capture protein-protein interactions in pull-down assays using biotinylated 'bait' proteins
  • Isolate biotin-labeled DNA-protein complexes from cell or tissue extracts
  • Capture single-stranded biotinylated DNA oligos
  • Isolate biotinylated PCR products

These streptavidin magnetic beads are validated and optimized for use with high-throughput magnetic platforms, such as the Thermo Scientific KingFisher 96 and KingFisher Flex Instruments, but the beads also enable premium performance for simple benchtop applications using an appropriate magnetic stand. The iron oxide, super-paramagnetic particles offer superior performance (high capacity and low nonspecific binding) compared with other commercial magnetic beads.

Pierce Streptavidin Magnetic Beads use a recombinant form of streptavidin with a mass of 53kDa and a near-neutral isoelectric point (pI). The protein is a tetramer having four biotin-binding sites. Unlike avidin, streptavidin has no carbohydrate groups, resulting in low nonspecific binding. The high-affinity interaction between streptavidin and biotin cannot be dissociated efficiently except with very harsh conditions, such as boiling in sample loading buffer for SDS-PAGE or 8M guanidine·HCl, pH 1.5. Consequently, it is often possible to elute binding partners in an interaction complex without also eluting the biotinylated component.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConcentrationSlurry: 10 mg/mL, 1% solids
Ligand TypeStreptavidin
Protein FormRecombinant
Quantity1 mL
TargetBiotin
Capacity (Metric)1 mL
FormLiquid
Particle Size1 μm
Product LinePierce™
TypeMagnetic Affinity Bead
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C. Product is shipped on an ice pack.
Have questions about this product? Ask our AI assisted search.
What are the average sizes of your streptavidin magnetic beads?
What is the estimated amount of streptavidin in your Pierce Streptavidin Magnetic Beads?
How many particles are present per mL of Pierce Streptavidin Magnetic Beads?
Are the MagnaBind Streptavidin Beads and Thermo Scientific Streptavidin Magnetic Beads sterile?
What is the magnetite content of Pierce Streptavidin Magnetic Beads?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Will the streptavidin-biotin bond be stable in ammonium carbonate buffer at 56 degrees C?

The streptavidin-biotin interaction is the strongest known non-covalent biological interaction between a protein and a ligand. Binding of biotin and streptavidin is very rapid and, once formed, the complex is unaffected by wide extremes of pH, temperature, organic solvents, and other denaturing agents. Normally, very harsh methods are required to dissociate the biotin from streptavidin, which will be irreversibly denatured by the procedure.

To dissociate biotinylated proteins from streptavidin, boil the beads in 0.1% SDS or SDS-PAGE buffer for 3 mins or incubate them in 8 M guanidinium hydrochloride, pH 1.5.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I have a dsDNA biotinylated on streptavidin Dynabeads. How can I dissociate the non-biotinylated DNA strand from the biotinylated one?

There are two methods to dissociate the non-biotinylated DNA from the biotinylated DNA strand. The following protocols are based on using 20 µL of Dynabeads Streptavidin, but are scalable. Both methods may release very small amounts of complementary biotinylated strand from streptavidin. If it is critical that no biotinylated strand is released, either adopt a different biotin modification using dual biotin (two biotin groups in sequence) or covalently bind DNA to e.g., Dynabeads M-270 Carboxylic Acid.

Using heat:

- Wash the DNA coated Dynabeads in 50 µL 1 x SSC.*
- Resuspend the beads in another 50 µL of 1 x SSC Incubate at 95 degrees C for 5 mins.
- Quickly put the tube in magnet stand for 1-2 mins and transfer the supernatant to a new tube.
- The supernatant contains your non-biotinylated DNA strand.

Using NaOH:

- Wash the DNA coated Dynabeads in 50 µL 1 x SSC.*
- Resuspend the beads in 20 µl of freshly prepared 0.15 M NaOH.
- Incubate at room temperature for 10 mins. Put the tube in magnet stand for 1-2 mins and transfer the supernatant to a new tube.
- The supernatant contains your non-biotinylated DNA strand. Neutralize the probe by adding 2.2 µL 10 x TE, pH 7.5 and 1.3 µL 1.25 M acetic acid.

Wash the Dynabeads coated with biotinylated strand once with 50 µL 0.1M NaOH, once with 50 µL of B&W buffer and once with 50 µL TE buffer.

*1 x SSC (0.15 M NaCl, 0.015 M sodium citrate. Dissolve the reagents in 800 mL water. Adjust pH to 7.0 with NaOH. Adjust the volume to 1 liter with water).

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

How do I measure the binding of biotinylated molecules on streptavidin Dynabeads?

Assay the supernatant for unbound molecules. This will determine the amount of molecule bound to the Dynabeads. For nucleic acids, the concentration can be checked by OD readings, or by running a gel. For proteins, the concentration in the supernatant can be determined by a spectrometer using a protein assay like BCA. Alternatively, you can label the molecule with radioactivity or fluorescence and measure the concentration of molecule directly on the beads (former) or in the supernatant (latter).

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

How many biotin binding-sites are there per streptavidin molecule?

Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Is the streptavidin protein in Pierce Streptavidin Magnetic Beads His-tagged?

The streptavidin in Pierce Streptavidin Magnetic Beads is not His-tagged.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3213779Certificate of AnalysisJul 03, 202588817
3213269Certificate of AnalysisJun 23, 202588816
3213780Certificate of AnalysisJun 18, 202588817
3213778Certificate of AnalysisJun 02, 202588817
3213251Certificate of AnalysisMay 29, 202588817
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (6)

Citations & References
Abstract
Proximity Labeling of the Chlamydia trachomatis Inclusion Membrane.
Authors:Olson MG,Jorgenson LM,Widner RE,Rucks EA
Journal:Methods in molecular biology (Clifton, N.J.)
PubMed ID:31385281
In the study of intracellular bacteria that reside within a membrane-bound vacuole, there are many questions related to how prokaryotic or eukaryotic transmembrane or membrane-associated proteins are organized and function within the membranes of these pathogen-containing vacuoles. Yet this host-pathogen interaction interface has proven difficult to experimentally resolve. For example, ... More
The rapid proximity labeling system PhastID identifies ATP6AP1 as an unconventional GEF for Rheb.
Authors:Feng R,Liu F,Li R,Zhou Z,Lin Z,Lin S,Deng S,Li Y,Nong B,Xia Y,Li Z,Zhong X,Yang S,Wan G,Ma W,Wu S,Songyang Z
Journal:Cell research
PubMed ID:38448650
Rheb is a small G protein that functions as the direct activator of the mechanistic target of rapamycin complex 1 (mTORC1) to coordinate signaling cascades in response to nutrients and growth factors. Despite extensive studies, the guanine nucleotide exchange factor (GEF) that directly activates Rheb remains unclear, at least in ... More
Proximity labeling reveals dynamic changes in the SQSTM1 protein network.
Authors:Rondón-Ortiz AN,Zhang L,Ash PEA,Basu A,Puri S,van der Spek SJF,Wang Z,Dorrian L,Emili A,Wolozin B
Journal:The Journal of biological chemistry
PubMed ID:39098523
Sequestosome1 (SQSTM1) is an autophagy receptor that mediates the degradation of intracellular cargo, including protein aggregates, through multiple protein interactions. These interactions form the SQSTM1 protein network, and these interactions are mediated by SQSTM1 functional interaction domains, which include LIR, PB1, UBA, and KIR. Technological advances in cell biology continue ... More
Isolation of secreted proteins from Drosophila ovaries and embryos through in vivo BirA-mediated biotinylation
Authors:Stevens LM, Zhang Y, Volnov Y, Chen G, Stein DS
Journal:PLoS ONE
PubMed ID:
The role of the PZP domain of AF10 in acute leukemia driven by AF10 translocations
Authors:Klein BJ, Deshpande A, Cox KL, Xuan F, Zandian M, Barbosa K, Khanal S, Tong Q, Zhang Y, Zhang P, Sinha A, Bohlander SK, Shi X, Wen H, Poirier MG, Deshpande A, Kutateladze TG
Journal:Nature Communications
PubMed ID:
6 total citations

Share catalog number, name or link

1x1 image pixel for data collection