Consistency and capacity for everyday applications
Thermo Scientific General Purpose Plus Centrifuges offer versatility with up to 15 rotor options. Their mid-performance drives are designed for applications demanding consistency, repeatability, and high throughput run after run, such as basic research and clinical applications with repetitive protocols. Ergonomic design and ease of use make them a robust, reliable solution for labs with multiple users. Choose either the 1.6 L or 4 L capacity configuration, available in ventilated and refrigerated models to match your needs. The 4 L models are available as benchtop and floor-standing versions.
Key product highlights
Easy-to-use LCD control panel
Standard, easy-to-use interface with button-functionality and bright LCD display.
Quick rotor exchange
Auto-Lock rotor exchange provides flexibility to quickly switch between applications in less than three seconds without the need for any tools.
Protective of user and samples
Biocontainment sealing options, including certified1 ClickSeal lids offer glove-friendly, one-handed operation.
Exceptionally versatile for multi-user settings
Compact 1.6 L capacity models support up to 15 optional swinging bucket and fixed angle rotors. For maximum capacity in 4 L models, the spacious TX-1000 rotor can run up to 196 blood tubes and forty 50 mL conical tubes. The 4 L models support up to 13 different rotors. The Auto-Lock system lets you easily switch between rotors in seconds, with one hand, for great flexibility. 
Bright, easy-to-use LCD control panel
Programs are directly accessible from six buttons. Simply scroll to enter speed, time, temperature, and acceleration and deceleration parameters. Select a pulse run or open the lid with a push of a button. The panel displays English, German, French, Spanish, Italian, Dutch, and Russian languages. 
Feature overview of compact centrifuges
Product literature for standard gp centrifuges
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.