Hydrophilic
A range of surfaces to optimize protein binding
Maximize passive adsorption and coating capacity of hydrophilic molecules such as antibodies, proteins, and glycans using these plates manufactured with hydrophilic surfaces including MaxiSorp, MediSorp, and MultiSorp.
Hydrophilic surface types
Slightly hydrophilic:
The slight hydrophilicity of these plate surfaces enhances their ability to bind a diverse range of biomolecules, including glycoproteins, serum-containing samples, and amphoteric molecules such as lipopolysaccharides. With these plates, non-specific adsorption of serum-containing samples is reduced, improving the signal-to-noise (S/N) ratio and consequently, the sensitivity levels.
As the surface is slightly hydrophilic, the maximum binding capacity for a glycoprotein (antibody) will be lower compared to the hydrophilic surface. This will mean that the maximum binding capacity for IgG is less than 650ng/cm2. The immobilization conditions should be calculated from the pI of the molecule that will be coated onto the surface. It is recommended that the pH>pI and a typical coating buffer will often be a carbonate buffer (pH= 9.6) when working with these surfaces.
Catalog # Name Size Price (USD) Qty
7572 White 96-Well Immuno Plates, Microlite 2+ Case of 50
313.20
Online Exclusive
348.00
Save 34.80 (10%)
9503100 Microlite™ White Microtiter™ Plates, Strip Removal Tool, Microlite 2 - -
7562 MicroTiter™ Luminescence Strip Microplates (Assembled), Microlite 2+ Assembly Each -
446470 Immuno Breakable Modules Clear, C8 LockWell, MediSorp Case of 60
499.20
Online Exclusive
624.00
Save 124.80 (20%)
6302 Immuno Clear Standard Modules, F12, 2 HB Case of 320
172.42
Online Exclusive
252.00
Save 79.58 (32%)
7567 Immuno Standard Modules White and Black, Immulon, F12, Microlite 2, 380μL Case of 320
241.79
Online Exclusive
353.00
Save 111.21 (32%)
467320 Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, 400μL, MediSorp Case of 60
367.20
Online Exclusive
408.00
Save 40.80 (10%)
467120 Immuno Clear Standard Modules, F8, MediSorp, framed Case of 60
580.50
Online Exclusive
645.00
Save 64.50 (10%)
3455 Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, 330μL, 2 HB Case of 50
232.20
Online Exclusive
258.00
Save 25.80 (10%)
7417 Microlite™ White Microtiter™ Plates, Plate, Microlite 2 Each -
7905 White 96-Well Immuno Plates, Immulon, Flat-bottom, Microfluor 2, 280μL Case of 50
315.00
Online Exclusive
350.00
Save 35.00 (10%)
6309 Immuno Clear Standard Modules, F12, 2 HB, framed Case of 100
537.25
Online Exclusive
786.00
Save 248.75 (32%)
7805 Black 96-Well Immuno Plates, 330μL, Microfluor 2 Case of 50
298.80
Online Exclusive
332.00
Save 33.20 (10%)
Hydrophilic:
The hydrophilic surfaces are optimized to bind a high amount of IgG (polyclonal), and these surfaces are ideal for antibody sandwich assays in the enzyme link immuno sorbent assay (ELISA). In addition, they show increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic characteristics. The high binding capacity is achieved by a physical modification of the surface.
The maximum binding capacity for IgG is 650ng/cm2. Due to the polarised nature of hydrophilic groups, hybrid hydrophilic and ionic bonds may also be established. Since hydrophilic and ionic groups are often scattered among hydrophobic moieties in macromolecules, a stable adsorption may not be obtained by hydrophilic and ionic bonds alone. A hydrophilic/hydrophobic surface should therefore be chosen, to match hydrophilic/hydrophobic patchwork of the molecule.
Catalog # Name Size Price (USD) Qty
15042 Pierce™ 96-Well Polystyrene Plates, White Opaque Each
151.65
Online Exclusive
177.00
Save 25.35 (14%)
95029490 Immuno Standard Modules, EB, black Case of 50 -
15041 Pierce™ 96-Well Polystyrene Plates, Corner Notch Each
508.65
Online Exclusive
556.00
Save 47.35 (9%)
434797 Immuno Clear Standard Modules, F8, MaxiSorp, framed Case of 120
876.07
Online Exclusive
1,283.00
Save 406.93 (32%)
95029330 Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, Microtiter, Flat-bottom, EB, High Flange, 380μL Case of 50
244.80
Online Exclusive
272.00
Save 27.20 (10%)
460984 Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, 400μL, MaxiSorp, certified, bulk Case of 180
1,139.00
Online Exclusive
1,340.00
Save 201.00 (15%)
468667 Immuno Clear Standard Modules, F8, MaxiSorp, framed, certified Case of 60
578.70
Online Exclusive
643.00
Save 64.30 (10%)
437591 Immuno Standard Modules, MaxiSorp, white Case of 60
717.30
Online Exclusive
797.00
Save 79.70 (10%)
467466 Immuno Clear Standard Modules, F16, MaxiSorp, framed Case of 60
580.50
Online Exclusive
645.00
Save 64.50 (10%)
449824 Clear Round-Bottom Immuno Nonsterile 96-Well Plates, MaxiSorp Case of 60
324.90
Online Exclusive
361.00
Save 36.10 (10%)
446612 Clear C-Shaped Immuno Nonsterile 96-Well Plates, MaxiSorp, certified Case of 60
367.20
Online Exclusive
408.00
Save 40.80 (10%)
446639 Immuno Breakable Modules Clear, Nunc; U8 LockWell; MaxiSorp; 320μL Case of 60 -
469264 Immuno Clear Standard Modules, U16, MaxiSorp Case of 320
411.30
Online Exclusive
457.00
Save 45.70 (10%)
437796 Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates, white Case of 60
359.20
Online Exclusive
449.00
Save 89.80 (20%)
6405 Immuno Clear Standard Modules, F12, 4 HBX, framed Case of 100
781.20
Online Exclusive
868.00
Save 86.80 (10%)
6404TS Immuno Clear Standard Modules, Max. working volume: 330μL Case of 320
253.80
Online Exclusive
282.00
Save 28.20 (10%)
439454 Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, 400μL, MaxiSorp, certified Case of 60
346.80
Online Exclusive
408.00
Save 61.20 (15%)
475515 Immuno Standard Modules, MaxiSorp, black Case of 60
684.00
Online Exclusive
760.00
Save 76.00 (10%)
3855 Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, 330μL, 4 HBX Case of 50
252.90
Online Exclusive
281.00
Save 28.10 (10%)
437111 Black 96-Well Immuno Plates, 400μL, Maxisorp Case of 80
497.70
Online Exclusive
553.00
Save 55.30 (10%)
6508 FTG,1⁄16 TUBE NUT⁄FER PEEK 1 unit
170.00
430341 Clear C-Shaped Immuno Nonsterile 96-Well Plates, MaxiSorp Case of 60
324.90
Online Exclusive
361.00
Save 36.10 (10%)
15031 Pierce™ 8-Well Polystyrene Strip Plates, Corner Notch Each
920.00
437915 Immuno Standard Modules, MaxiSorp, clear Case of 60
765.00
Online Exclusive
850.00
Save 85.00 (10%)
464718 Clear Flat-Bottom Immuno Nonsterile 384-Well Plates, MaxiSorp Case of 30
236.70
Online Exclusive
263.00
Save 26.30 (10%)
95029140 Strip Assembly, 1 x 12, EB Case of 50
430.20
Online Exclusive
478.00
Save 47.80 (10%)
456537 Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, 400μL, MaxiSorp, certified Case of 180
1,098.00
Online Exclusive
1,220.00
Save 122.00 (10%)
475078 Immuno Clear Standard Modules, U8, MaxiSorp, framed Case of 60
548.25
Online Exclusive
645.00
Save 96.75 (15%)
460518 384-Well Immuno Plates, Black Case of 30
281.70
Online Exclusive
313.00
Save 31.30 (10%)
95029100 Immuno Clear Standard Modules, F8, EB, framed Case of 50
423.00
Online Exclusive
470.00
Save 47.00 (10%)
469949 Immuno Clear Standard Modules, F8, MaxiSorp Case of 640
406.33
Online Exclusive
436.00
Save 29.67 (7%)
446471 Immuno Breakable Modules, MaxiSorp, black Case of 60
522.00
Online Exclusive
580.00
Save 58.00 (10%)
460372 384-Well Immuno Plates, White Case of 30
281.70
Online Exclusive
313.00
Save 31.30 (10%)
473709 Immuno Clear Standard Modules, C12, MaxiSorp, framed Case of 60
603.90
Online Exclusive
671.00
Save 67.10 (10%)
441653 Immuno Clear Standard Modules, C8 StarWell, MaxiSorp, framed Case of 60
594.42
Online Exclusive
722.00
Save 127.58 (18%)
445101 Immuno Clear Standard Modules, C8, MaxiSorp, framed Case of 60
580.50
Online Exclusive
645.00
Save 64.50 (10%)
463201 Immuno Breakable Modules, MaxiSorp, white Case of 60
526.50
Online Exclusive
585.00
Save 58.50 (10%)
473768 Immuno Breakable Modules Clear, C8 BreakApart, MaxiSorp Case of 60
647.97
Online Exclusive
949.00
Save 301.03 (32%)
446469 Immuno Breakable Modules Clear, C8 LockWell, MaxiSorp Case of 60
561.60
Online Exclusive
624.00
Save 62.40 (10%)
442404 Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, 400μL, MaxiSorp Case of 60
324.90
Online Exclusive
361.00
Save 36.10 (10%)
436110 White 96-Well Immuno Plates, Maxisorp, Flat-Bottom, MaxiSorp, 350μL Case of 80
495.00
Online Exclusive
550.00
Save 55.00 (10%)
95029180 Immuno Breakable Modules Clear Case of 50
433.80
Online Exclusive
482.00
Save 48.20 (10%)
Very hydrophilic:
This is the most hydrophilic surface in the portfolio, which many hydrophilic proteins will bind to with a high affinity. The most commonly-used molecules that will bind to these surfaces are glycans. It is also suggested to use this surface for the immobilization of highly polar molecules or for homogenous assays of hydrophobic analytes which will not adhere to the hydrophobic surface.
Binding to the very hydrophilic surface tends to be more pH sensitive. It is important to consider the molecule orientation as this may be controlled to some extent by the buffer composition and pH, so that when large, complex molecules are physically adsorbed, the orientation could be random so that not all the absorbed molecules will be capable of binding the analyte and no signal will be seen.
Catalog # Name Size Price (USD) Qty
467340 Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, 400μL, MultiSorp Case of 60
370.60
Online Exclusive
436.00
Save 65.40 (15%)
467140 Immuno Clear Standard Modules, F8, MultiSorp, framed Case of 60
620.10
Online Exclusive
689.00
Save 68.90 (10%)
446490 Immuno Breakable Modules Clear, C8 LockWell, MultiSorp Case of 60
426.20
Online Exclusive
624.00
Save 197.80 (32%)
For Research Use Only. Not for use in diagnostic procedures.