The dyes in this section are all amphiphilic probes—molecules that comprise a charged fluorophore that localizes the probe at the membrane's surface and lipophilic aliphatic "tails" that insert into the membrane and thus anchor the probe to the membrane. In addition to labeling model membranes, most of these probes are very useful for cell tracing applications (Tracers for Membrane Labeling—Section 14.4). Summary of our lipophilic carbocyanine and aminostyryl tracers—Table 14.3 lists all of our lipophilic carbocyanine and aminostyryl tracers and compares their properties and uses. Our FM dyes, which are also amphiphilic styryl dyes but with less lipophilic character than the dyes in this section, are particularly useful for labeling membranes of live cells and for following synaptosome recycling (Probes for Following Receptor Binding and Phagocytosis—Section 16.1).
Dialkylcarbocyanine Probes
Carbocyanines are among the most strongly light-absorbing dyes known and have proven to be useful tools in several different areas of research. Carbocyanines with short alkyl tails attached to the imine nitrogens are employed both as membrane-potential sensors (Slow-Response Probes—Section 22.3) and as organelle stains for mitochondria and the endoplasmic reticulum (Probes for Mitochondria—Section 12.2, Probes for the Endoplasmic Reticulum and Golgi Apparatus—Section 12.4). Those with longer alkyl tails (≥12 carbons) have an overall lipophilic character that makes them useful for neuronal tracing ref and long-term labeling of cells in culture ref (Tracers for Membrane Labeling—Section 14.4), as well as for noncovalent labeling of lipoproteins (Probes for Following Receptor Binding and Phagocytosis—Section 16.1). This section describes the use and properties of dialkylcarbocyanines as general-purpose probes of membrane structure and dynamics.
DiI, DiO, DiD, DiR and Analogs
The most widely used carbocyanine membrane probes have been the octadecyl (C18) indocarbocyanines (D282, D3911) and oxacarbocyanines (D275) often referred to by the generic acronyms DiI and DiO, or more specifically as DiIC18(3) and DiOC18(3), where the subscript is the number of carbon atoms in each alkyl tail and the bracketed numeral is the number of carbon atoms in the bridge between the indoline or benzoxazole ring systems. We also offer several variations on these basic structures (Tracers for Membrane Labeling—Section 14.4, Summary of our lipophilic carbocyanine and aminostyryl tracers—Table 14.3):
  • DiI and DiO analogs with unsaturated alkyl tails (FAST DiO, D3898; FAST DiI, D3899, D7756)
  • DiI and DiO analogs with shorter alkyl tails (DiIC12(3), D383; DiIC16(3), D384)
  • Long-wavelength light–excitable carbocyanines (DiD, D307, D7757)
  • Infrared light–excitable carbocyanine (DiR, D12731)
  • Chloromethylbenzamido DiI and sulfonated DiI and DiO derivatives (see below)
Spectral Properties of Dialkylcarbocyanines
The spectral properties of dialkylcarbocyanines are largely independent of the lengths of the alkyl chains, and are instead determined by the heteroatoms in the terminal ring systems and the length of the connecting bridge. The DiICn(3) probes have absorption and fluorescence spectra compatible with rhodamine (TRITC) optical filter sets, whereas DiOCn(3) analogs can be used with fluorescein (FITC) optical filter sets. The emission maxima of DiIC18(3) and DiOC18(3) incorporated in dioctadecenoylphosphocholine (dioleoyl PC or DOPC) liposomes (Figure 13.4.1) are similar to those of the dyes in methanol.
The very large molar extinction coefficients of carbocyanine fluorophores are their most outstanding spectral property. Their fluorescence quantum yields are only modest—about 0.07 for DiI in methanol and about three-times greater in amphiphilic solvents such as octanol.ref Their fluorescence in water is quite weak.ref The excited-state lifetimes of carbocyanine fluorophores in lipid environments are short (~1 nanosecond), which is an advantage for flow cytometry applications because it allows more excitation/de-excitation cycles during flow transit; the overall decay is multi-exponential.ref Dialkylcarbocyanines are also exceptionally photostable.ref
The red He-Ne laser–excitable indodicarbocyanines such as DiD (DiIC18(5); D307, D7757) have long-wavelength absorption and red emission (Figure 13.4.1). Their extinction coefficients are somewhat larger and fluorescence quantum yields much larger than those of carbocyanines such as DiI.ref Moreover, photoexcitation of DiD seems to cause less collateral damage than photoexcitation of DiI in live cells.ref The DiIC18(7) tricarbocyanine probe (DiR, D12731) has excitation and emission in the infrared, which may make the dye useful as an in vivo tracer for labeled cells and liposomes in live organisms.ref
dialkylcarbocyanine.par.12267.image.275.257.1.s000980-normalized-fluorescence-gif
Figure 13.4.1 Normalized fluorescence emission spectra of DiO (D275), DiI (D282), DiD (D307) and DiR (D12731) bound to phospholipid bilayer membranes.
Substituted DiI and DiO Derivatives
We have synthesized various derivatives of DiI, DiO and DiD. All of these derivatives have octadecyl (C18) tails identical to those of DiI (D282D3911) and DiO (D275), thereby preserving the excellent membrane retention characteristics of the parent molecules. A variety of substitutions have been made on the indoline or benzoxazole ring systems:
  • Chloromethylbenzamido DiI derivatives (CellTracker CM-DiI; C7000C7001)
  • Anionic sulfophenyl derivatives of DiI and DiO ref (SP-DiIC18(3), D7777; SP-DiOC18(3), D7778)
  • Sulfonate derivatives of DiI and DiD (DiIC18(3)-DS; DiIC18(5)-DS, D12730)
Although these derivatives have primarily been developed to provide improved fixation and labeling in long-term cell tracing applications (Tracers for Membrane Labeling—Section 14.4), they also offer several features that can potentially be exploited for investigating membrane structure and dynamics. For researchers wishing to carry out comparative evaluations, our Lipophilic Tracer Sampler Kit (L7781) provides 1 mg samples of each of nine different carbocyanine derivatives, including several of the newer substituted derivatives:
  • DiI (DiIC18(3))
  • DiD (DiIC18(5))
  • DiR (DiIC18(7))
  • DiO (DiOC18(3))
  • DiA (4-Di-16-ASP)
  • DiIC18(3)-DS
  • SP-DiIC18(3)
  • SP-DiOC18(3)
  • 5,5'-Ph2-DiIC18(3)
The fluorescence quantum yields of the sulfophenyl and phenyl derivatives (measured in methanol) are generally two- to threefold greater than those of DiI and DiO. In particular, we have found that the sulfophenyl derivatives (SP-DiIC18(3), D7777; SP-DiOC18(3), D7778) bound to phospholipid model membranes have approximately fivefold higher quantum yields than DiI and DiO. DiIC18(5)-DS (D12730) has been used in combination with an NBD-labeled glycerophosphoserine probe in a novel resonance energy transfer assay that detects inner monolayer membrane hemifusion, avoiding erroneous indications of membrane fusion due to lipid mixing and other environmental effects in the outer monolayer.ref The negative charge and greater water solubility of the sulfonated carbocyanines results in modified lateral and transverse distributions of these probes in lipid bilayers relative to those of DiI and DiO. This characteristic has been exploited to identify plasma membrane lipid domains that are responsive to electrical stimulation of outer hair cells in the inner ear.ref
DiI and DiO as Probes of Membrane Structure
The orientation of DiIC18(3) in membranes has been determined by fluorescence polarization microscopy.ref The long axis of the fluorophore is parallel to the membrane surface, and the two alkyl chains protrude perpendicularly into the lipid interior (Figure 13.2.1D in Fatty Acid Analogs and Phospholipids—Section 13.2). There are conflicting reports in the literature regarding the ease of transbilayer migration ("flip-flop") of lipophilic indocarbocyanines.ref The lateral partitioning behavior of dialkylindocarbocyanines in membranes has been investigated by fluorescence recovery after photobleaching (FRAP),ref calorimetry,ref lifetime measurements ref and fluorescence resonance energy transfer techniques ref (Fluorescence Resonance Energy Transfer (FRET)—Note 1.2). These studies demonstrate that the probe distribution between coexisting fluid and gel phases depends on the similarity of the alkyl chain lengths of the probe and the lipid. In general, the more dissimilar the lengths, the greater the preference for fluid-phase over gel-phase lipids. For example, the shorter-chain DiIC12(3) has a substantial preference for the fluid phase (~6:1) in DOPC, whereas DiIC18(3) is predominantly distributed in the gel phase ref (~1:10). Consequently, long-chain dialkylcarbocyanines are among the best probes for detecting particularly rigid gel phases.
Lipophilic carbocyanines have been used to visualize membrane fusion and cell permeabilization that occurs in response to electric fields,ref as well as fusion of liposomes with planar bilayers.ref Membrane fusion can also be measured by fluorescence resonance energy transfer to DiIC18(3) from dansyl- or NBD-labeled phospholipid donors ref or by direct imaging.ref In Langmuir–Blodgett films, excited-state energy transfer from DiIC18(3) to DiIC18(5) is exceptionally efficient because of the favorable orientations of the fluorophores.ref Energy transfer from DiIC18(5) to DiIC18(7) should be similarly effective. Lipophilic carbocyanines have also been used to elicit photosensitized destabilization of liposomes,ref to sensitize photoaffinity labeling of the viral glycoprotein hemagglutinin,ref to image membrane domains in lipid monolayers ref and to develop a fiber-optic potassium sensor.ref
DiI and DiO as Probes of Membrane Dynamics
Despite their reasonably good photostability, dialkylcarbocyanines are widely employed to measure lateral diffusion processes using fluorescence recovery after photobleaching (FRAP) techniques.ref Their lateral diffusion coefficients in isolated fluid- and gel-phase bilayers are independent of the carbocyanine alkyl chain length.ref Phase-separated populations of lipophilic carbocyanine dyes can be distinguished by their diffusion rates and can therefore be used to define lateral domains in cell membranes.ref Combined lateral diffusion measurements of labeled proteins and lipids have demonstrated that transformed ref and permeabilized ref cells show marked changes in protein diffusion whereas lipid diffusion rates remain unchanged. In other cases, coupling of lipid and protein mobility has been identified in the form of relatively immobilized lipid domains in yeast plasma membranes ref and around IgE receptor complexes.ref A different photobleaching technique, which depends on the absence of diffusional fluorescence recovery, was employed to determine lipid flow direction in locomoting cells by following the movement of a photobleached stripe of DiIC16(3) ref (D384).
(Top)
Dialkylaminostyryl Probes
The lipophilic aminostyryl probes 4-Di-10-ASP, DiA (4-Di-16-ASP, D3883) and FAST DiA insert in membranes with their two alkyl tails and their fluorophore oriented parallel to the phospholipid acyl chains ref (Figure 13.2.1H in Fatty Acid Analogs and Phospholipids—Section 13.2). When these dialkylaminostyryl probes bind to membranes, they exhibit a strong fluorescence enhancement; their fluorescence in water is minimal. The interfacial solvation of the aminostyryl fluorophore causes a large blue shift of the absorption spectrum of the membrane-bound probe.ref For example, the absorption maximum of DiA is 456 nm when incorporated into DOPC liposomes and 490 nm when in methanol. The fluorescence emission maximum of DiA in the membrane environment is 590 nm, which is quite close to that observed for probes with shorter alkyl tails such as 4-Di-10-ASP;ref however, the fluorescence spectrum of DiA is very broad, with appreciable intensity from about 510 nm to 690 nm. Consequently, DiA can be detected as green, orange or even red fluorescence, depending on the optical filter employed. Like the lipophilic carbocyanines, DiA is commonly used for neuronal membrane tracing (Tracers for Membrane Labeling—Section 14.4). FAST DiA, the diunsaturated analog of DiA, is intended to facilitate these studies by accelerating dye diffusion within the membrane.
The FM 1-43, FM 1-43FX, FM 4-64 and FM 5-95 dyes, which are discussed in detail in Probes for Following Receptor Binding and Phagocytosis—Section 16.1, are styryl dyes that also exhibit high Stokes shifts and broad fluorescence emission but have less lipophilic character than the 4-Di-10-ASP and DiA probes. The FM dyes are commonly used to define the outer membranes of liposomes and live cells and to detect synaptosome recycling.
Data Table
For a detailed explanation of column headings, see Definitions of Data Table Contents
Cat #MWStorageSolubleAbsECEmSolventNotes
C7000
CellTracker CM-DiI
1051.50F,D,LDMSO, EtOH553134,000570MeOH 
C7001
CellTracker CM-DiI
1051.50F,D,LDMSO, EtOH553134,000570MeOH 
D275
DiO
881.72LDMSO, DMF484154,000501MeOH 
D282
DiI
933.88LDMSO, EtOH549148,000565MeOH 
4-Di-10-ASP618.73LDMSO, EtOH49253,000612MeOH1
D307
DiD
959.92LDMSO, EtOH644260,000665MeOH2
D383
DiIC12(3)
765.56LDMSO, EtOH549144,000565MeOH3
D384
DiIC16(3)
877.77LDMSO, EtOH549148,000565MeOH 
DiOC16(3)
825.61LDMSO, DMF484156,000501MeOH 
D3883
DiA
787.05LDMSO, EtOH49152,000613MeOH1
Δ9-DiI925.49F,L,AADMSO, EtOH549144,000564MeOH2
D3898
FAST DiO solid
873.65F,L,AADMSO, DMF484138,000499MeOH 
D3899
FAST DiI oil
925.82F,L,AADMSO, EtOH549143,000564MeOH2
D3911
DiI
933.88LDMSO, EtOH549148,000565MeOH 
D7756
FAST DiI solid
1017.97F,L,AADMSO, EtOH549148,000564MeOH 
D7757
DiD
1052.08LDMSO, EtOH644193,000663MeOH 
FAST DiA solid899.80F,L,AADMSO, EtOH49241,000612MeOH1
DiIC18(3)-DS993.54LDMSO, EtOH555144,000570MeOH 
D7777
SP-DiIC18(3)
1145.73LDMSO, EtOH556164,000573MeOH 
D7778
SP-DiOC18(3)
1115.55LDMSO, EtOH497175,000513MeOH 
D12730
DiIC18(5)-DS
1019.58LDMSO, EtOH650247,000670MeOH 
D12731
DiR
1013.41LDMSO, EtOH748270,000780MeOH 
  1. Abs and Em of styryl dyes are at shorter wavelengths in membrane environments than in reference solvents such as methanol. The difference is typically 20 nm for absorption and 80 nm for emission, but varies considerably from one dye to another. Styryl dyes are generally nonfluorescent in water.
  2. This product is intrinsically a liquid or an oil at room temperature.
  3. This product is intrinsically a sticky gum at room temperature.
Ordering Information
Catalog # Name Size Price (USD) Qty
C7001 CellTracker™ CM-DiI Dye, 1mg Each
349.65
Online exclusive
363.00
Save 13.35 (4%)
C7000 CellTracker™ CM-DiI Dye, 20 x 50μg Each
395.65
Online exclusive
408.00
Save 12.35 (3%)
D3883 DiA; 4-Di-16-ASP (4-(4-(Dihexadecylamino)styryl)-N-Methylpyridinium Iodide) Each
339.00
D291 4-Di-10-ASP (4-(4-(Didecylamino)styryl)-N-Methylpyridinium Iodide) 25 mg -
D307 DiD' oil; DiIC18(5) oil (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindodicarbocyanine Perchlorate) Each
287.00
D7757 DiD' solid; DiIC18(5) solid (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindodicarbocyanine, 4-Chlorobenzenesulfonate Salt) Each
222.65
Online exclusive
230.00
Save 7.35 (3%)
D3886 Δ9-DiI (1,1'-Dioleyl-3,3,3',3'-Tetramethylindocarbocyanine methanesulfonate) 25 mg -
D282 Dil Stain (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindocarbocyanine Perchlorate ('DiI'; DiIC18(3))), 100 mg Each
387.65
Online exclusive
399.00
Save 11.35 (3%)
D3911 Dil Stain (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindocarbocyanine Perchlorate ('DiI'; DiIC18(3))), 25 mg Each
317.65
Online exclusive
327.00
Save 9.35 (3%)
D383 DiIC12(3) (1,1'-Didodecyl-3,3,3',3'-Tetramethylindocarbocyanine Perchlorate) Each
403.00
D384 DiIC16(3) (1,1'-Dihexadecyl-3,3,3',3'-Tetramethylindocarbocyanine Perchlorate) Each
394.00
D7776 DiIC18(3)-DS) 1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindocarbocyanine-5,5'-Disulfonic Acid) 5 mg -
D12730 DiIC18(5)-DS (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindodicarbocyanine-5,5'-Disulfonic Acid) Each
397.00
D7758 FAST DiA™ solid; DiΔ9,12-C18ASP, CBS (4-(4-(Dilinoleylamino)styryl)-N-Methylpyridinium 4-Chlorobenzenesulfonate) 5 mg -
D3898 FAST DiO™ Solid; DiOΔ9,12-C18(3), ClO4 (3,3'-Dilinoleyloxacarbocyanine Perchlorate) Each
394.00
D7756 FAST DiI™ solid; DiIΔ9,12-C18(3), CBS (1,1'-Dilinoleyl-3,3,3',3'-Tetramethylindocarbocyanine, 4-Chlorobenzenesulfonate) Each
395.00
D3899 FAST DiI™ oil; DiIΔ9,12-C18(3), ClO4 (1,1'-Dilinoleyl-3,3,3',3'-Tetramethylindocarbocyanine Perchlorate) Each
394.00
D275 DiO'; DiOC18(3) (3,3'-Dioctadecyloxacarbocyanine Perchlorate) Each
402.00
D1125 DiOC16(3) (3,3'-Dihexadecyloxacarbocyanine Perchlorate) 25 mg
399.00
D7778 SP-DiOC18(3) (3,3'-Dioctadecyl-5,5'-Di(4-Sulfophenyl)Oxacarbocyanine, Sodium Salt) Each
407.00
D7777 SP-DiIC18(3) (1,1'-Dioctadecyl-6,6'-Di(4-Sulfophenyl)-3,3,3',3'-Tetramethylindocarbocyanine) Each
397.00
D12731 DiR'; DiIC18(7) (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindotricarbocyanine Iodide) Each
304.65
Online exclusive
314.00
Save 9.35 (3%)
L7781 Lipophilic Tracer Sampler Kit Each
444.00
For Research Use Only. Not for use in diagnostic procedures.