Organism TypeMouseEmbryonic mouse fibroblast; Properties: contact inhibition; transformation; transfection; cell biology; differentiation to adipocytes. Susceptible to: murine leukemia virus, murine sarcoma virus, vesicular stomatitis, vaccinia, herpes simplex, N-tropic oncornaviruses C
TissuesEmbryo
PhenotypeAdherent
Primaryno
ApplicationCell Culture / Growth Conditions,Protein Expression,RNAi,Transient Transfection
Products
Catalog # Name Size Price (USD) Qty
11668019 Lipofectamine™ 2000 Transfection Reagent, 1.5 mL Each
842.65
Online Exclusive
924.00
Save 81.35 (9%)
11668027 Lipofectamine™ 2000 Transfection Reagent, 0.75 mL Each
545.65
Online Exclusive
590.00
Save 44.35 (8%)
11965084 DMEM, high glucose, 1000 mL Each
55.65
Online Exclusive
60.75
Save 5.10 (8%)
11965092 DMEM, high glucose, 500 mL Each
34.65
Online Exclusive
37.14
Save 2.49 (7%)
11965118 DMEM, high glucose, 10 x 500 mL Each
291.65
Online Exclusive
320.00
Save 28.35 (9%)
11965126 DMEM, high glucose, 6 x 1000 mL Each
280.65
Online Exclusive
308.00
Save 27.35 (9%)
15596018 TRIzol™ Reagent, 200 mL Each
493.00
21700018 Ham's F-12 Nutrient Mix, powder, 50 L Each
220.65
Online Exclusive
234.00
Save 13.35 (6%)
K494500 BLOCK-iT™ U6 RNAi Entry Vector Kit Each
1,018.00
R25001 Zeocin™ Selection Reagent, 8 x 1.25 mL Each
343.65
Online Exclusive
380.00
Save 36.35 (10%)
R25005 Zeocin™ Selection Reagent, 50 mL Each
1,256.65
Online Exclusive
1,390.00
Save 133.35 (10%)
V79020 pcDNA™3.1 (+) Mammalian Expression Vector 20 µg
1,110.00
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.