Paternity-and-kinship-solutions
Trusted, integrated systems for paternity and kinship analysis
As the worldwide leader in DNA typing solutions, Thermo Fisher Scientific delivers the most comprehensively validated products, expertise and application support available to paternity testing laboratories. Precise results, consistent performance and proven success, sample after sample, are what you expect from us.
Get a quote
Applied Biosystems direct PCR amplification kits combine a high power of discrimination with superior performance and fast time to results, meaning you can get accurate results from your paternity and kinship samples in less than two hours.
Verifiler Globalfiler workflow graphic
 Click image to enlargeSimple and accurate workflow with the Applied Biosystems family of paternity STR kits
Speed up your workflow, expand your throughput
Prep-n-Go Buffer and Prep-n-Go Buffer for Buccal Swabs
The Prep-n-Go Buffer and Prep-n-Go Buffer for Buccal Swabs enable high quality direct PCR amplification of non-FTA reference and single source samples without the need for heat incubation, extraction or purification. Increase your STR profile quality with fewer intermediate steps.
Increased efficiency and discriminatory power
Amplification kits for paternity testing
Direct STR amplification kits offer improved efficiency by eliminating the need for DNA extraction and quantification.
A32014-10in
VeriFiler Express PCR Amplification Kit
For paternity customers who demand high confidence in relationship testing conclusions, the new 6-dye VeriFiler Express PCR Amplification Kit—with 25 STR markers including two highly discriminating Penta markers—enables superior genotyping results backed by superior service and support.
COL12342-4479567-VeriFiler-Direct-PCR-Amplification-Kit
GlobalFiler Express PCR Amplification Kit
As part of the only fully integrated and validated forensic workflow, this 6-dye, 24-locus technology is designed to deliver superior lab performance—all facilitated by high quality manufacturing at our ISO18385 certified facility, along with comprehensive training, service, and support.
Same STR workflow, same trusted CE technology
SeqStudio-Genetic-Analyzer-R5-JPEGs-00211024_5-(1)
Now with an innovative all-in-one cartridge that helps reduce run setup time from hours to minutes
The Applied Biosystems SeqStudio Genetic Analyzer for HID is a 4-capillary, easy-to-use, and convenient benchtop system that delivers gold-standard STR fragment analysis with just one click.  The SeqStudio instrument uses an innovative all-in-one cartridge. Load up your samples and cathode buffer, click in the cartridge, and go. The cartridge is stable for up to six months on the instrument and can process 250 injections (1000 samples). The SeqStudio Genetic Analyzer for HID is optimized for performance with Applied Biosystems STR kits and incorporates a resolution algorithm and a marker-to-marker pull-up reduction that is optimized for the markers in each kit.
Get the same answers you trust with the outstanding data quality, service, and support you’ve come to expect from Applied Biosystems instruments.
By combining Applied Biosystems 3500 Data Collection Software v4.0 or SeqStudio Data Collection Software v 1.2 with Applied Biosystems STR kits, GeneMapper ID-X software helps significantly enhance productivity and confidence, as well as IT support and compliance, in your paternity laboratory’s data interpretation workflow. These software solutions offer the following benefits:
Improved data interpretation—reduced pull-up edits with Applied Biosystems STR kits, Improved first-pass success rate for labs processing single source samples with off-scale data recovery
Enhanced user experience—flexible plate loading, streamlined workflow, support languages, 6-dye installation standard
IT support and compliance—Windows 10 support, internationalization
COL23060-D10NOOSRsamescale
 Click image to enlargeEnhanced dynamic range for off-scale data recovery
Complete analysis solution for kinship and paternity testing
Converge Analysis Software
Utilising validated algorithms, Converge software’s Kinship and Paternity module offers laboratories the ability to conduct various types of relationship testing in a fraction of the time as compared to current methods.
Request your demo at thermofisher.com/converge ›
Image from COL31723
 Click image to enlargeKinship and paternity analysis made simple with Applied Biosystems Converge Software
Make the right purchase for your lab
Catalog # Name Size Price (USD) Qty
A32607 NUCLEIC-CARD™ Collection Device System Each
497.00
4408399 GeneScan™ 600 LIZ™ dye Size Standard v2.0 Each
704.00
4473973 NUCLEIC-CARD™ matrix, 1 spot, 100 Cards Each
546.00
4473974 NUCLEIC-CARD™ COLOR matrix, 1 spot Each
350.00
4467079 Prep-n-Go™ Buffer (5 tubes) Each
8,560.00
4441352 PrepFiler Express™ Forensic DNA Extraction Kit Each
768.00
4441351 PrepFiler Express BTA™ Forensic DNA Extraction Kit Each
826.00
A32014 VeriFiler™ Express PCR Amplification Kit Each
4,600.00
A32070 VeriFiler™ Express PCR Amplification Kit with Prep-n-Go™ Buffer (5 tubes) Each
5,280.00
A33032 VeriFiler™ Express PCR Amplification Kit with Prep-n-Go™ Buffer (for buccal swabs) Each
5,690.00
4476609 GlobalFiler™ Express PCR Amplification Kit, 200 reactions Each
5,210.00
4479649 GlobalFiler™ Express PCR Amplification Kit and Prep-n-Go™ Buffer (for buccal swabs), 200 reactions Each
5,990.00
A31832 NGM Detect™ PCR Amplification Kit Each
6,580.00
4425042 DS-36 Matrix Standard (Dye Set J6) Each
269.00
A46228 SeqStudio™ Genetic Analyzer for HID, laptop, with training Each -
A46229 SeqStudio™ Genetic Analyzer for HID, desktop, with training Each -
A46227 SeqStudio™ Genetic Analyzer for HID, with training Each -
A39975 GeneMapper™ ID-X Software v1.6, full installation Each
42,040.00
A39976 GeneMapper™ ID-X Software v1.6, client installation, 1 License Each
25,080.00
A35131 Converge™ Software Each -
A31001 Converge™ Software Case Management License and Kinship & Paternity Analysis License, 1 user, 1-year license Each
4,210.00
A31002 Converge™ Software Case Management License and Kinship & Paternity Analysis License, 1 user, 3-year license Each
8,420.00