Brilliant Violet 786 excitation shown as dotted line and emission shown as solid red histogram
3405780/60, 750 LP406788(in buffer) 3
 flow cytometry
Invitrogen Brilliant Violet™ 786 (BV786) is a tandem dye excited by the 405 nm violet laser and emits at 786 nm. This dye has a medium relative brightness and can be used for cell surface, intracellular, and nuclear staining.
When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye- conjugated antibodies in a staining panel, we recommend using Super Bright Complete Staining Buffer or Brilliant Stain Buffer to minimize any non-specific polymer interactions.
We offer BV786 dye conjugated to primary antibodies for use in flow cytometry. 
Search Brilliant Violet™ 786 primary antibodies
Spectral signature of Brilliant Violet™ 786 dye. Data acquired on a 5-laser Cytek Aurora and normal human peripheral blood cells stained with CD8a Monoclonal Antibody, Brilliant Violet™ 786 were used for analysis.
Dot plot of Ki-67 BV786 vs CD45R (B220) FITC in unstimulated and stimulated cells
Intracellular staining of mouse splenocytes using Brilliant Violet™ 786. C57BL/6 mouse splenocytes were unstimulated (left) or stimulated for 48 hours with CD3e Monoclonal Antibody, Functional Grade (right). Cells were then surface-stained with CD45R (B220) Monoclonal Antibody, FITC and stained intracellularly, using the Foxp3/Transcription Factor Staining Buffer Set and protocol, with 1.0 µg of Ki-67 Monoclonal Antibody (SolA15), Brilliant Violet™ 786. Viable cells in the lymphocyte gate were used for analysis, as determined by LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation.
Dot plot of samples stained with IgG2a control vs CD4-FITC and CD8a-BV786 vs CD4-FITC
Cell surface staining of mouse splenocytes using Brilliant Violet™ 786. C57BL/6 mouse splenocytes were stained with CD4 Monoclonal Antibody, FITC and 0.25 µg of Rat IgG2a kappa Isotype Control, Brilliant Violet™ 786 (left) or 0.25 µg of CD8a Monoclonal Antibody, Brilliant Violet™ 786 (right). Viable cells in the lymphocyte gate were used for analysis, as determined by 7-AAD Viability Staining Solution.
Brilliant Ultra Violet™ dye background
Brilliant Ultra Violet dyes are a polymer dye-based technology and compatible with both spectral flow cytometry as well as traditional flow cytometry. The UV laser has unique channels far from heavily occupied detectors, allowing for larger panels.
Additional resources
Flow cytometry protocols handbook
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
Request the handbook
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.