The Drama Is In the Details
The analysis of cell proliferation is crucial for cell differentiation and cancer studies and is commonly used for evaluating compound toxicity in the drug development process. Tools for measuring cell proliferation include probes for analyzing the average DNA content and cellular metabolism in a population, as well as single-cell indicators of DNA synthesis and cell cycle–specific proteins.
Here we describe a number of assays that each provide a unique window through which to view the process of cell proliferation (Tables 1A–1E). These reagents can be used individually or together to form the basis of an assay for cell proliferation, cytotoxicity, or drug efficacy.
Determining Cell Proliferation Based on DNA Content
The CyQUANT® cell proliferation assay provides a fluorescence microplate–based method for accurately counting cells in a population, based on cellular DNA content. Because cellular DNA content is highly regulated, the CyQUANT® assay can be used at multiple time points to calculate the average proliferation rate of a cell population. Determination of cell number with the CyQUANT® assay is also a highly sensitive indicator of cytotoxicity.
Three generations of CyQUANT® assays (Table 1A) provide options for different workflows: the original CyQUANT® Kit is designed for multi-day and endpoint assays; the CyQUANT® NF Kit provides a live-cell assay; and the CyQUANT® Direct Kit (Figure 1) provides a live-cell assay configured with a background suppression dye. Binding of CyQUANT® dye to DNA is independent of metabolic state, producing consistent signal windows and fluorescence intensities across a range of conditions and cell types.
Linearity and sensitivity of the CyQUANT® Direct assay 
Figure 1. Linearity and sensitivity of the CyQUANT® Direct assay. CHO cells were plated at densities of 0–20,000 per well in a 384-well microplate. Cells were labeled with the CyQUANT® Direct dye according to the microplate protocol. Fluorescence intensities, measured with a fluorescence microplate reader using a FITC filter set, varied linearly with respect to cell number between 40 and 20,000 cells. The inset shows the measurement range from 0 to 1,250 cells/well. The results shown represent averages of eight experiments per data point.
Table 1A. Determining Cell Proliferation Based on Total DNA Content.
Kit or reagent Ex/Em (Abs) * Stains live cells Stains fixed cells Instrument platform † Assay features Cat. No.
CyQUANT® Direct Cell Proliferation Assay Kit
508/527
Yes
M
An add-and-read, lysis-free cell proliferation assay designed for use with multi-well plates, making it ideal for high-throughput screening applications.
C35011
C35012
CyQUANT® NF Cell Proliferation Assay Kit
480/520
Yes
Yes
M
Live-cell assay using a cell-permeant DNA stain that eliminates the need for cell lysis and the freeze/thaw step.
C35007
C35006
CyQUANT® Cell Proliferation Assay Kit
480/520
Yes
M
Run multi-day and endpoint assays with the same set of standards using this frozen assay format.
C7026
* Fluorescence excitation and emission maxima in nm, with absorption maximum in parentheses where applicable. † FC = flow cytometry. FM = fluorescence microscopy. HCS = high-content screening and analysis. M = microplate assay.
Determining cell proliferation based on total DNA content
Catalog # Name Size Price (USD) Qty
C35011 CyQUANT™ Direct Cell Proliferation Assay, 10 microplates kit Each
546.65
Online Exclusive
568.00
Save 21.35 (4%)
C35012 CyQUANT™ Direct Cell Proliferation Assay, 100 microplates kit Each
3,630.65
Online Exclusive
3,790.00
Save 159.35 (4%)
C35007 CyQUANT™ NF Cell Proliferation Assay, 200 assays kit Each
205.65
Online Exclusive
212.00
Save 6.35 (3%)
C35006 CyQUANT™ NF Cell Proliferation Assay, 1,000 assays Each
529.65
Online Exclusive
546.00
Save 16.35 (3%)
C7026 CyQUANT™ Cell Proliferation Assay, for cells in culture Each
469.65
Online Exclusive
486.00
Save 16.35 (3%)
Determining Cell Proliferation Based on Metabolic Activity
The PrestoBlue™ and alamarBlue® reagents (Table 1B) are used for fluorescence- or absorption-based microplate assays that measure the reductive capacity of cells (Figure 2). These cell-permeant, resazurin-based compounds are blue in color and virtually nonfluorescent. When added directly to cells, resazurin is converted to resorufin by the reducing environment of a viable cell, turning red in color and becoming highly fluorescent. Damaged and nonviable cells—which are also likely to be non-proliferating cells—have decreased reductive capacity and thus generate a proportionally lower signal. Thus, PrestoBlue™ and alamarBlue® reagents provide a measure of relative cell number that can be used to assess the average proliferation rate of the populations when performed at multiple time points. Because of the live-cell nature of the cell proliferation assays with PrestoBlue™ or alamarBlue® reagents, you can perform downstream functional analyses that are not possible with assays that require cell lysis. Thus, these assays are extremely valuable when using cells available in limited supply, such as primary cells.
The new PrestoBlue™ reagent provides robust data in as little as 10 minutes, whereas most other resazurin-based cell viability reagents on the market require a 1- to 4-hour incubation. PrestoBlue™ reagent is the ideal choice when using cells available in limited supply, such as primary cells, or when assay times are critical. For protocols that require a traditional tetrazolium-based metabolic assay, we offer the Vybrant® MTT Cell Proliferation Assay Kit.
Cell number determination with PrestoBlue™ reagent
Figure 2. Cell number determination with PrestoBlue™ reagent in less time. CHO-K1 cells were plated in cell culture medium in a 384-well plate in quadruplicate starting at 10,000 cells/well with a two-fold serial dilution. Cells were incubated with PrestoBlue™ reagent, alamarBlue® reagent, or CellTiter-Blue® reagent for (left) 10 min or (right) 4 hr. Fluorescence measurements obtained in 10 min with PrestoBlue™ reagent take 4 hr with alamarBlue® and CellTiter-Blue® reagents.
Table 1B. Determining Cell Proliferation based  on Metabolic Activity.
Kit or reagent Ex/Em (Abs) * Stains live cells Stains fixed cells Instrument platform † Assay features Cat. No.
PrestoBlue™ cell viability reagent
•560/590
•(570)
Yes
M
With an incubation step as short as 10 min, PrestoBlue™ reagent saves time compared to other resazurin-based assays such as alamarBlue® reagent.
A13261
A13262
alamarBlue® reagent
•560/590
•(570)
Yes
M
Easily enters into live cells, eliminating the need to lyse or further process cells.
DAL1025
DAL1100
Vybrant® Cell Metabolic Assay Kit
•563/587
•(563)
Yes
M, FC
Provides C12-resazurin, which is better retained than resazurin, yielding brighter signals and lower detection limits.
V23110
Vybrant® MTT Cell Proliferation Assay Kit
(570)
Yes
M
Absorption-based tetrazolium salt assay based on the same reduction principle as the alamarBlue® and PrestoBlue™ assays.
V13154
M6494
* Fluorescence excitation and emission maxima in nm, with absorption maximum in parentheses where applicable. † FC = flow cytometry. FM = fluorescence microscopy. HCS = high-content screening and analysis. M = microplate assay.
Determining cell proliferation based on metabolic activity
Catalog # Name Size Price (USD) Qty
A13261 PrestoBlue™ Cell Viability Reagent, 25 mL Each
346.65
Online Exclusive
357.00
Save 10.35 (3%)
A13262 PrestoBlue™ Cell Viability Reagent, 100 mL Each
708.65
Online Exclusive
732.00
Save 23.35 (3%)
DAL1025 alamarBlue™ Standard Cell Viability Reagent, 25 mL Each
318.65
Online Exclusive
328.00
Save 9.35 (3%)
DAL1100 alamarBlue™ Standard Cell Viability Reagent, 100 mL Each
633.65
Online Exclusive
656.00
Save 22.35 (3%)
V23110 Vybrant™ Cell Metabolic Assay Kit, with C12-resazurin 1 kit
460.00
V13154 CyQUANT™ MTT Cell Viability Assay, 570 nm 1,000 assays
394.00
M6494 MTT (3-(4,5-Dimethylthiazol-2-yl)-2,5-Diphenyltetrazolium Bromide) Each
195.00
Following Cell Division with CellTrace™ Stains
The amine-reactive succinimidyl ester of carboxyfluorescein diacetate (CFSE) is the most widely used probe for generational analysis of cells, including one study which reported the resolution of 8–10 successive generations of lymphocytes [1]. CFSE binds covalently to both intracellular and cell-surface proteins, resulting in more homogeneous cellular labeling and, consequently, better intergenerational resolution than other cell-tracking dyes such as the membrane marker PKH26. When cells divide, CFSE labeling is distributed equally between the daughter cells, which are therefore half as fluorescent as the parents. As a result, each successive generation in a population of proliferating cells is readily detected using a flow cytometer, fluorescence microscope, or fluorescence microplate reader (excitation/emission maxima ~495/525 nm). CFSE is available as a component of the CellTrace™ CFSE Cell Proliferation Kit (Table 1C).
Functionally analogous to CFSE, CellTrace™ Violet stain partitions equally between daughter cells during division, producing successive two-fold reductions in cell-associated fluorescence intensity. When analyzed by flow cytometry, this label partitioning provides a direct indication of cell proliferation (Figure 3). CellTrace™ Violet stain generates blue fluorescence; consequently, it can be used to analyze proliferation of GFP-expressing cells or, in combination with CFSE, to track cells from different origins after mixing.
CellTrace™ Violet stain 
Figure 3. Generational analysis using CellTrace™ Violet stain. Human peripheral blood lymphocytes were harvested and stained with CellTrace™ Violet stain. The violet peaks represent successive generations of cells stimulated with mouse anti–human CD3 antibody and interleukin-2, and grown in culture for 7 days. The peak outlined in black represents cells that were grown in culture for 7 days with no stimulus.
Table 1C. Following Cell Division with Cytoplasmic Tracers.
Kit or reagent Ex/Em (Abs) * Stains live cells Stains fixed cells Instrument platform † Assay features Cat. No.
CFSE (CFDA SE)
495/525
Yes ‡
FC, FM, M
Amine-reactive, cell-permeant fluorescein derivative that provides homogeneous cellular labeling.
C1157
CellTrace™ CFSE Cell Proliferation Kit
492/517
Yes ‡
FC
Validated for generational analysis, with a protocol optimized for flow cytometry.
C34554
CellTrace™ Violet Cell Proliferation Kit
405/455
Yes ‡
FC
Functionally analogous to CFSE. Can be excited with violet diode lasers and multiplexed with green-fluorescent probes.
C34557
* Fluorescence excitation and emission maxima in nm, with absorption maximum in parentheses where applicable. † FC = flow cytometry. FM = fluorescence microscopy. M = microplate assay. ‡ Fixable probe. After fixation, the GFP and RFP fusions of the Premo™ FUCCI Cell Cycle Sensor can be detected with anti-GFP antibodies and anti-RFP antibodies.
Tracking Cell Cycle Distribution with Vybrant® DyeCycle™ Stains
Live-cell studies of DNA content and cell cycle distribution are useful for detecting variations in growth patterns, such as in the study of tumor behavior and suppressor gene mechanisms. In a given population, cells will be distributed among three major phases of the cell cycle: the G0/G1 phase (one set of paired chromosomes per cell), the S phase (DNA synthesis with a variable amount of DNA per cell), and the G2/M phase (two sets of paired chromosomes per cell prior to cell division).
We offer four Vybrant® DyeCycle™ stains for flow cytometric analysis of DNA content in live cells (Table 1). The cell-permeant Vybrant® DyeCycle™ stains are nonfluorescent until bound to double-stranded DNA, and the fluorescence signal generated upon DNA binding is proportional to DNA content. Fluorescence data can then be used to generate a DNA profile histogram that reveals the phases of the cell cycle (Figure 4). For cell cycle analysis in fixed cells, we offer the FxCycle™ Violet and FxCycle™ Far Red stains (Table 1D).
Vybrant® DyeCycle™ Orange stain
Figure 4. Sorting of live-cell populations with Vybrant® DyeCycle™ Orange stain. Cells were sorted based on G0/G1 and G2/M gates using a BD FACSVantage™ flow cytometer (BD Biosciences) with 488 nm excitation and a 585/42 nm bandpass filter.
Table 1D. Determining Cell Cycle Distribution.
Kit or reagent Ex/Em (Abs) * Stains live cells Stains fixed cells Instrument platform † Assay features Cat. No.
Vybrant® DyeCycle™ Violet stain
Vybrant® DyeCycle™ Green stain
Vybrant® DyeCycle™ Orange stain
Vybrant® DyeCycle™ Ruby stain
369/437
506/534
519/563
638/686
Yes
Yes
FC
Cell-permeant DNA-binding dyes for live-cell studies of DNA content and cell cycle distribution. Once bound to dsDNA, these dyes emit a fluorescence signal that is proportional to DNA content. Fluorescence data can be used to generate a DNA profile histogram that reveals the phases of the cell cycle.
V35003
V35004
V35005
V10309
V10273
FxCycle™ Violet stain
FxCycle™ Far Red stain
358/461
640/658
Yes
FC
FxCycle™ Violet (DAPI) preferentially stains dsDNA in fixed cells, whereas FxCycle™ Far Red stain requires addition of RNase A.
F10347
F10348
* Fluorescence excitation and emission maxima in nm, with absorption maximum in parentheses where applicable. † FC = flow cytometry. FM = fluorescence microscopy. HCS = high-content screening and analysis. M = microplate assay.
Determining cell cycle distribution
Detecting Active DNA Synthesis at the Single-Cell Level
Although Vybrant® DyeCycle™ stains can be used to determine cell cycle distributions within a population, the accuracy of this measurement decreases in complex populations, such as those with polyploidy. The only way to accurately detect cells in S phase is to actually measure DNA synthesis. Like traditional [3H] thymidine and bromodeoxyuridine (BrdU) assays, the Click-iT® EdU assay (Table 1E) is based on the incorporation of a nucleoside analog into newly synthesized DNA. In this case, however, the nucleoside analog is EdU (5-ethynyl-2’-deoxyuridine) and it is detected not with radioactivity or an antibody but by a click reaction—a copper-catalyzed covalent reaction between an azide and an alkyne [2]. In the Click-iT® EdU assay, the EdU nucleoside contains the alkyne and the fluorescent detection reagent contains the azide. The Click-iT® EdU signal can be directly visualized using fluorescence microscopy (Figure 5) or flow cytometry and is especially useful for high-content screening (HCS) assays. It has proven effective in diverse species including plants, bacteria, yeast, and a variety of animals.
Proliferating cells in rat ileum 
Figure 5. Proliferating cells in rat ileum. Cell proliferation was detected with the Click-iT® EdU Alexa Fluor® 594 Imaging Kit. Proliferating cells fluoresce red; nuclei are stained with the blue-fluorescent counterstain Hoechst 33342.
Visualizing Cell Cycle Progression with Premo™ FUCCI Cell Cycle Sensor
The Premo™ FUCCI Cell Cycle Sensor allows visualization of cell cycle progression in live cells using traditional fluorescence microscopy or high-content imaging. FUCCI, the fluorescence ubiquitination cell cycle indicator, is a sensor that employs GFP and RFP, each of which is fused to one of two regulators of the cell cycle: geminin and Cdt1 [3]. This cell cycle indicator exhibits a dynamic color change from red to green, marking an individual cell’s progression through the cell cycle (Figure 6). The Premo™ FUCCI Cell Cycle Sensor combines the Cdt1 and geminin FP constructs with the powerful BacMam gene delivery and expression system. Transduction with BacMam reagents is efficient and reproducible in most cell types, without apparent cytopathic effects.
Premo™ FUCCI Cell Cycle Sensor
Figure 6. Imaging cell cycle progression in live cells with Premo™ FUCCI Cell Cycle Sensor. (A) Schematic of cell cycle-dependent expression of the FUCCI fluorescent protein reporters. (B) U2OS cells were transduced with Premo™ FUCCI Cell Cycle Sensor. Images were collected over 15 hr.
Table 1E. Specific Cell Cycle Markers.
Kit or reagent Ex/Em (Abs) * Stains live cells Stains fixed cells Instrument platform † Assay features Cat. No.
Click-iT® EdU Alexa Fluor® 488 Imaging Kit
Click-iT® EdU Alexa Fluor® 555 Imaging Kit
Click-iT® EdU Alexa Fluor® 594 Imaging Kit
Click-iT® EdU Alexa Fluor® 647 Imaging Kit
495/519
555/565
590/615
650/670
Yes **
FM
Following EdU incorporation in cells or whole animals, Click-iT® EdU staining of cells and tissue sections is complete in 60 min, whereas BrdU protocols require harsh denaturation, long blocking steps, and an overnight incubation with anti-BrdU followed by a secondary antibody detection step.
C10337
C10338
C10339
C10340
Click-iT® EdU Alexa Fluor® 488 HCS Assay
Click-iT® EdU Alexa Fluor® 555 HCS Assay
Click-iT® EdU Alexa Fluor® 594 HCS Assay
Click-iT® EdU Alexa Fluor® 647 HCS Assay
495/519
555/565
590/615
650/670
Yes **
HCS
Click-iT® EdU Kits for high-content imaging and analysis are available with Alexa Fluor® 488, Alexa Fluor® 555, Alexa Fluor® 594, and Alexa Fluor® 647 azides for detection of the incorporated EdU.
C10350§
C10352§
C10354§
C10356§
Click-iT® EdU Pacific Blue™ FC Assay Kit
Click-iT® EdU Alexa Fluor® 488 FC Assay Kit
Click-iT® EdU Alexa Fluor® 647 FC Assay Kit
410/455
495/515
650/670
Yes **
FC
Click-iT® EdU Kits for flow cytometry analysis are available with Pacific Blue™, Alexa Fluor® 488, and Alexa Fluor® 647 azides for detection of the incorporated EdU.
C10418§
C10425§
C10424§
Click-iT® EdU Microplate Assay Kit
568/585
Yes **
M
Detect incorporated EdU with Oregon Green® azide, followed by HRP anti–Oregon Green® and Amplex® UltraRed substrate.
C10214
Premo™ FUCCI Cell Cycle Sensor
485/520
555/584
Yes ‡
FC, FM, HCS
As a cell progresses through the cell cycle, fluorescence of the Premo™ FUCCI Sensor changes from red to green.
P36232
HCS Mitotic Index Kit
495/519
Yes
HCS, FM
Anti–phosphohistone H3 serves as a sensitive index of mitosis and is detected with Alexa Fluor® 488 secondary antibody.
H10293
* Fluorescence excitation and emission maxima in nm, with absorption maximum in parentheses where applicable. † FC = flow cytometry. FM = fluorescence microscopy. HCS = high-content screening and analysis. M = microplate assay. ‡ Fixable probe. After fixation, the GFP and RFP fusions of the Premo™ FUCCI Cell Cycle Sensor can be detected with anti-GFP antibodies and anti-RFP antibodies. § The Click-iT® EdU HCS Assay Kits are available in 2-plate (Cat. No. shown) and 10-plate sizes. The Click-iT® EdU Flow Cytometry Assay Kits are available in a 50-assay (Cat. No. shown) and a 100-assay size. ** In the Click-iT® EdU assay, EdU is incorporated in live cells, but then detected following cell fixation and permeabilization.
Ordering Information for All Products
Catalog # Name Size Price (USD) Qty
C7026 CyQUANT™ Cell Proliferation Assay, for cells in culture Each
469.65
Online Exclusive
486.00
Save 16.35 (3%)
C35006 CyQUANT™ NF Cell Proliferation Assay, 1,000 assays Each
529.65
Online Exclusive
546.00
Save 16.35 (3%)
C35011 CyQUANT™ Direct Cell Proliferation Assay, 10 microplates kit Each
546.65
Online Exclusive
568.00
Save 21.35 (4%)
C35012 CyQUANT™ Direct Cell Proliferation Assay, 100 microplates kit Each
3,630.65
Online Exclusive
3,790.00
Save 159.35 (4%)
C35007 CyQUANT™ NF Cell Proliferation Assay, 200 assays kit Each
205.65
Online Exclusive
212.00
Save 6.35 (3%)
A13261 PrestoBlue™ Cell Viability Reagent, 25 mL Each
346.65
Online Exclusive
357.00
Save 10.35 (3%)
A13262 PrestoBlue™ Cell Viability Reagent, 100 mL Each
708.65
Online Exclusive
732.00
Save 23.35 (3%)
DAL1025 alamarBlue™ Standard Cell Viability Reagent, 25 mL Each
318.65
Online Exclusive
328.00
Save 9.35 (3%)
DAL1100 alamarBlue™ Standard Cell Viability Reagent, 100 mL Each
633.65
Online Exclusive
656.00
Save 22.35 (3%)
V23110 Vybrant™ Cell Metabolic Assay Kit, with C12-resazurin 1 kit
460.00
V13154 CyQUANT™ MTT Cell Viability Assay, 570 nm 1,000 assays
394.00
M6494 MTT (3-(4,5-Dimethylthiazol-2-yl)-2,5-Diphenyltetrazolium Bromide) Each
195.00
C1157 5(6)-CFDA, SE; CFSE (5-(and-6)-Carboxyfluorescein Diacetate, Succinimidyl Ester), mixed isomers Each
369.65
Online Exclusive
382.00
Save 12.35 (3%)
C34554 CellTrace™ CFSE Cell Proliferation Kit, for flow cytometry, 180 reactions kit Each
272.00
C34557 CellTrace™ Violet Cell Proliferation Kit, for flow cytometry, 180 assays kit Each
550.00
V35003 Vybrant™ DyeCycle™ Violet Stain 200 µL
622.00
V35004 Vybrant™ DyeCycle™ Green Stain 400 µL
622.00
V35005 Vybrant™ DyeCycle™ Orange Stain 400 µL
614.00
V10309 Vybrant™ DyeCycle™ Ruby Stain, 100 reactions 100 assays
389.00
V10273 Vybrant™ DyeCycle™ Ruby Stain, 400 reactions 400 assays
640.00
F10347 FxCycle™ Violet Stain Each
518.00
F10348 FxCycle™ Far Red Stain Each
514.00
C10337 Click-iT™ EdU Cell Proliferation Kit for Imaging, Alexa Fluor™ 488 dye Each
892.00
C10338 Click-iT™ EdU Cell Proliferation Kit for Imaging, Alexa Fluor™ 555 dye Each
900.00
C10339 Click-iT™ EdU Cell Proliferation Kit for Imaging, Alexa Fluor™ 594 dye Each
862.65
Online Exclusive
890.00
Save 27.35 (3%)
C10340 Click-iT™ EdU Cell Proliferation Kit for Imaging, Alexa Fluor™ 647 dye Each
910.00
C10350 Click-iT™ EdU Alexa Fluor™ 488 HCS Assay, 2 x 96 well plates Each
789.65
Online Exclusive
874.00
Save 84.35 (10%)
C10352 Click-iT™ EdU Alexa Fluor™ 555 HCS Assay, 2 x 96 well plates Each
842.00
C10354 Click-iT™ EdU Alexa Fluor™ 594 HCS Assay, 2 x 96 well plates Each
838.00
C10356 Click-iT™ EdU Alexa Fluor™ 647 HCS Assay, 2 x 96 well plates Each
832.65
Online Exclusive
858.00
Save 25.35 (3%)
C10418 Click-iT™ EdU Pacific Blue™ Flow Cytometry Assay Kit Each
764.00
C10425 Click-iT™ EdU Alexa Fluor™ 488 Flow Cytometry Assay Kit, 50 assays Each
764.00
C10424 Click-iT™ EdU Alexa Fluor™ 647 Flow Cytometry Assay Kit, 50 assays Each
764.00
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.