Gibco fetal bovine serum products
Through our unwavering commitment to quality, we continue to provide scientists with the consistent reliability, service, value, and innovation that have made Gibco products a global market leader for over 50 years.
Gibco fetal bovine serum products
Choose the right sera for your specific cell culture needs—from basic research to specialty assays. Whether you need sera with the least viral risk, the lowest endotoxin levels, or sera qualified for specialty applications and assays, Gibco products offer superior value.
Value FBS
Sera for standard research applications
  • Up to 50 quality tests including 9CFR virus testing, endotoxin, performance
  • Triple 0.1 micron filtration*
Premium FBS
Sera with the least risk of BSE and lower viral risk
  • Meets USP/EP guidelines
  • Up to 90 quality tests including EMA virus testing; USP/EP mycoplasma, endotoxin, performance; biochemical/hormonal profiling; Oritain fingerprinting
  • Triple 0.1 micron filtration
Specialty FBS
Sera qualified for specialty research
  • Specific assays, including stem cell research, immunoassays, antibodies, and others
*Brazilian FBS manufactured in Brazil is double 0.1 micron filtered, not triple (Cat. Nos. 12657011 and 12657029).
Gibco FBS cross-reference tool
The Gibco FBS cross reference tool can be used to find a direct replacement for other serum suppliers.
Enter supplier's product number (SKU) or supplier name:
If your product number does not appear, please email us so we can include it in our tool, click here
BrandSizeSupplier SKUSupplier ProductGibco SKUGibco Product
HyClone50 mLSH30071.01Characterized Fetal Bovine Serum, US Origin, 50 mLA3160501Gibco Fetal Bovine Serum, qualified, US origin, 50 mL (Value Plus FBS)
HyClone50 mLSH30071.01HICharacterized Fetal Bovine Serum, Heat inactivated, US Origin, 50 mLA3840101Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 50 mL (Value Plus FBS)
HyClone100 mLSH30071.02Characterized Fetal Bovine Serum, US Origin, 100 mL26140087Gibco Fetal Bovine Serum, qualified, US origin, 100 mL (Value Plus FBS)
HyClone100 mLSH30071.02HICharacterized Fetal Bovine Serum, Heat inactivated, US Origin, 100 mL16140063Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 100 mL (Value Plus FBS)
HyClone500 mLSH30071.03Characterized Fetal Bovine Serum, US Origin, 500 mL26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
HyClone500 mLSH30071.03HICharacterized Fetal Bovine Serum, Heat inactivated, US Origin, 500 mL16140071Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 500 mL (Value Plus FBS)
HyClone100 mLSH30084.02Characterized Fetal Bovine Serum, AUS Origin, 100 mL10099133Gibco Fetal Bovine Serum, qualified, Australia origin, 100 mL (Premium FBS)
HyClone500 mLSH30084.03Characterized Fetal Bovine Serum, AUS Origin, 500 mL10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
HyClone500 mLSH30084.03HICharacterized Fetal Bovine Serum, Heat inactivated AUS, Origin, 500 mL10100147Gibco Fetal Bovine Serum, qualified, heat inactivated Australia origin, 500 mL (Premium FBS)
HyClone500 mLSH30084.03IRCharacterized Fetal Bovine Serum, Gamma irradiated, AUS, Origin, 500 mL10101145Gibco Fetal Bovine Serum, qualified, gamma irradiated Australia origin, 500 mL (Premium FBS)
HyClone1000 mLSH30084.04Characterized Fetal Bovine Serum, AUS Origin, 1000 mL10099158Gibco Fetal Bovine Serum, qualified, Australia origin, 1000 mL (Premium FBS)
HyClone1000 mLSH30084.04HICharacterized Fetal Bovine Serum, Heat inactivated AUS, Origin, 1000 mL10100154Gibco Fetal Bovine Serum, qualified, heat inactivated Australia origin, 1000 mL (Premium FBS)
HyClone100 mLSH30406.01Characterized Fetal Bovine Serum, NZ Origin, 100 mL10091130Gibco Fetal Bovine Serum, qualified, New Zealand origin, 100 mL (Premium FBS)
HyClone500 mLSH30406.02Characterized Fetal Bovine Serum, NZ Origin, 500 mL10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
HyClone500 mLSH30406.02IRCharacterized Fetal Bovine Serum, Gamma Irradiated, NZ origin, 500 mL10094142Gibco Fetal Bovine Serum, gamma irradiated, qualified, New Zealand, 500 mL (Premium FBS)
HyClone500 mLSH30406.05Characterized Fetal Bovine Serum, NZ Origin, 500 mL10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
HyClone500 mLSH40496.03HICharacterized Fetal Bovine Serum, Canadian Origin, 500 mL12483020Gibco Fetal Bovine Serum, qualified, Canada origin, 500 mL (Value FBS)
HyClone50 mLSH30068.01Charcoal/Dextran Treated FBS, US Origin, 50 mLA3382101Gibco Fetal Bovine Serum, charcoal stripped, USDA-approved regions, 50 mL (Specialty FBS)
HyClone100 mLSH30068.02Charcoal/Dextran Treated Fetal Bovine Serum, US Origin, 100 mLA3382101Gibco Fetal Bovine Serum, charcoal stripped, USDA-approved regions, 50 mL (Specialty FBS)
HyClone500 mLSH30068.03Charcoal/Dextran Treated Fetal Bovine Serum, US Origin, 500 mL12676029Gibco Fetal Bovine Serum, charcoal stripped, USDA-approved regions, 500 mL (Specialty FBS)
HyClone50 mLSH30070.01Defined Fetal Bovine Serum, US Origin, 50 mLA3160401Gibco Fetal Bovine Serum, certified, US origin, 50 mL (Premium FBS)
HyClone50 mLSH30070.01HIDefined Fetal Bovine Serum, US Origin, Heat inactivated, 50 mL (Premium FBS)A3840001Gibco Fetal Bovine Serum, certified, heat inactivated, US origin, 50 mL (Premium FBS)
HyClone100 mLSH30070.02Defined Fetal Bovine Serum, US Origin, 100 mL16000036Gibco Fetal Bovine Serum, certified, US origin, 100 mL (Premium FBS)
HyClone100 mLSH30070.02HIDefined Fetal Bovine Serum, US Origin, Heat inactivated, 100 mL10082139Gibco Fetal Bovine Serum, certified, heat inactivated, US origin, 100 mL (Premium FBS)
HyClone500 mLSH30070.03 Defined Fetal Bovine Serum, US Origin, 500 mL16000044Gibco Fetal Bovine Serum, certified, US origin, 500 mL (Premium FBS)
HyClone500 mLSH30070.03HIDefined Fetal Bovine Serum, US Origin, Heat inactivated, 500 mL10082147Gibco Fetal Bovine Serum, certified, heat inactivated, US origin, 500 mL (Premium FBS)
HyClone100 mLSH30088.02Standard Fetal Bovine Serum, US Origin, 100 mL10437010Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 100 mL (Value FBS)
HyClone500 mLSH30088.03Standard Fetal Bovine Serum, US Origin, 500 mL10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
HyClone100 mLSH30088.02HIStandard Fetal Bovine Serum, US Origin, Heat inactivated 100 mL10437018Gibco Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions, 100 mL (Value FBS)
HyClone500 mLSH30088.03HIStandard Fetal Bovine Serum, US Origin, Heat inactivated 500 mL10438026Gibco Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions, 500 mL (Value FBS)
HyClone500 mLSH30070.03MHuman Mesenchymal Stem Cell Screened Fetal Bovine Serum, US Origin, 500 mL12662029Gibco Fetal Bovine Serum, mesenchymal stem cell-qualified, USDA-approved regions, 500 mL (Specialty FBS)
HyClone50 mLSV30160.01Research Grade Fetal Bovine Serum, South American Origin, 50 mLA3160801Gibco Fetal Bovine Serum, qualified, Brazil origin, 50 mL (Value FBS)
HyClone100 mLSV30160.02Research Grade Fetal Bovine Serum, South American Origin, 100 mL10270098Gibco Fetal Bovine Serum, qualified, Brazil origin, 100 mL (Value FBS)
HyClone100 mLSV30160.02HIResearch Grade Fetal Bovine Serum, South American Origin, Heat inactivated, 100 mL10500056Gibco Fetal Bovine Serum, qualified, heat inactivated, Brazil origin, 100 mL (Value FBS)
HyClone500 mLSV30160.03Research Grade Fetal Bovine Serum, South American Origin, 500 mL10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
HyClone500 mLSV30180.03HyClone Fetal Bovine Serum, South American Origin10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
HyClone500 mLSV30160.03HIResearch Grade Fetal Bovine Serum, South American Origin, Heat inactivated, 500 mL10500064Gibco Fetal Bovine Serum, qualified, heat inactivated, Brazil origin, 500 mL (Value FBS)
HyClone50 mLSH30079.01Dialyzed Fetal Bovine Serum, US Origin, 50 mLA3382001Gibco Fetal Bovine Serum, dialyzed, US origin, 50 mL (Specialty FBS)
HyClone100 mLSH30079.02Dialyzed Fetal Bovine Serum, US Origin, 100 mLA3382001Gibco Fetal Bovine Serum, dialyzed, US origin, 50 mL (Specialty FBS)
HyClone500 mLSH30079.03Dialyzed Fetal Bovine Serum, US Origin, 500 mL26400044Gibco Fetal Bovine Serum, dialyzed, US origin, 500 mL (Specialty FBS)
HyClone100 mLSH30070.02EES Screened Fetal Bovine Serum, US Origin, 100 mL16141061Gibco Fetal Bovine Serum, embryonic stem-cell, qualified US origin, 100 mL (Specialty FBS)
HyClone500 mLSH30070.03EES Screened Fetal Bovine Serum, US Origin, 500 mL16141079Gibco Fetal Bovine Serum, embryonic stem-cell, qualified, US origin, 500 mL (Specialty FBS)
HyClone500 mLSH30898.03Fetal Bovine Serum, Super Low IgG, US origin, 500 mL16250078Gibco Fetal Bovine Serum, ultra-low IgG, US origin, 500 mL (Specialty FBS)
HyClone100 mLSH30898.02Fetal Bovine Serum, Super Low IgG, US origin, 100 mLA3381901Gibco Fetal Bovine Serum, ultra-low IgG, US origin, 50 mL (Specialty FBS)
HyClone500 mLSH30910.03Fetal Bovine Serum, USDA Tested, 500 mL10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
Sigma/SAFC50 mLF2442-50MLFetal Bovine, USA origin, 50 mLA3160401Gibco Fetal Bovine Serum, certified, US origin, 50 mL (Premium FBS)
Sigma/SAFC100 mLF2442-100MLFetal Bovine, USA origin, 100 mL16000036Gibco Fetal Bovine Serum, certified, US origin, 100 mL (Premium FBS)
Sigma/SAFC500 mLF2442-500MLFetal Bovine, USA origin, 500 mL16000044Gibco Fetal Bovine Serum, certified, US origin, 500 mL (Premium FBS)
Sigma/SAFC1LF2442-1LFetal Bovine, USA origin, 1L16000069Gibco Fetal Bovine Serum, certified, US origin, 500 mL (Premium FBS)
Sigma/SAFC100 mLF4135-100MLFetal Bovine, USA origin, Heat inactivated, 100ML16140063Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 100 mL (Value Plus FBS)
Sigma/SAFC500 mLF4135-500MLFetal Bovine, USA origin, Heat inactivated, 500ML16140071Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 500 mL (Value Plus FBS)
Sigma/SAFC1LF4135-1LFetal Bovine, USA origin, Heat inactivated, 1L16140085Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 1000 mL (Value Plus FBS)
Sigma/SAFC100 mL12303C-100MLFetal Bovine, USDA approved countries, 100ML10437010Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 100 mL (Value FBS)
Sigma/SAFC500 mL12303C-500MLFetal Bovine, USDA approved countries, 500ML10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
Sigma/SAFC100 mLF1051-100MLFetal Bovine,Canada origin, 100ML12483012Gibco Fetal Bovine Serum, qualified, Canada origin, 100 mL (Value FBS)
Sigma/SAFC500 mLF1051-500MLFetal Bovine, Canada origin, 500ML12483020Gibco Fetal Bovine Serum, qualified, Canada origin, 500 mL (Value FBS)
Sigma/SAFC50 mLF0926-50MLFetal Bovine Serum, USDA approved, 50 mLA3160601Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 50 mL (Value FBS)
Sigma/SAFC100 mLF0926-100MLFetal Bovine Serum, USDA approved, 100 mL10437010Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 100 mL (Value FBS)
Sigma/SAFC500 mLF0926-500MLFetal Bovine Serum, USDA approved, 500 mL10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
Sigma/SAFC50 mL12306C-50MLFetal Bovine, USDA approved countries, Heat inactivated, 50MLA3840201Gibco Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions, 50 mL (Value FBS)
Sigma/SAFC100 mL12306C-100MLFetal Bovine, USDA approved countries, Heat inactivated, 100ML10437018Gibco Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions, 100 mL (Value FBS)
Sigma/SAFC500 mL12306C-500MLFetal Bovine, USDA approved countries, Heat inactivated, 500ML10438026Gibco Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions, 500 mL (Value FBS)
Sigma/SAFC100 mLF9423-100MLFetal Bovine Serum, Australia origin, 100 mL10099133Gibco Fetal Bovine Serum, qualified, Australia origin, 100 mL (Premium FBS)
Sigma/SAFC500 mLF3297-500MLFetal Bovine Serum, Australia origin, y-irradiated, 500 mL10101145Gibco Fetal Bovine Serum, qualified, gamma irradiated Australia origin, 500 mL (Premium FBS)
Sigma/SAFC100 mL12003C-100MLFetal Bovine Serum, Australia origin, 100 mL10099133Gibco Fetal Bovine Serum, qualified, Australia origin, 100 mL (Premium FBS)
Sigma/SAFC500 mLF9423-500MLFetal Bovine Serum, Australia origin, 500 mL10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
Sigma/SAFC500 mL12003C-500MLFetal Bovine Serum, Australia origin, 500 mL10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
Sigma/SAFC1000 mLF9423-1000MLFetal Bovine Serum, Australia origin, 1000 mL10099158Gibco Fetal Bovine Serum, qualified, Australia origin, 1000 mL (Premium FBS)
Sigma/SAFC100 mL12103C-100MLFetal Bovine Serum, USA origin, 100 mL26140087Gibco Fetal Bovine Serum, qualified, US origin, 100 mL (Value Plus FBS)
Sigma/SAFC500 mL12103C-500MLFetal Bovine Serum, USA origin, 500 mL26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
Sigma/SAFC50 mLF8067-50MLFetal Bovine Serum, New Zealand origin, 50 mLA3160901Gibco Fetal Bovine Serum, qualified, New Zealand origin, 50 mL (Premium FBS)
Sigma/SAFC100 mLF8067-100MLFetal Bovine Serum, New Zealand origin, 100 mL10091130Gibco Fetal Bovine Serum, qualified, New Zealand origin, 100 mL (Premium FBS)
Sigma/SAFC100 mL12203C-100MLFetal Bovine Serum, New Zealand origin, 100 mL10091130Gibco Fetal Bovine Serum, qualified, New Zealand origin, 100 mL (Premium FBS)
Sigma/SAFC500 mLF8067-500MLFetal Bovine Serum, New Zealand origin, 500 mL10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
Sigma/SAFC500 mL12207C-500MLFetal Bovine Serum, New Zealand origin, 500 mL10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
Sigma/SAFC500 mL12203C-500MLFetal Bovine Serum, New Zealand origin, 500 mL10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
Sigma/SAFC1000 mL12203C-1000MLFetal Bovine Serum, New Zealand origin, 1000 mL10091155Gibco Fetal Bovine Serum, qualified, New Zealand origin, 1000 mL (Premium FBS)
Sigma/SAFC1001 mL12207C-1000MLFetal Bovine Serum, New Zealand origin, 1000 mL10091155Gibco Fetal Bovine Serum, qualified, New Zealand origin, 1000 mL
Sigma/SAFC100 mLF8317-100MLFetal Bovine Serum, New Zealand origin, Heat inactivated, 100 mL10093136Gibco Fetal Bovine Serum, qualified, heat inactivated, New Zealand origin, 100 mL (Premium FBS)
Sigma/SAFC500 mLF8317-500MLFetal Bovine Serum, New Zealand origin, Heat inactivated, 500 mL10093177Gibco Fetal Bovine Serum, qualified, heat inactivated, New Zealand origin, 500 mL (Premium FBS)
Sigma/SAFC100 mLF8192-100MLFetal Bovine Serum, Australia origin, Heat inactivated, 100 mL10100139Gibco Fetal Bovine Serum, qualified, heat inactivated Australia origin, 100 mL (Premium FBS)
Sigma/SAFC500 mLF8192-500MLFetal Bovine Serum, Australia origin, Heat inactivated, 500 mL10100147Gibco Fetal Bovine Serum, qualified, heat inactivated Australia origin, 500 mL (Premium FBS)
Sigma/SAFC500 mL12006C-500mLFetal Bovine Serum, Australia origin, Heat inactivated, 500 mL10100147Gibco Fetal Bovine Serum, qualified, heat inactivated Australia origin, 500 mL (Premium FBS)
Sigma/SAFC100 mLF0392-100MLFetal Bovine Serum, USA origin, Dialyzed, 100 mLA3382001Gibco Fetal Bovine Serum, dialyzed, 100 mL (Specialty FBS)
Sigma/SAFC100 mL12105C-100MLFetal Bovine Serum, USA origin, Dialyzed, 100 mLA3382001Gibco Fetal Bovine Serum, dialyzed, 100 mL (Specialty FBS)
Sigma/SAFC500 mLF0392-500MLFetal Bovine Serum, USA origin, Dialyzed, 500 mL26400044Gibco Fetal Bovine Serum, dialyzed, 500 mL (Specialty FBS)
Sigma/SAFC100 mLF6765-100MLFetal Bovine Serum, USA origin, Charcoal Stripped, 100 mLA3382101Gibco Fetal Bovine Serum, charcoal stripped, USDA-approved regions, 50 mL (Specialty FBS)
Sigma/SAFC500 mLF6765-500MLFetal Bovine Serum, USA origin, Charcoal Stripped, 500 mL12676029Gibco Fetal Bovine Serum, charcoal stripped, USDA-approved regions, 500 mL (Specialty FBS)
Sigma/SAFC100 mLF6178-100MLFetal Bovine Serum, USA origin, 100 ml26140087Gibco Fetal Bovine Serum, qualified, US origin, 100 mL (Value Plus FBS)
Sigma/SAFC500 mLF6178-500MLFetal Bovine Serum, USA origin, 500 ml26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
Sigma/SAFC100 mL12106C-100MLFetal Bovine Serum, USA origin, Heat inactivated, 100 mL16140063Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 100 mL (Value Plus FBS)
Sigma/SAFC500 mL12106C-500MLFetal Bovine Serum, USA origin, Heat inactivated, 500 mL16140071Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 500 mL (Value Plus FBS)
Sigma/SAFC100 mLES-009-CFetal Bovine Serum, US Origin, EmbryoMac ES Cell Qualified FBS, 100 mL16141061Gibco Fetal Bovine Serum, embryonic stem cell qualified, US origin, 100 mL (Specialty FBS)
Sigma/SAFC500 mLES-009-BFetal Bovine Serum, US Origin, EmbryoMac ES Cell Qualified FBS, 500 mL16141079Gibco Fetal Bovine Serum, embryonic stem cell qualified, US origin, 500 mL (Specialty FBS)
Sigma/SAFC500 mLTMS-013-BFetal Bovine Serum, US Origin, 500 mL26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
Sigma/SAFC500 mLF7524-500MLFetal Bovine Serum, South American origin, 500 mL10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
Sigma/SAFC100 mLF7524-100MLFetal Bovine Serum, South American origin, 100 mL10270098Gibco Fetal Bovine Serum, qualified, Brazil origin, 100 mL (Value FBS)
Sigma/SAFC50 mLF0804-50MLFetal Bovine Serum, South American origin, 50 mLA3160801Gibco Fetal Bovine Serum, qualified, Brazil origin, 50 mL (Value FBS)
Sigma/SAFC500 mL12306C-500MLFetal Bovine Serum, Heat inactivated, South American origin, 500 mL10500064Gibco Fetal Bovine Serum, qualified, heat inactivated, Brazil origin, 500 mL (Value FBS)
Sigma/SAFC500 mLF0679-500MLFetal Bovine Serum, Heat inactivated, y-irradiated, South American origin, 500 mL10499044Gibco Fetal Bovine Serum, qualified, gamma irradiated, Brazil origin, 500 mL (Value FBS)
Gemini500 mL100-106Benchmark Fetal Bovine Serum10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
Gemini500 mL100-108Dialyzed FBS26400044Gibco Fetal Bovine Serum, dialyzed, US origin, 500 mL (Specialty FBS)
Gemini500 mL100-119Charcoal Dextran Stripped FBS12676029Gibco Fetal Bovine Serum, charcoal stripped, USDA-approved regions, 500 mL (Specialty FBS)
Gemini500 mL100-120AbSolute Low IgG FBS16250078Gibco Fetal Bovine Serum, ultra-low IgG, US origin, 500 mL (Specialty FBS)
Gemini500 mL100-125Stasis Stem Cell Qualified FBS, USDA10439024Gibco Fetal Bovine Serum, embryonic stem cell qualified, USDA approved regions, 500 mL (Specialty FBS)
Gemini500 mL100-500GemCell United States Origin FBS16000044Gibco Fetal Bovine Serum, certified, US origin, 500 mL (Premium FBS)
Gemini500 mL100-525Stasis Stem Cell Qualified FBS, US16141079Gibco Fetal Bovine Serum, embryonic stem-cell, qualified, US origin, 500 mL (Specialty FBS)
Gemini500 mL100-700No-Worries NZ origin FBS10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
Gemini500 mL900-108Foundation FBS (Grade IV)10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
Atlanta Biologicals500 mLS10250Embryonic Stem Cell Qualified FBS10439024Gibco Fetal Bovine Serum, embryonic stem cell qualified, USDA approved regions, 500 mL (Specialty FBS)
Atlanta Biologicals500 mLS11050Advantage FBS, United States or USDA10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
Atlanta Biologicals500 mLS11050HAdvantage Heat Inactivated FBS, United States or USDA10438026Gibco Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions, 500 mL (Value FBS)
Atlanta Biologicals500 mLS11550Premium Select FBS, United States or USDA16000044Gibco Fetal Bovine Serum, certified, US origin, 500 mL (Premium FBS)
Atlanta Biologicals500 mLS11550HPremium Select FBS, Heat Inactivated FBS, United States or USDA10082147Gibco Fetal Bovine Serum, certified, heat inactivated, US origin, 500 mL (Premium FBS)
Atlanta Biologicals500 mLS11610Charcoal/Dextran Treated FBS 12676029Gibco Fetal Bovine Serum, charcoal stripped, USDA-approved regions, 500 mL (Specialty FBS)
Atlanta Biologicals500 mLS12450Optima FBS, US26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
Atlanta Biologicals500 mLS12450HOptima Heat Inactivated FBS, US16140071Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 500 mL (Value Plus FBS)
Atlanta Biologicals500 mLS12650FBS, Dialyzed 10 kD26400044Gibco Fetal Bovine Serum, dialyzed, US origin, 500 mL (Specialty FBS)
VWR/Seradigm500 mL1600-500FBS Ultimate Grade16000044Gibco Fetal Bovine Serum, certified, US origin, 500 mL (Premium FBS)
VWR/Seradigm500 mL1600-500HFBS Ultimate Grade, Heat Inactivated10082147Gibco Fetal Bovine Serum, certified, heat inactivated, US origin, 500 mL (Premium FBS)
VWR/Seradigm500 mL1500-500FBS Premium Grade26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
VWR/Seradigm500 mL1500-500HFBS Premium Grade, Heat Inactivated16140071Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 500 mL (Value Plus FBS)
VWR/Seradigm500 mL1300-500FBS, USDA Approved Origin10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
Corning500 mL35-010-CVFetal Bovine Serum, Regular , USDA approved source, 500 mL10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
Corning500 mL35-015-CVFetal Bovine Serum, Premium, US sourced, 500 mL26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
Corning500 mL35-016-CVFetal Bovine Serum, Premium, Heat Inactivated, 500 mL16140071Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 500 mL (Value Plus FBS)
Corning500 mL35-076-CVFetal Bovine Serum, Australia Source, 500 mL10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
Corning500 mL35-079-CVFetal Bovine Serum, South American Source, EU Compliant, 500 mL10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
Corning500 mL35-073-CVFetal Bovine Serum, Premium (Low IgG), 500 mL16250078Gibco Fetal Bovine Serum, ultra-low IgG, US origin, 500 mL (Specialty FBS)
Corning500 mL35-081--CVFetal Bovine Serum, New Zealand Source, 500 mL10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
PAN-Biotech500 mLP40-36500FBS Good, New Zealand origin, 500 mL10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
PAN-Biotech100 mLP40-36100FBS Good, New Zealand origin, 100 mL10091130Gibco Fetal Bovine Serum, qualified, New Zealand origin, 100 mL (Premium FBS)
PAN-Biotech500 mLP30-1302FBS, Australia, 500 mL10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
PAN-Biotech100 mLP30-1301FBS, Australia, 100 mL10099133Gibco Fetal Bovine Serum, qualified, Australia origin, 100 mL (Premium FBS)
PAN-Biotech500 mLP40-39500FBS Good, Australia origin, 500 mL10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
PAN-Biotech100 mLP40-39100FBS Good, Australia origin, 100 mL10099133Gibco Fetal Bovine Serum, qualified, Australia origin, 100 mL (Premium FBS)
PAN-Biotech500 mLP30-3306FBS Standard, South American, 500 mL10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
PAN-Biotech500 mLP30-1402Fetal bovine serum, US origin, 500 mL26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
PAN-Biotech500 mlP30-2802Fetal bovine serum, Ultra low IgG, 500 mL16250078Gibco Fetal Bovine Serum, ultra-low IgG, US origin, 500 mL (Specialty FBS)
PAN-Biotech100 mLP30-3305FBS Standard, South American, 100 mL10270098Gibco Fetal Bovine Serum, qualified, Brazil origin, 100 mL (Value FBS)
PAN-Biotech100 mLP30-1401Fetal bovine serum, US origin, 100 mL26140087Gibco Fetal Bovine Serum, qualified, US origin, 100 mL (Value Plus FBS)
PAN-Biotech100 mLP30-2801Fetal bovine serum, Ultra low IgG, 100 mLA3381901Gibco Fetal Bovine Serum, ultra-low IgG, US origin, 50 mL (Specialty FBS)
Cellmax500 mLSA102.02Australian premium fetal bovine serum, 500 mL10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
Cellmax100 mLSA102.01Australian premium fetal bovine serum, 100 mL10099133Gibco Fetal Bovine Serum, qualified, Australia origin, 100 mL (Premium FBS)
Bovogen 500 mLSFBS-NZFBS-New Zealand Bovine Material10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
Bovogen 500 mLSFBS-AUFBS-Australia origin10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
Bovogen 500 mLSFBS-IgGFBS-Ultra Low IgG16250078Gibco Fetal Bovine Serum, ultra-low IgG, US origin, 500 mL (Specialty FBS)
Bovogen 500 mLSFBS-NZFBS-New Zeland origin10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
EXCEL500 mLFND500Fetal Bovine Serum (Defined), Australian origin10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
EXCEL100 mLFND100Fetal Bovine Serum (Defined), Australian origin10099133Gibco Fetal Bovine Serum, qualified, Australia origin, 100 mL (Premium FBS)
Life Science Production500 mLS-001A-BREU Grade FBS Origin: Brazil 10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
Life Science Production500 mLS-001A-HI-BREU Grade FBS Origin: Brazil, Heat inactivated10500064Gibco Fetal Bovine Serum, qualified, heat inactivated, Brazil origin, 500 mL
Life Science Production500 mLS-001-USStandard FBS, US origin16000044Gibco Fetal Bovine Serum, certified, US origin, 500 mL (Premium FBS)
Life Science Production500 mLS-001-USDAStandard FBS, USDA26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
Life Science Production500 mLS-001-AUAustralian FBS10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
Life Science Production100 mLS-001B-BREU Grade FBS, Brazilian Origin10270098Gibco Fetal Bovine Serum, qualified, Brazil origin, 100 mL (Value FBS)
Life Science Production100 mLS-001B-HI-BREU Grade FBS, Heat inactivated, Brazilian Origin10500056Gibco Fetal Bovine Serum, qualified, heat inactivated, Brazil origin, 100 mL (Value FBS)
BioSera500 mLFB-1001/500Fetal Bovine Serum (Sth American)-500ml10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
BioSera500 mLFB-1370/500Fetal Bovine Serum (Australian origin, USDA approved)-500ml10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
BioSera100 mLFB-1001/100Fetal Bovine Serum (Sth American)-100ml10270098Gibco Fetal Bovine Serum, qualified, Brazil origin, 100 mL (Value FBS)
BioSera100 mLFB-1350Fetal Bovine Serum (USA)26140087Gibco Fetal Bovine Serum, qualified, US origin, 100 mL (Value Plus FBS)
BioSera100 mLFB-1370/100Fetal Bovine Serum (Australian origin, USDA approved)-500ml10099133Gibco Fetal Bovine Serum, qualified, Australia origin, 100 mL (Premium FBS)
BioWest500 mLS1810-500Fetal Bovine Serum (FBS) South America, 500 mL10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
BioWest500 mLS1520-500Fetal Bovine Serum (FBS) USA, 500 mL26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
BioWest100 mLS1810-100Fetal Bovine Serum (FBS) South America, 100 mL10270098Gibco Fetal Bovine Serum, qualified, Brazil origin, 100 mL (Value FBS)
BioWest100 mLS1520-100Fetal Bovine Serum (FBS) USA, 100 mL26140087Gibco Fetal Bovine Serum, qualified, US origin, 100 mL (Value Plus FBS)
Labtech500 mLFCS-SA/500Fetal Bovine Serum, South American origin 500 mL10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
Clontech500 mL631101FBS US-sourced, 500 mL26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
Clontech100 mL631105FBS US-sourced, 100 mL26140087Gibco Fetal Bovine Serum, qualified, US origin, 100 mL (Value Plus FBS)
Biological Industries500 mL04-007-1AFetal Bovine Serum, South American10270106Gibco Fetal Bovine Serum, qualified, Brazil origin, 500 mL (Value FBS)
Biological Industries500 mL04-001-1AFetal Bovine Serum, US origin26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
NQBB International Biological500 mLA6806-11Fetal Bovine Serum, Australia origin10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
JR Scientific500 mL490016-872Fetal Bovine Serum, USDA approved10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
JR Scientific50 mL490016-868Fetal Bovine Serum, USDA approvedA3160601Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 50 mL (Value FBS)
Atlas Biologicals50 mLF-0050-DFetal Bovine Serum-USDA Approved (Grade) SourcingA3160601Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 50 mL (Value FBS)
Atlas Biologicals500 mLF-0500-DFetal Bovine Serum-USDA Approved (Grade) Sourcing10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
Atlas Biologicals50 mLF-0050-AFetal Bovine Serum-US originA3160501Gibco Fetal Bovine Serum, qualified, US origin, 50 mL (Value Plus FBS)
Atlas Biologicals500 mLF-0500-AFetal Bovine Serum-US origin26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
YoungIn(OEM)500 mLUS-FBS-500Fetail Bovine Serum-US origin26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
GenDEPOT500 mLF0600-050Fetal Bovine Serum, US Origin, F060016000044Gibco Fetal Bovine Serum, certified, US origin, 500 mL (Premium FBS)
WEKGENE500 mLS 001-07Fetal Bovine Serum, Certified16000045Gibco Fetal Bovine Serum, certified, US origin, 500 mL (Premium FBS)
RMBIO500 mLFBS-BBTFBS, US origin26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
RMBIO500 mLFBS-CBTFBS, USDA Approved Origins10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
RMBIO500 mLFBS-ABT FBS, Australia and New Zealand10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
RMBIO500 mLFBS-ABT FBS, Australia and New Zealand10091148Gibco Fetal Bovine Serum, qualified, New Zealand origin, 500 mL (Premium FBS)
MPBio500 mL0929162CELLECT™ FBS, SILVER, HEAT INACTIVATED16140071Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 500 mL (Value Plus FBS)
MPBio500 mL0929168CELLECT™ FBS, GOLD, HEAT INACTIVATED16140071Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 500 mL (Value Plus FBS)
MPBio500 mL0929167 CELLect™ FBS, Gold, US Origin26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
MPBio100 mL02180735FBS, Silver, Uruguay Origin10270098Gibco Fetal Bovine Serum, qualified, Brazil origin, 100 mL (Value FBS)
MPBio500 mL0929161CELLECT™ FBS, SILVER, MEXICO ORIGIN10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
HiMedia500 mLRM9952Fetal Bovine Serum, USDA Approved10437028Gibco Fetal Bovine Serum, qualified, USDA-approved regions, 500 mL (Value FBS)
HiMedia500 mLRM9951Fetal Bovine Serum, Origin : Australia10099141Gibco Fetal Bovine Serum, qualified, Australia origin, 500 mL (Premium FBS)
HiMedia500 mLRM10409Fetal Bovine Serum, Origin : US, Heat inactivated10082147Gibco Fetal Bovine Serum, certified, heat inactivated, US origin, 500 mL (Premium FBS)
HiMedia500 mLRM9954Fetal Bovine Serum, Origin : Canada12483020Gibco Fetal Bovine Serum, qualified, Canada origin, 500 mL (Value FBS)
HiMedia500 mLRM9955Fetal Bovine Serum, EU Approved, Heat inactivated16140071Gibco Fetal Bovine Serum, qualified, heat inactivated, US origin, 500 mL (Value Plus FBS)
HiMedia500 mLRM10685Fetal Bovine Serum, Dialyzed26400044Gibco Fetal Bovine Serum, dialyzed, US origin, 500 mL (Specialty FBS)
HiMedia100 mLRM10432Fetal Bovine Serum, Origin: Brazil, EU Approved10270098Gibco Fetal Bovine Serum, qualified, Brazil origin, 100 mL (Value FBS)
HiMedia500 mLRM10416Fetal Bovine Serum, EU Approved, Charcoal treated12676029Gibco Fetal Bovine Serum, charcoal stripped, USDA-approved regions, 500 mL (Specialty FBS)
HiMedia500 mLRM10434Fetal Bovine Serum, Origin : US26140079Gibco Fetal Bovine Serum, qualified, US origin, 500 mL (Value Plus FBS)
other animal serum
Other animal sera
Although FBS is the most commonly used serum product, many other products are sold as a lower-cost alternative. These products include bovine serum, horse serum, newborn calf serum, goat serum, rabbit serum, porcine serum and chicken serum.
View all other animal sera
Recurring orders help take the worry out of reordering your favorite FBS products on time.  You can add FBS products you want to automatically reorder, tell us when you want them delivered, and we'll set up the orders for you.  Your freezer will thank you!
Save space now
What makes us recommended most by scientist worldwide
Click the buttons below and discover the benefits of Gibco fetal bovine serum products.
Gibco FBS
Resources
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.