Dermal Fibroblast Culture Systems
Optimized for your experiments
Our primary human dermal fibroblast cell culture systems are optimized to synthesize the extra cellular matrix and collagen, the structural framework for animal tissues that play a critical role in healing.
  • >500,000 cells/vial, cryopreserved at the end of 1' culture
  • >70% viability
  • >12 Adult or 16 Neonatal population doublings
  • Optimized for superior performance over leading competitors
Request more information    Ordering information
Phase contrast images of Human Dermal Fibroblast (HDFn) in culture
Fibroblast Cell
Day 3 - HDFn
Fibroblast Cell
Day 5 - HDFn
Fibroblast Cell
Day 7 - HDFn
HDFn cells grown in Medium 106 ( M-106-500) with the addition of Low Serum Growth Supplement ( LSGS). First culture after thaw.
Ordering Information
Catalog # Name Size Price (USD) Qty
C0135C Human Dermal Fibroblasts, adult (HDFa) Each
758.00
C0045C Human Dermal Fibroblasts, neonatal (HDFn) Each
715.65
Online Exclusive
758.00
Save 42.35 (6%)
M106500 Human Fibroblast Expansion Basal Medium (formerly Medium 106) Each
100.65
Online Exclusive
106.00
Save 5.35 (5%)
R01510 Gentamicin/Amphotericin Solution Each
117.00
R001100 Trypsin/EDTA Solution (TE) Each
41.30
R002100 Trypsin Neutralizer Solution Each
38.65
Online Exclusive
41.28
Save 2.63 (6%)
S00310 Low Serum Growth Supplement (LSGS) Each
126.00
S003K Low Serum Growth Supplement Kit (LSGS Kit) Each
145.00
Resources
Brochure: Key Products for Dermatological Cell Culture
Cell Culture & Transfection Learning Center
Access cell culture and transfection application resources for more success as you plan and execute your experiments.
For Research Use Only. Not for use in diagnostic procedures.