Mesenchymal Stem Cell Media
Gibco brand media provide the broadest selection of specialty products that are optimized for mesenchymal stem cell populations, many of which are free of animal derived components. These media are designed to minimize adaptation time, maximize cell performance, and meet regulatory requirements.
Gibco MSC Media are manufactured according to cGMP for medical devices, 21 CFR Part 820.
Reduced serum and serum free media
  • StemPro MSC SFM XenoFree: XenoFree media for human mesenchymal stem cell (MSC) culture
  • StemPro MSC SFM: a serum-free medium specially formulated for the growth and expansion of human mesenchymal stem cells (MSCs)
  • MesenPRO RS Medium:  Reduced serum (2%) medium for mesenchymal stem cell expansion
Ordering information
Catalog # Name Size Price (USD) Qty
A1067501 StemPro™ MSC SFM XenoFree Each
521.90
Special offer
Online exclusive
Ends: 21-Jun-2025
614.00
Save 92.10 (15%)
A1033201 StemPro™ MSC SFM, 1 Kit Each
472.82
Special offer
Online exclusive
Ends: 21-Jun-2025
528.00
Save 55.18 (10%)
12746012 MesenPRO RS™ Medium Each
265.09
Special offer
Online exclusive
Ends: 21-Jun-2025
307.00
Save 41.91 (14%)
Serum and serum replacement
Ordering information
Matrices and substrates
  • CELLstart Substrate: a defined substrate, containing only components of human origin (xeno-free) that supports hESCs attachment and expansion of undifferentiated colonies in serum-free medium without the need for feeder cells
Ordering information
Catalog # Name Size Price (USD) Qty
A1014201 CELLstart™ Substrate Each
374.85
Special offer
Online exclusive
Ends: 21-Jun-2025
441.00
Save 66.15 (15%)
Differentiation kits
Ordering information
Catalog # Name Size Price (USD) Qty
A1007001 StemPro™ Adipogenesis Differentiation Kit Each
238.39
Special offer
Online exclusive
Ends: 21-Jun-2025
264.00
Save 25.61 (10%)
A1007101 StemPro™ Chondrogenesis Differentiation Kit Each
249.81
Special offer
Online exclusive
Ends: 21-Jun-2025
275.00
Save 25.19 (9%)
A1007201 StemPro™ Osteogenesis Differentiation Kit Each
238.74
Special offer
Online exclusive
Ends: 21-Jun-2025
264.00
Save 25.26 (10%)
For Research Use Only. Not for use in diagnostic procedures.