E-Gel EX Gels
Ultra-sensitive detection of DNA
E-Gel EX gels were developed for ultimate sensitivity and demonstrate over 5-fold greater sensitivity than comparable gels containing ethidium bromide. The exceptional sensitivity allows you to use lower amounts of sample, saving time and money.
Order now  Circular RNA article
Benefits of E-Gel EX gels
  • Complete separation in as little as 10 minutes
  • Ultra-sensitive detection of DNA
  • Live monitoring of DNA migration without the hazards of UV light
  • Easy access to the gel for extraction of DNA bands
  • Quick check of RNA samples
Features of E-Gel EX gels
Take advantage of our E-Gel EX Double Comb Agarose Gels. These gels offer twice the number of sample wells per cassette compared to standard E-Gels, helping to reduce analysis limitations lower the cost per reaction.
Video Player is loading.
Current Time 0:00
Duration 1:32
Loaded: 0%
Stream Type LIVE
Remaining Time 1:32
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    E-Gel EX gels are the fastest, most sensitive precast agarose gels we offer for complete resolution of DNA or RNA samples typically in as little as 10 minutes. These gels are ideal for a quick check of the integrity of RNA samples or routine analysis of PCR products and plasmid preparations (Figure 1). E-Gel EX gels were developed for superior sensitivity over comparable gels containing ethidium bromide (Figure 2), allowing you to use less of your valuable sample.
    E-Gel EX gel image with one lane of ladder and 10 lanes of RNA samples.
    Figure 1. Check the integrity of your RNA samples using E-Gel EX gels. RNA samples were separated on a 2% E-Gel EX gel using the preset E-Gel EX program on the E-Gel iBase Power System. Lanes M, 5, and 10: 0.1–2 kb RNA Ladder; lanes 1–4, 6–9: mouse total RNA, 200 ng per lane.
    E-Gel EX gel image
    Figure 2. Sensitivity of DNA detection with E-Gel EX Gels. Two-fold serial dilutions of a 1 kb fragment, ranging from 1,000 ng to 1 ng, were separated on a 1% E-Gel EX Gel. Blue-light transillumination (with a Safe Imager Blue-Light Transilluminator) was used during documentation.
    The innovative design of the E-Gel EX gels allows the gel cassette to be opened and bands of choice to be easily excised. 
    Composite of two photos. Left: Researcher uses knife to dislodge gel from cassette. Right: Researcher holds gel.
    Follow these simple steps to excise your DNA band of choice:
    1. Place the cassette on a bench with the wells facing up
    2. Insert the sharp edge of the gel knife in the groove between the cassette halves, and lever the knife up and down
    3. Open the cassette and excise the band
    DNA electrophoresis has never been so simple. Your pre-cast agarose gel is ready to use, for any application and throughput. You can separate, purify, and recover your nucleic acid fragments right from the gel—fast.
    Take advantage of our E-Gel EX Double Comb Agarose Gels. These gels offer twice the number of sample wells per cassette compared to standard E-Gels, helping to reduce analysis limitations lower the cost per reaction.
    Video Player is loading.
    Current Time 0:00
    Duration 1:32
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:32
    1x
      • Chapters
      • descriptions off, selected
      • en (Main), selected
      E-Gel EX gels are the fastest, most sensitive precast agarose gels we offer for complete resolution of DNA or RNA samples typically in as little as 10 minutes. These gels are ideal for a quick check of the integrity of RNA samples or routine analysis of PCR products and plasmid preparations (Figure 1). E-Gel EX gels were developed for superior sensitivity over comparable gels containing ethidium bromide (Figure 2), allowing you to use less of your valuable sample.
      E-Gel EX gel image with one lane of ladder and 10 lanes of RNA samples.
      Figure 1. Check the integrity of your RNA samples using E-Gel EX gels. RNA samples were separated on a 2% E-Gel EX gel using the preset E-Gel EX program on the E-Gel iBase Power System. Lanes M, 5, and 10: 0.1–2 kb RNA Ladder; lanes 1–4, 6–9: mouse total RNA, 200 ng per lane.
      E-Gel EX gel image
      Figure 2. Sensitivity of DNA detection with E-Gel EX Gels. Two-fold serial dilutions of a 1 kb fragment, ranging from 1,000 ng to 1 ng, were separated on a 1% E-Gel EX Gel. Blue-light transillumination (with a Safe Imager Blue-Light Transilluminator) was used during documentation.
      The innovative design of the E-Gel EX gels allows the gel cassette to be opened and bands of choice to be easily excised. 
      Composite of two photos. Left: Researcher uses knife to dislodge gel from cassette. Right: Researcher holds gel.
      Follow these simple steps to excise your DNA band of choice:
      1. Place the cassette on a bench with the wells facing up
      2. Insert the sharp edge of the gel knife in the groove between the cassette halves, and lever the knife up and down
      3. Open the cassette and excise the band
      DNA electrophoresis has never been so simple. Your pre-cast agarose gel is ready to use, for any application and throughput. You can separate, purify, and recover your nucleic acid fragments right from the gel—fast.
      E-Gel EX Agarose Gels: Selection guide
      ProductStain#Sample wellsRun time (min)ResolutionCat. No.
      E-Gel EX Starter Kit, 1%SYBR Gold II1110100 bp–5 kbG8141ST
      E-Gel EX Starter Kit, 2%SYBR Gold II111050 bp–2 kbG8142ST
      E-Gel EX Gel, 1%, 10-PakSYBR Gold II1110100 bp–5 kbG401001
      E-Gel EX Gel, 2%, 10-PakSYBR Gold II111050 bp–2 kbG401002
      E-Gel EX Gel, 4%, 10-PakSYBR Gold II111010 bp-400 kbG410004
      E-Gel EX Double Comb Agarose Gels, 1% 10-PackSYBR Gold II225400 bp–5 kbA42345
      E-Gel EX Double Comb Agarose Gels, 1% 20-PackSYBR Gold II225400 bp–5 kbA44887
      E-Gel EX Double Comb Agarose Gels, 1% 50-PackSYBR Gold II225400 bp–5 kbA44888
      E-Gel EX Double Comb Agarose Gels, 2% 10-PackSYBR Gold II22550 bp–2 kbA42346
      E-Gel EX Double Comb Agarose Gels, 2% 20-PackSYBR Gold II22550 bp–2 kbA44889
      E-Gel EX Double Comb Agarose Gels, 2% 50-PackSYBR Gold II22550 bp–2 kbA44890
      5 min
      5 min
      5 min
      Education icon
       PCR enzymes education resources
      Access PCR learning resources—a knowledge base of all things PCR.
      Webinar icon
      Molecular biology webinars
      View our webinars for current date to learn more about molecular biology topics.
      Video icon
      PCR enzymes videos
      Discover product features and tips to optimize your PCR experiments.
      Tools icon
      Support center
      Find tips, troubleshooting help, and resources for your end-point PCR and cDNA applications.
      Computer mouse icon
      Contact us
      Email or call our Technical Application Scientists for questions regarding PCR enzymes and master mixes.
      Check icon
      OEM and custom PCR products
      Discover custom manufacturing solutions, product labeling, and packaging capabilities tailored to your specifications.
      Stylesheet for Classic Wide Template adjustments
      For Research Use Only. Not for use in diagnostic procedures.