E-Gel™ EX Agarose Gels
E-Gel™ EX Agarose Gels
Invitrogen™

E-Gel™ EX Agarose Gels

Invitrogen E-Gel EX precast agarose gels are designed for ultrasensitive and convenient DNA and RNA sample electrophoresis in as little as 10 to 20 minutes.
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySeparation RangeGel Percentage
G401002Promo Image10 Gels/Pk.50 to 2000 bp2%
G401001Promo Image10 Gels/Pk.100 to 5000 bp1%
G402021Promo Image20 Gels/Pk.100 to 5000 bp1%
G402022Promo Image20 Gels/Pk.50 to 2000 bp2%
G401004Promo Image10 Gels/Pk.20 to 800 bp4%
Catalog number G401002
Price (USD)
212.00
Special offer
Online exclusive
237.00
Save 25.00 (11%)
Each
Add to cart
Quantity:
10 Gels/Pk.
Separation Range:
50 to 2000 bp
Gel Percentage:
2%
Recurring order eligible. Learn more »
Price (USD)
212.00
Special offer
Online exclusive
237.00
Save 25.00 (11%)
Each
Add to cart
Ask our AI about this Product
Invitrogen E-Gel EX precast agarose gels are designed for ultrasensitive and convenient DNA and RNA sample electrophoresis in as little as 10 to 20 minutes. Each E-Gel agarose gel cassette contains all components and stain required for efficient gel separation and analysis–just load your samples and run. E-Gel agarose gels are ideal for analyzing PCR products, restriction digests, and plasmid preparations, or for DNA fragment library analysis. E-Gel agarose gels are also suitable for a quick check of RNA samples.

Features of E-Gel EX agarose gels include:

  • Instantaneous setup—precast E-Gel agarose gels are ready for use when you are, and do not require gel preparation or additional buffers
  • High sensitivity—detect 1 ng of sample DNA
  • Rapid analysis—complete DNA sample separation in 10–20 minutes
  • In-process control—view DNA sample migration in real time
  • Easy sample access—openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
  • For DNA and RNA—ideal for DNA sample analysis and suitable for quick check of RNA samples

Ultra-sensitive detection of DNA

E-Gel EX gels pre-stained with SYBR Gold DNA stain were developed for ultimate sensitivity and demonstrate up to 5-fold greater sensitivity than comparable gels containing ethidium bromide. This superior sensitivity allows you to use lower amounts of sample, saving time and money.

Fast and convenient analysis

E-Gel EX gels are our fastest-resolving agarose gels, enabling sample analysis in as little as 10–20 minutes. Each E-Gel agarose gel contains agarose, electrodes, SYBR Gold DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not required any gel or buffer preparation or gel straining steps. Just load your samples and run.

Live monitoring of DNA migration

The E-Gel EX agarose gels are pre-stained with SYBR Gold II DNA gel stain with an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, safe real-time monitoring of DNA or RNA migration is enabled without risk to eyes or possible damage of the sample, unlike UV illumination.

Quick check of RNA samples

Besides DNA analysis, E-Gel EX gels can also be used for fast analysis of RNA samples to check their integrity before proceeding with downstream applications.

Easy access to the gel

E-Gel cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), for easy excision of specific bands or transfer of the gel to a membrane for southern blot analysis.

Running and imaging devices

E-Gel EX gels require the Invitrogen E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.

Available in a variety of gel formats

E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 11-well E-Gel EX agarose gels are available in 1%, 2%, and 4% gel percentages.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible LaddersE-Gel™ 1 Kb Plus Express Ladder, Century™-Plus RNA Markers
Mode of SeparationMolecular Weight
Product TypeEX Agarose Gel
Quantity10 Gels/Pk.
Sample Buffer TypePreparative without dyes, Non-preparative
Sample Loading Volume20 μL
Sample TypeDouble-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA
Shipping ConditionRoom Temperature
Stain TypeSYBR™ Gold
Well DesignSingle Comb
Gel Percentage2%
Gel TypeE-Gel
Separation Range50 to 2000 bp
Wells11-well
Unit SizeEach
Contents & Storage
• E-Gel™ EX 10-Paks contain 10 gels

Store gels at room temperature. Guaranteed stable for 3 months when properly stored.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
T220625-01Certificate of AnalysisJun 24, 2025G401001
U120625-01Certificate of AnalysisJun 16, 2025G401002
U290525-01Certificate of AnalysisJun 12, 2025G401002
T030625-01Certificate of AnalysisJun 09, 2025G401001
H040625-02Certificate of AnalysisJun 08, 2025G401004
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection