icon
Cell culture contamination is easily the most common set-back encountered in cell culture laboratories, sometimes with very serious consequences. Cell culture contaminants can be divided into two main categories, chemical contaminants such as impurities in media, sera, and water, endotoxins, plasticizers, and detergents, and biological contaminants such as bacteria, molds, yeasts, viruses, mycoplasma, as well as cross contamination by other cell lines. While it is impossible to eliminate contamination entirely, it is possible to reduce its frequency and seriousness by gaining a thorough understanding of their sources and by following proper aseptic technique.
How to identify contamination in cell culture
To identify cell culture contamination, you must have knowledge of the morphology of cells you are studying and the potential contaminants the culture may have been exposed to. With this information, you can decide which methods will be most effective for testing and monitoring your cell cultures for contamination. This is essential to ensure that you catch any potential contamination before it becomes unmanageable. 
However, some biological contaminants, such as mycoplasma, can be extremely difficult to detect even with regular monitoring and require alternative identification and treatment.
Learn more about mycoplasma contamination
    Cell culture contamination can be identified by performing tests such as cell analysis through microscopy, microbial testing, cell line authentication, and immunostaining. Various methods may be necessary to determine which contaminant is present or if there are multiple contaminants present in the cell culture. 
    A culture should be tested for contamination if you suspect that it was exposed to a potential contaminant. Testing for contaminants should also be routine procedure in your regular culturing protocols. It is recommended to test cell lines or primary cultures prior to beginning an experiment and to routinely analyze all cell cultures for potential contamination.
    Bacterial contamination in cell culture
    Bacteria are a large and ubiquitous group of unicellular microorganisms. They are typically a few micrometers in diameter and can have a variety of shapes ranging from spheres to rods and spirals. Bacteria are one of the most common contaminants in cell culture because of their ubiquity, size, and fast growth rates.
    Bacterial contamination in cell cultures is easily detected by visual inspection within a few days of it becoming infected.
    • Infected cultures usually appear cloudy (i.e., turbid), sometimes with a thin film on the surface.
    • Sudden drops in the pH of the culture medium are also frequently encountered.
    • Under a low-power microscope, the bacteria appear as tiny, moving granules between the cells, and observation under a high-power microscope can resolve the shapes of individual bacteria.
     The simulated images below show an adherent 293 cell culture contaminated with E. coli.
    Two panels with simulated phase contrast images of adherent 293 cells contaminated with E. coli
    Figure 1. Simulated phase contrast images of adherent 293 cells contaminated with E. coli. The spaces between the adherent cells show tiny, shimmering granules under low power microscopy, but the individual bacteria are not easily distinguishable (panel A). Further magnification of the area enclosed by the black square resolves the individual E. coli cells (panel B), which are typically rod-shaped and are about 2 µm long and 0.5 µm in diameter. Each side of the black square in panel A is 100 µm.
    Viral contamination in cell culture
    Viruses are microscopic infectious agents dependent on a host cell for replication. Their extremely small size makes it difficult to detect and remove them from cultures and reagents. Since most viruses have very stringent requirements for their host, they usually do not adversely affect cell cultures from species other than their host. However, using virally infected cell cultures can present a serious health hazard to the laboratory personnel, especially if human or primate cells are cultured in the laboratory.
    Viral infection of cell cultures can be detected by electron microscopy, immunostaining with a panel of antibodies, ELISAs, or PCR with appropriate viral primers.
    Fungal contamination in cell culture
    Fungi are widespread eukaryotic organisms that can be uni- or multi-cellular. To reproduce, fungi produce spores that are released and can grow in many different nutrient-dense environments—including cell culture medias and matrices. The most common fungal contamination in cell culture comes from molds and yeasts.
    Yeast contamination
    Yeasts are unicellular eukaryotic microorganisms in the kingdom of Fungi, ranging in size from a few micrometers (typically) up to 40 µm (rarely).
    Like bacteria, yeast contamination in cell culture turns the medium turbid, especially if the contamination is in an advanced stage. There is very little change in the pH of a culture contaminated by yeast until the contamination becomes heavy—at which stage the pH usually increases. Under microscopy, yeast appears as individual ovoid or spherical particles, that may bud off smaller particles.
    Simulated phase contrast image of yeast contamination in cell culture of 293 cells
    Figure 2. Simulated phase contrast images of 293 cells 24 hours after plating in adherent culture that is contaminated with yeast. The contaminating yeast cells appear as ovoid particles, budding off smaller particles as they replicate. 
    Mold contamination
    Molds are eukaryotic microorganisms in the kingdom of Fungi that grow as multicellular filaments called hyphae. A connected network of these filaments contains genetically identical nuclei and are called a colony or mycelium. 
    Similar to yeast, mold contamination in cell culture presents with a stable pH level in the initial stages of contamination, then rapidly increases as the culture becomes more heavily infected and turbid. Under microscopy, the mycelia usually appear as thin, wisp-like filaments, and sometimes as denser clumps of spores. Spores of many mold species can survive extremely harsh and inhospitable environments in their dormant stage, only to become activated when they encounter suitable growth conditions.
    Using antibiotics and antimycotics in cell culture
    Antibiotics and antimycotics should not be used routinely in cell culture, because their continuous use encourages the development of antibiotic-resistant strains and allows low-level contamination to persist, which can develop into full-scale contamination once the antibiotic is removed from media and may hide mycoplasma infections and other cryptic contaminants. Further, some antibiotics might cross react with the cells and interfere with the cellular processes under investigation.
    Antibiotics should only be used as a last resort and only for short term applications, and they should be removed from the culture as soon as possible. If they are used in the long term, antibiotic-free cultures should be maintained in parallel as a control to help investigate cryptic infections.
    The decision to use antibiotics to prevent cell culture contamination should be based on the individual researcher's needs and experience. The following table is a general guide for use of Gibco antibiotics in cell culture media. Solutions that use one or more antibiotics in conjunction with an antimycotic are also available. For all media types, optimal concentrations of antibiotics and antimycotics should be determined empirically. Please always reference antibiotic and antimycotic product information sheets to help determine working concentrations.
    Decontaminating cell cultures
    When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.
    • First, determine if the cell culture contamination is bacteria, fungus, mycoplasma, or yeast. 
    • Isolate the contaminated culture from other cell lines.
    • Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
    • Antibiotics and antimycotics at high concentrations can be toxic to some cell lines; therefore, perform a dose response test to determine the level at which an antibiotic or antimycotic becomes toxic.
    The following is a suggested procedure for determining toxicity levels and decontaminating cultures:
    1. Dissociate, count, and dilute the cells in antibiotic-free medium. Dilute the cells to the concentration used for regular cell passage.
    2. Dispense the cell suspension into a multi-well culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. 
    3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
    4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
    5. Culture the cells for one passage in antibiotic-free media.
    6. Repeat step 4.
    7. Culture the cells in antibiotic-free medium for 4 to 6 passages to determine if the contamination has been eliminated.
    Cross-contamination
    While not as common as microbial contamination, extensive cross-contamination of many cell lines with other fast growing cell lines is a clearly established problem with serious consequences. Obtaining cell lines from reputable cell banks, periodically checking the characteristics of the cell lines, and practicing good aseptic technique are practices that will help you avoid cross-contamination. DNA fingerprinting, karyotype analysis, and isotype analysis can confirm the presence or absence of cross-contamination in your cell cultures.
    Why, when, and how to passage cells grown in both adherent and suspension cultures, including preparing new culture vessels for passaged cells.
    Reference
    Resources
    Cell Culture & Transfection Learning Center
    Access cell culture and transfection educational resources to enable excellent experiment planning and execution.
    Gibco Cell Culture Basics
    Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
    Cell Culture Videos
    Explore this collection of cell culture videos providing valuable insights into techniques, resources, and stay up to date with the latest advancements in the field.
    Cell Culture Selection Tools
    Find the right cell culture products for your research with these selection tools. Search for media, FBS, plastics, transfection reagents, and more.
    Support
    Cell Culture & Transfection Support Center
    Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
    Need technical support? 
    Contact our expert team for technical and application support of Laboratory Products.
    Related products
    For Research Use Only. Not for use in diagnostic procedures.
    Stylesheet for Classic Wide Template adjustments