Claisen rearrangement reaction
In 1912, German chemist Rainer Ludwig Claisen published the rearrangement of allyl phenyl ethers to their corresponding C-allyl phenols, as well as the conversion of the O-allylated acetoacetic ester to its C-allylated isomer upon treatment with ammonium chloride, followed by distillation. The thermal rearrangement of allyl vinyl ethers to their corresponding γ,δ-unsaturated carbonyl compounds has since been known as the Claisen rearrangement.
The precursor allyl vinyl ethers can be prepared in several ways, such as from the allylic alcohols via mercuric ion-catalyzed exchange with ethyl vinyl ether, or by Wittig olefination of allyl formates and carbonyl compounds.
A modification is the Johnson-Claisen rearrangement, in which an allylic alcohol is heated with trialkyl orthoacetate under mildly acidic conditions to produce a γ,δ-unsaturated ester.
The Claisen rearrangement has been used in many successful total synthesis campaigns, including K.C. Nicolaou and colleagues' synthesis of 1-O-methylforbesione via tandem Claisen rearrangement/Diels-Alder reactions.
Review available Thermo Scientific products for the Claisen rearrangement reaction:
Catalog # Name Size Price (USD) Qty
A15000.30 Ammonium chloride, 98+% Each
31.65
Special offer
Online exclusive
Ends: 30-Jun-2025
36.90
Save 5.25 (14%)
A15000.0B Ammonium chloride, 98+% Each
41.65
Special offer
Online exclusive
Ends: 26-Sep-2025
48.50
Save 6.85 (14%)
A15000.0I Ammonium chloride, 98+% Each
127.65
Special offer
Online exclusive
Ends: 26-Sep-2025
150.00
Save 22.35 (15%)
A11766.30 Allyl bromide, 99%, stab. with 300-1000ppm Propylene oxide Each
38.65
Special offer
Online exclusive
Ends: 26-Sep-2025
45.70
Save 7.05 (15%)
A11766.0B Allyl bromide, 99%, stab. with 300-1000ppm Propylene oxide Each
166.00
A15691.AK Ethyl vinyl ether, 99%, stab. Each
28.65
Online Exclusive
32.20
Save 3.55 (11%)
A15691.AP Ethyl vinyl ether, 99%, stab. Each
43.65
Special offer
Online exclusive
Ends: 30-Jun-2025
50.70
Save 7.05 (14%)
A15691.0F Ethyl vinyl ether, 99%, stab. Each
137.65
Special offer
Online exclusive
Ends: 26-Sep-2025
162.00
Save 24.35 (15%)
L06004.AE Triethyl orthoacetate, 97% Each
38.65
Special offer
Online exclusive
Ends: 26-Sep-2025
44.90
Save 6.25 (14%)
L06004.AP Triethyl orthoacetate, 97% Each
117.65
Online Exclusive
130.00
Save 12.35 (10%)
L06004.0F Triethyl orthoacetate, 97% Each
333.65
Special offer
Online exclusive
Ends: 30-Jun-2025
392.00
Save 58.35 (15%)
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now