UltraPure™ TBE Buffer, 10X
UltraPure™ TBE Buffer, 10X
Invitrogen™

UltraPure™ TBE Buffer, 10X

UltraPure™ 10X TBE Buffer is a sterile-filtered solution of 1 M Tris, 0.9 M boric acid, and 0.01 M EDTA深入閱讀
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
155810441 L
1558102810 L
產品號碼 15581044
價格 (TWD)
1,710.00
線上優惠
Ends: 30-Jun-2025
2,190.00
您節省 480.00 (22%)
Each
-
新增至購物車
Quantity:
1 L
Recurring order eligible. Learn more »
Request bulk or custom format
價格 (TWD)
1,710.00
線上優惠
Ends: 30-Jun-2025
2,190.00
您節省 480.00 (22%)
Each
新增至購物車
Ask our AI about this Product

UltraPure™ 10X TBE Buffer is a sterile-filtered solution of 1 M Tris, 0.9 M boric acid, and 0.01 M EDTA used to prepare 1X buffer for polyacrylamide and agarose gel electrophoresis. UltraPure™ 10X TBE Buffer is available in a 1 L plastic bottle or in a 10 L Cubitainer™ box.

Performance and quality testing: No DNase, RNase, or protease activity detected.

For Research Use Only. Not for use in diagnostic procedures.
規格
BufferRunning Buffer
Concentration10 X
For Use With (Application)Nucleic Acid Gel Electrophoresis, Blotting
Gel CompatibilityPolyacrylamide Gels, Agarose Gels
Product LineUltraPure™
Product TypeTBE Buffer
Quantity1 L
Shipping ConditionRoom Temperature
FormatBottle
Unit SizeEach
內容物與存放
Contents: One bottle (1 L) UltraPure™ 10X TBE Buffer
Storage: Room Temperature
Have questions about this product? Ask our AI assisted search.
What is the difference between ProLong and SlowFade antifade reagents?
What do ProLong Gold, ProLong Diamond and ProLong Glass antifade mountant reagents contain?
Can I use ProLong Gold, ProLong Diamond, or ProLong Glass mounting medium in a multi-well plate?
My dye is photobleaching too fast with confocal imaging. What can I do?
What happens when ProLong, ProLong Gold, ProLong Diamond, or ProLong Glass antifade mountant is curing? Is there a chemical reaction?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



文件與下載

證書

批號Certificate TypeDateCatalog Number(s)
3205034Certificate of AnalysisJun 05, 202515581044
3192262Certificate of AnalysisMay 28, 202515581028
3127877Certificate of AnalysisMay 11, 202515581044
3137180Certificate of AnalysisApr 12, 202515581044
3127874Certificate of AnalysisMar 29, 202515581044
顯示 5 個結果, 在上方搜尋特定憑證

安全資料表

常見問答集 (常見問題)

The precipitation of concentrated TBE stocks may be due to nucleation of salt crystals by dust particles or other insoluble materials. Therefore, filtering the solution through a 0.2 µm cellulose acetate or cellulose nitrate filter after preparation helps avoid this precipitate. [Mayeda A, Krainer A (1991) BioTechniques 10.2, 1820]

引用資料與參考文獻 (3)

引用資料與參考文獻
Abstract
Semiquantitative analysis of gene expression in cultured chondrocytes by RT-PCR.
Authors:Rolland-Valognes G,
Journal:Methods Mol Med
PubMed ID:15280588
'Reverse transcriptase-polymerase chain reaction (RT-PCR) is a powerful, sensitive, and rapid method to monitor small amounts of nucleic acids. This is of particular interest for small amounts of cells, as in cartilage. We present here two protocols to isolate total RNA and a protocol to study matrix metalloproteinase and type ... More
A pancoronavirus RT-PCR assay for detection of all known coronaviruses.
Authors:Vijgen L, Moës E, Keyaerts E, Li S, Van Ranst M,
Journal:Methods Mol Biol
PubMed ID:19057882
'The recent discoveries of novel human coronaviruses, including the coronavirus causing SARS, and the previously unrecognized human coronaviruses HCoV-NL63 and HCoV-HKU1, indicate that the family Coronaviridae harbors more members than was previously assumed. All human coronaviruses characterized at present are associated with respiratory illnesses, ranging from mild common colds to ... More
Isolating aptamers using capillary electrophoresis-SELEX (CE-SELEX).
Authors:Mosing RK, Bowser MT,
Journal:Methods Mol Biol
PubMed ID:19377982
SELEX (systematic evolution of ligands by exponential enrichment) is a process for isolating DNA or RNA sequences with high affinity and selectivity for molecular targets from random sequence libraries. These sequences are commonly referred to as aptamers. The process typically requires 10-15 cycles of enrichment, PCR amplification and nucleic acid ... More
3 total citations

Other products to consider



分享目錄編號、產品名稱或連結。

1x1 image pixel for data collection