1x1 image pixel for data collection
Newborn Calf Serum, New Zealand origin
Newborn Calf Serum, New Zealand origin
Gibco™

Newborn Calf Serum, New Zealand origin

Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
16010159500 mL
160101421 L
16010167100 mL
Catalog number 16010159
Price (USD)
170.00
Each
In stock
Add to cart
Quantity:
500 mL
Price (USD)
170.00
Each
Add to cart
Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
  • Sera Category: Secure
  • Origin: New Zealand
  • Collected from calves 20 days old or less.
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
Shipping ConditionFrozen
SpeciesCattle/Bovine
AgeNewborn (20 days or less)
Country of OriginNew Zealand
FormLiquid
FormatBottle
Product TypeNewborn Calf Bovine Serum
Quantity500 mL
Serum TreatmentStandard (Sterile-filtered)
SterilitySterile
Unit SizeEach
Contents & Storage
Storage conditions: ≤-10°C
Shipping conditions: Frozen

Frequently asked questions (FAQs)

Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection