Nunc™ Lab-Tek™ Chamber Slide System
Nunc™ Lab-Tek™ Chamber Slide System
Thermo Scientific™

Nunc™ Lab-Tek™ Chamber Slide System

Grow anchorage dependent cells right on the microscope slide using the unique removable chamber of the Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide System.
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWellsMaterialUnit Size
177429Promo Image2Permanox PlasticCase of 96
177372Promo Image1Soda Lime GlassCase of 96
177372PKPromo Image1Soda Lime GlassPack of 16
177380Promo Image2Soda Lime GlassCase of 96
177380PKPromo Image2Soda Lime GlassPack of 16
177399Promo Image4Soda Lime GlassCase of 96
177399PKPromo Image4Soda Lime GlassPack of 16
177402Promo Image8Soda Lime GlassCase of 96
177402PKPromo Image8Soda Lime GlassPack of 16
177410Promo Image1Permanox PlasticCase of 96
177410PKPromo Image
also known as 177410
1Permanox PlasticPack of 16
177429PKPromo Image
also known as 177429
2Permanox PlasticPack of 16
177437Promo Image4Permanox PlasticCase of 96
177437PKPromo Image4Permanox PlasticPack of 16
177445Promo Image8Permanox PlasticCase of 96
177445PKPromo Image8Permanox PlasticPack of 16
178599Promo Image16Soda Lime GlassCase of 96
178599PKPromo Image16Soda Lime GlassPack of 16
Catalog number 177429
Price (USD)
1,359.65
Online Exclusive
1,400.00
Save 40.35 (3%)
Case of 96
Estimated availability date  Pending  
Add to cart
Wells:
2
Material:
Permanox Plastic
Unit Size:
Case of 96
Recurring order eligible. Learn more »
Price (USD)
1,359.65
Online Exclusive
1,400.00
Save 40.35 (3%)
Case of 96
Add to cart
Ask our AI about this Product

Grow, stain, and examine cells directly on the Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide™, which consists of a polystyrene media chamber attached to a microscopic slide. Slide surface is specially treated for adherent cell culture. Choose either specialty washed Soda Lime Glass slide or cell attachment optimized Permanox Plastic slide. Media chamber is easily removable without the need for a tool.

Features:
  • The solvent-resistant Permanox™ Plastic Chamber Slides are available in 1-, 2-, 4- and 8-well formats.
  • The specialty washed Soda Lime Glass Chamber Slides are available in 1-, 2-, 4-, 8- and 16-well formats.
  • Media chambers can be removed from slides without the need for a tool
  • Medical-grade non-toxic silicone sealing gasket can be removed with a pair of pointy tweezers before applying coverslip.
  • "Sand blasted" writing area provides easy identification of samples.
  • Coverglass 171080 is to be used with the 16-well glass Chamber Slide 178599.
  • Ordering Information:

    • Can be purchased in case of 96 or pack of 16.
    • Shipped sterile in convenient stackable incubation trays.
  • Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorClear
Description2-well Chamber Slide w/ removable wells
IncludesClear polystyrene lid with handle on the side for aseptic manipulations
Lid MaterialPolystyrene
No. per Pack16
Culture Area4.2 cm2
Dimensions (LxW)75 x 25 mm
MaterialPermanox Plastic
SterilitySterile
Surface TreatmentNunclon Delta
Volume (Metric) Working1.2 to 2.0 mL
Wells2
Unit SizeCase of 96
ECM
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
1302667Certificate of ComplianceDec 11, 2024177402
1327530Certificate of ComplianceDec 11, 2024177445
1328336Certificate of ComplianceDec 11, 2024177372
1323017Certificate of ComplianceDec 11, 2024177402
1323015Certificate of ComplianceDec 11, 2024177437
5 results displayed, search above for a specific certificate

Frequently asked questions (FAQs)

Unfortunately, we do not have a recommended cell density for our LabTek Chamber Slides. However, you can refer to the following link, which provides recommended cell densities for our other cell culture products:
https://www.thermofisher.com/us/en/home/references/gibco-cell-culture-basics/cell-culture-protocols/cell-culture-useful-numbers.html

Please review the following key differences between the Nunc Lab-Tek Chamber Slide System and the Nunc Lab-Tek II Chamber Slide System:
- The Nunc Lab-Tek Chamber Slide System is available in Permanox plastic or glass and incorporates a silicone gasket.
- The Nunc Lab-Tek II Chamber Slide System is available in glass only and do not have a silicone gasket.
- The Nunc Lab-Tek II Chamber Slide System has a blue hydrophobic border and the upper structure is attached by an adhesive. Supplied with this product is a key and holder system which is used to remove the upper structure from the glass slide.
- The Nunc Lab-Tek Chamber Slide System is available in 1-, 2-, 4-, 8- or 16-well formats.
- The Nunc Lab-Tek II Chamber Slide System is available in 1-, 2-, 4- or 8-well formats.

The following are possible causes and solutions:
- The cells could be stressed due to media substitution, especially from serum-free formulations, or other changes to the culture environment such as in temperature, humidity, and CO2 percentage. If possible, always plate cells in serum-containing medium. Switching cells to a serum-free media should be done gradually.
- The cells could be plated at very low densities. Most cell lines have a minimal platting density below which normal cell proliferation cannot occur, and therefore should be avoided.
- The cells could be poorly adherent. Treatment of cells that may reduce adhesion and therefore, should not be done immediately after plating, but after the cells have resumed exponential growth.

The upper structure of our Nunc Lab-Tek Chamber Slide products are made of polystyrene so it is not compatible with many fixation reagents. These chamber slide products are designed for upper structure and gasket removal before fixing, staining, and coverslipping.

Our Nunc Lab-Tek Chamber Slide products are used for fluorescence analyses at the microscopic level.
The following are commonly used excitation and emittance wavelengths:
FITC
- Excitation: 420-490 nm
- Emittance: 520-550 nm
Rhodamine
- Excitation: 510-560 nm
- Emittance: 570-610 nm
Hoechst
- Excitation: 330-380 nm
- Emittance: 480-510 nm

Share catalog number, name or link

1x1 image pixel for data collection