Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous
Thermo Scientific Chemicals

Sodium phosphate, dibasic, 99+%, for analysis, anhydrous

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 204851000
Price (USD)
31.65
Online Exclusive
35.00
Save 3.35 (10%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
31.65
Online Exclusive
35.00
Save 3.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or granular powder
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
Sulfate (SO4)=<50 ppm
Loss on drying=<0.2 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471476Certificate of AnalysisMay 17, 2025204855000, 204850050, 204851000
A0471477Certificate of AnalysisMay 17, 2025204855000, 204850050, 204851000
A0467782Certificate of AnalysisJan 31, 2025204855000, 204850050, 204851000
A0467783Certificate of AnalysisJan 31, 2025204855000, 204850050, 204851000
A0453969Certificate of AnalysisAug 25, 2023204855000, 204850050, 204851000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link