MEM, glutamine, HEPES, powder
MEM, glutamine, HEPES, powder
Gibco™

MEM, glutamine, HEPES, powder

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be usedRead more
Have Questions?
Catalog NumberQuantity
2101104450 L
Catalog number 21011044
Price (USD)
624.00
Each
-
Add to cart
Quantity:
50 L
Recurring order eligible. Learn more »
Customize this product
Price (USD)
624.00
Each
Add to cart
Ask our AI about this Product
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM is modified as follows:

With Without
• L-glutamine • Sodium Bicarbonate
• Phenol Red
• HEPES

The complete formulation is available.

Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow’s MEM, MEM α, DMEM, and Temin’s Modification. MEM is available with Earle’s salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle’s salts.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

cGMP Manufacturing and Quality System
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Powder forms of Gibco™ L-15 cell culture medium require pH adjustment and filtration at the time of preparation (see protocol for details).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Culture EnvironmentCO2
Osmolality240 - 280 mOsm/kg
Product LineGibco™
Product TypeMinimum Essential Medium (MEM)
Quantity50 L
Shelf Life36 Months
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
Culture TypeMammalian Cell Culture
FormPowder
Serum LevelStandard Serum Supplementation
With AdditivesGlutamine, HEPES, Phenol Red
Without AdditivesNo Sodium Bicarbonate, No Sodium Pyruvate
pH5.2 to 5.6
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C.
Shipping conditions: Ambient.
Shelf life: 36 months from date of manufacture.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3112389Certificate of AnalysisMar 14, 202521011044
3122279XCertificate of AnalysisFeb 01, 202521011044
2973100Certificate of AnalysisOct 18, 202421011044
2973098Certificate of AnalysisOct 18, 202421011044
3053067Certificate of AnalysisOct 18, 202421011044
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection