Pierce™ IgG Elution Buffer, pH 2.0
Pierce™ IgG Elution Buffer, pH 2.0
Thermo Scientific™

Pierce™ IgG Elution Buffer, pH 2.0

Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such asRead more
Have Questions?
Catalog NumberQuantity
210281 L
Catalog number 21028
Price (USD)
109.65
Online Exclusive
126.00
Save 16.35 (13%)
Each
In stock
Add to cart
Quantity:
1 L
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
109.65
Online Exclusive
126.00
Save 16.35 (13%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads. The Pierce IgG Elution Buffer provides efficient dissociation and elution of antibodies from all three IgG-binding proteins, including Protein L. This low-pH elution buffer is also effective for most antibody-antigen or protein-protein affinity purification systems.

Shared features of all Thermo Scientific Pierce IgG Binding and Elution Buffers:

Ready to use—carefully formulated and prefiltered 1X solutions
Three binding buffers—optimized for Protein A, Protein G or Protein A/G
Two elution buffer options—acidic (pH 2.8) or high salt (near-neutral)
Versatile—elution buffers are appropriate for antibody-antigen affinity purification

Products available for each antibody purification method:
Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA
Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA
Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide
IgG Elution Buffer: pH 2.8; amine-based
Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide
Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)

Related Products
Pierce™ IgG Elution Buffer
Pierce™ Gentle Ag/Ab Elution Buffer, pH 6.6
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LinePierce™
Quantity1 L
FormLiquid
Product TypeElution Buffer
Solution TypeElution Buffer
Unit SizeEach
Contents & Storage
Store at 4°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3214634Certificate of AnalysisJul 10, 202521028
3212525Certificate of AnalysisMay 19, 202521028
AC406572Certificate of AnalysisMar 28, 202521028
AB405509Certificate of AnalysisFeb 14, 202521028
AA403519Certificate of AnalysisJan 31, 202521028
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link