Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
21083027 | 500 mL |
Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.
With | Without |
• Galactose | • Glucose |
• L-glutamine | • Phenol Red |
• Sodium Pyruvate | • HEPES |
• Sodium Bicarbonate |
The complete formulation is available.
Using L-15
L-15 contains no proteins, lipids, or growth factors. Therefore, L-15 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).
We do provide osmolality information on the certificate of analysis. All lots of Leibovitz's L-15 Medium will meet the osmolality specification of 300-340 mOsm/kg.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
No, Leibovitz L15 medium (liquid or powder) does not contain sodium bicarbonate (the formulation for each catalog number is published online). Developed for the support of cell growth in non CO2-equilibrated environments, L-15 formulations were developed without a sodium bicarbonate buffer system. L-15 medium is buffered by phosphates and free-base amino acids (L-arginine primarily, but also L-histidine and L-cysteine). Galactose and sodium pyruvate are used to replace glucose in order to reduce the production of acidic metabolites.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
We offer the following Leibovitz media:
- Leibovitz's L-15 Medium, powder: Cat. Nos. 41300039 (10x 1L) and 41300070 (1x 10L)
- Leibovitz's L-15 Medium, no phenol red: Cat. No 21083027 (1x 500 mL)
- Leibovitz's L-15 Medium: Cat. Nos.11415064 (1x 500 mL) and 11415114 (10x 500 mL) (for sale in N. America, Latin America, and Asia-Pacific regions; Cat. No. 11415049 (for sale in Europe, Africa, and the Middle East
- Leibovitz's L-15 Medium with GlutaMAX Supplement: Cat. No. 31415029 (1x 500 mL)
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.
Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.
We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.
Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.
Share catalog number, name or link