PageRuler™ Unstained Broad Range Protein Ladder
PageRuler™ Unstained Broad Range Protein Ladder
Thermo Scientific™

PageRuler™ Unstained Broad Range Protein Ladder

Thermo Scientific PageRuler Unstained Broad Range Protein Ladder is a mixture of 11 proteins (5 to 250 kDa) for useRead more
Promo ImagePromotion
Promo code:RBLOT25
Save up to 24% on Western Blot Reagents and Gels
From now until June 30th, get 16% off when you spend $400 or more, and get 24% off when you spend $750 or more on select western blot reagent and protein gels.Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
26630Promo Image2 x 250 μL
26630X48 x 250 μL
Catalog number 26630
Price (USD)
130.65
Online Exclusive
155.00
Save 24.35 (16%)
Each
In stock
Add to cart
Quantity:
2 x 250 μL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
130.65
Online Exclusive
155.00
Save 24.35 (16%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific PageRuler Unstained Broad Range Protein Ladder is a mixture of 11 proteins (5 to 250 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Compare and view all other protein standards and ladders ›

Product features
• Reference bands—the 100, 50, and 20 kDa bands are more intense for easy orientation
• Tagged—bands 10, 15, 20, 30, 40, 50, 70, 100, 150, and 250 contain an integral Strep-tag II sequence and can be detected on western blots using Strep-Tactin conjugates or an antibody against Strep-tag II sequence

Applications
• Accurate protein sizing on SDS-polyacrylamide gels and western blots

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel CompatibilityBolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol)250, 150, 100, 70, 50, 40, 30, 20, 15, 10, 5 kDa
Product LinePageRuler™
Product TypeProtein Ladder
Quantity2 x 250 μL
Ready to LoadYes
Shipping ConditionApproved for shipment on Wet or Dry Ice
Stain TypeUnstained
System TypeWestern Blotting, SDS-PAGE
Number of Markers11
Size Range5 to 250 kDa
Unit SizeEach
Contents & Storage
Contents: two vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol

Storage: Upon receipt store at -20°C

Have questions about this product? Ask our AI assisted search.
When should unstained or prestained ladder/marker be used?
Do Thermo Scientific protein ladders have glycosylated proteins?
What is the composition of the PageRuler Unstained Broad Range Protein Ladder?
Are any animal-derived proteins present in the different PageRuler Unstained Protein Ladders?
Can I use any of your protein standards for estimation of protein quantity (protein quantitation)?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

The upper bands of the ladder are missing. What could be the reason?

The upper bands of the ladder may be degraded by proteases. Ladder, gel, buffer, pipettes, pipette tips, or equipment can be contaminated by proteases during usage. A general recommendation would be to avoid working with proteases in the same room. We would recommend preparing fresh solutions, cleaning the equipment, and using clean pipettes and tips. If the ladder itself is contaminated, please use a new tube of the ladder.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Some additional bands or smears were observed on the gel when using a PageRuler unstained ladder. What may have caused this?

Additional bands can appear due to dithiothreitol (DTT) oxidation in the storage buffer. Please add newly prepared DTT solution to the final concentration of 100 mM and boil for 5 min at 95 degrees C. This should solve the issue. Addition of DTT is NOT recommended for prestained protein ladders, since too high a concentration of reducing agents can cause protein destaining.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Do the proteins in Thermo Scientific protein ladders have a His-Tag or would otherwise react with an anti-His-Tag antibody?

No, proteins in Thermo Scientific protein ladders are not His tagged. However, non-specific interaction between the ladder proteins and primary or secondary antibodies is possible and some His-Tag detection systems, such as Thermo Scientific 6xHis Protein Tag Stain Reagent Kit, show non-specific interaction. The protein ladder bands are more readily detected when using high antibody concentrations. The non-specific cross-reactivity is difficult to predict, it often has a different pattern dependent on the antibodies used in each individual experiment. The most general way to handle this problem would be to use lower concentrations of antibodies and to use lower amount of protein ladders. It may also be useful to leave one empty well between the ladder and the sample to overcome a possible leakage of the signal to the nearby sample lane.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can Thermo Scientific protein ladders be detected by Strep-Tactin conjugates?

PageRuler Unstained protein ladders can be detected directly on Western blots by using Strep-Tactin conjugates or an antibody against the Strep-tag II sequence. All PageRuler and Spectra ladder proteins contain an integral Strep-tag II sequence, however the prestained ladders cannot be detected by Strep-Tactin conjugates.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Why is non-specific binding detected after Western blot?

Protein ladder bands can sometimes be detected with chemiluminescent techniques due to non-specific interactions of ladder proteins with either primary or secondary antibodies (or with both). The ladder bands are only rarely detected by chromogenic substrates. The extremely high sensitivity of the chemiluminescent assays is needed to see the bands, so the actual degree of cross-reactivity is low. The non-specific cross-reactivity is difficult to predict, it often has a different pattern depending on the antibodies used. If antibodies recognize a linear epitope, the cross-reactivity may be due to sequence homology. If antibodies react with a denaturation-resistant conformational epitope it could be impossible to identify the exact reason for detected cross-reactivity. The most general way to handle this problem would be to use lower concentrations of antibodies.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Figures

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3277146Certificate of AnalysisJul 09, 202526630
3248467Certificate of AnalysisMay 28, 202526630
3227115Certificate of AnalysisApr 28, 202526630
3218513Certificate of AnalysisApr 15, 202526630
3194051Certificate of AnalysisMar 20, 202526630
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link