RPMI 1640 Medium, no glutamine, no phenol red
Save up to 29% on your order of cell culture products. View promotion details >
RPMI 1640 Medium, no glutamine, no phenol red
Gibco™

RPMI 1640 Medium, no glutamine, no phenol red

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as aRead more
Have Questions?
Catalog NumberQuantity
32404014500 mL
Catalog number 32404014
Price (USD)
36.31
Each
-
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Customize this product
Price (USD)
36.31
Each
Add to cart
Ask our AI about this Product

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This RPMI 1640 is modified as follows:
Without
• L-glutamine
• HEPES
• Phenol Red


The complete formulation is available.

Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12, and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineHeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell TypeLeukemic Cells
Concentration1 X
Culture EnvironmentCO2
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality260 - 300 mOsm/kg
Product LineGibco™
Product TypeRPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesSodium Bicarbonate
Without AdditivesNo Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.
Have questions about this product? Ask our AI assisted search.
What is the density (g/L) for RPMI 1640 Medium?
Should I be concerned about phenol red in my media when labeling my live cells with fluorescent dyes?
I understand that some media are worse than others for fluorescence imaging. How do I choose?
Do you offer media without phenol red (phenol red-free media)?
How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3144444Certificate of AnalysisMay 10, 202532404014
3090879Certificate of AnalysisMar 16, 202532404014
3023267Certificate of AnalysisFeb 19, 202532404014
2555063Certificate of AnalysisJan 29, 202532404014
2549528Certificate of AnalysisJan 29, 202532404014
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Most media contain phenol red, which can quench fluorescent dyes in the visible wavelengths. Most media also contain autofluorescent components, such as riboflavin, which can reduce signal-to-background. We offer FluoroBrite DMEM and HEPES-based Live Cell Imaging Solution, which have been optimized for fluorescent imaging. We also offer a number of media without phenol red. But if none of these are reasonable options for your experiment, then we also offer BackDrop Background Suppressor ReadyProbes Reagent, which can be added to quench media autofluorescence.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Some cell types accumulate phenol red, and this can pose a problem in the use of many fluorescent probes. Phenol red can quench visible-wavelength dyes and, although phenol red is non-fluorescent, various impurities may be fluorescent. We have many phenol red-free media to choose from. Our Live Cell Imaging Solution (HEPES-based) and our FluoroBrite DMEM have been optimized to be phenol red-free as well as to be non-autofluorescent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection