3500xL Genetic Analyzer 24-Capillary Array, 36 cm
3500xL Genetic Analyzer 24-Capillary Array, 36 cm
Applied Biosystems™

3500xL Genetic Analyzer 24-Capillary Array, 36 cm

The internally uncoated capillaries are supplied in preassembled sets of 24 capillaries per array, with a built-in frame for easyRead more
Have Questions?
Catalog NumberQuantity
44046871 Array
Catalog number 4404687
Price (USD)
3,015.00
Each
-
Add to cart
Quantity:
1 Array
Price (USD)
3,015.00
Each
Add to cart
Ask our AI about this Product
The internally uncoated capillaries are supplied in preassembled sets of 24 capillaries per array, with a built-in frame for easy installation.

Applications of the 36 cm capillaries:
HID⁄ Forensic Applications.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capillary Length36 cm
Description24-Capillary Array, 36cm Capillary, Internally Uncoated Capillaries, Human Identification Applications, For Use With 3500xL Genetic Analyzer, Room Temperature
For Use With (Application)Sequencing, Human Identification
For Use With (Equipment)3500xL Genetic Analyzer
Product Type3500xL Genetic Analyzer
Quantity1 Array
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to your genetic analyser's maintenance, troubleshooting and reference guide.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
M317F12Certificate of AnalysisJan 04, 20184404687
M317H13Certificate of AnalysisJan 03, 20184404687
M317G05Certificate of AnalysisJan 02, 20184404687
M317E11Certificate of AnalysisDec 29, 20174404687
M317H11Certificate of AnalysisDec 29, 20174404687
5 results displayed, search above for a specific certificate

Frequently asked questions (FAQs)

If the array is stuck in the pump, even after loosening the knob, squirt around the edge of the array head with distilled water and try to remove it again. If the array will not move, a service call may be required. Do not use excessive force or tools to try and remove it.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

If we do not specify the expiry date or number of uses, the warranty will last for 12 months from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

The Applied Biosystems 3500/3500xl Genetic Analyzer capillary arrays are warrantied for a specific number of runs or until the expiration date printed on the array package, whichever comes first. If you notice that you are starting to get reduced data quality (e.g., shorter reads, trailing peaks in all 4 colors) even with the Sequencing Standard, these may be signs that the array requires changing.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

Thermo Fisher Scientific can only provide warranty on performance on non-expired reagents. Using reagents past their expiration date is a risk and may compromise your data integrity.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

Share catalog number, name or link

1x1 image pixel for data collection