1x1 image pixel for data collection
Reagent Reservoirs
Reagent Reservoirs
Thermo Scientific™

Reagent Reservoirs

Choose these reagent reservoirs, designed to provide a variety of solutions to suit all of your pipetting procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)MaterialSterilityPackaging
9512809525 mLPolystyreneNon-sterile10 Bags of 10
8096-1125 mLPlasticSterileIndividually Wrapped
8093-1125 mLPolystyreneSterile10 Bags of 10
95128085100 mLPolystyreneNon-sterile10 Bags of 10
9512809325 mLPolystyreneNon-sterileBulk
Catalog number 95128095
Price (USD)
255.61
Online Exclusive
320.00
Save 64.39 (20%)
Case of 100
In stock
Add to cart
Capacity (Metric):
25 mL
Material:
Polystyrene
Sterility:
Non-sterile
Packaging:
10 Bags of 10
Recurring order eligible. Learn more »
Price (USD)
255.61
Online Exclusive
320.00
Save 64.39 (20%)
Case of 100
Add to cart
Ask our AI about this Product

Thermo Scientific™ Reagent Reservoirs provide a variety of solutions to suit all of your pipetting procedures.

Innovation goes beyond our pipetters and tips. Even our reagent reservoirs have been designed to provide solutions tailored to your procedures, rather than the other way around. Four variations of reservoirs are offered, 25mL, 25mL with divider, 100mL and a disposable polystyrene 75mL reservoir.

Benefits

  • Trough within a trough maximizes the amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
  • 25mL Divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizes recovery if using only single channel and small amount of reagent
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
ColorWhite
Packaging10 Bags of 10
Capacity (Metric)25 mL
DisposableSingle-use
MaterialPolystyrene
SterilityNon-sterile
TypeReagent Reservoir with Divider
Unit SizeCase of 100
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2501012Certificate of ComplianceJun 26, 202595128093
2430612Certificate of ComplianceMay 22, 202595128095
2404312Certificate of ComplianceMay 22, 202595128095
2329212Certificate of ComplianceMay 22, 202595128095
2431312Certificate of ComplianceMay 22, 202595128093
5 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection