FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent
FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent
Gibco™

FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent

FoamAway™ Irradiated AOF is a ready-to-use antifoaming agent designed to offer high performance and convenience in combating foam in yourRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A10369012.5 L bag
A10369020.5 L in a 1 L bag
Catalog number A1036901
Price (USD)
840.00
Each
-
Add to cart
Quantity:
2.5 L bag
Recurring order eligible. Learn more »
Price (USD)
840.00
Each
Add to cart
Ask our AI about this Product
FoamAway™ Irradiated AOF is a ready-to-use antifoaming agent designed to offer high performance and convenience in combating foam in your cell culture system. It is prediluted, presterilized, and packaged to connect directly to your bioreactor with no further processing required.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
ClassificationAnimal Origin-Free
Culture TypeSuspension Cell Culture
For Use With (Application)Cell Culture
FormLiquid
Phenol Red IndicatorNo Phenol Red
Product LineGibco™
Product TypeAntifoaming Agent
Quantity2.5 L bag
Shipping ConditionRoom Temperature
SterilityGamma-irradiated
Volume (Metric)2.5 L
Unit SizeEach
Contents & Storage
Store at room temperature. Shake well before use. Do not filter.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3130481Certificate of AnalysisJun 21, 2025A1036902, A1036901
3130466Certificate of AnalysisJun 20, 2025A1036902, A1036901
3130451Certificate of AnalysisJun 04, 2025A1036902, A1036901
3130472Certificate of AnalysisMay 30, 2025A1036902, A1036901
3130475Certificate of AnalysisMay 30, 2025A1036902, A1036901
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

The target shipping percentage of simethicone in FoamAway Irradiated AOF (animal origin-free) Antifoaming Agent is 3%. This supports the defoaming assay noted on the COA for the shelf life of the product.

FoamAway Irradiated AOF (animal origin-free) Antifoaming Agent is formulated to a 4.0% simethicone concentration during manufacture, representing the upper end of the product's concentration range, to account for loss of simethicone during processing.
The final concentration of simethicone may be variable in the finished product, but is ≤4.0%. The final product must pass a defoaming activity assay as a quality release criterion, ensuring defoaming activity throughout the 8-month shelf life.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

Unacceptable defoam activity was observed after 8 months. Therefore, the shelf life was reduced from 12 to 8 months from the date of irradiation in 2019.

The connectors are male and female luer locks.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

We recommend shaking to thoroughly mix the product before each addition. Approximately 1:1,000 dilution may be the midrange of the working concentration, about 30 ppm. The range may be about a 1:20,000 to 1:600 depending on the process. For an extended culture, we recommend re-adding material during the run as it is incorporated with the foaming components; we recommend adding as little as possible to keep the downstream processes functioning.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Los productos recomendados



Share catalog number, name or link

1x1 image pixel for data collection