Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified
This product is a replacement for Geltrex™ hESC-qualified Reduced Growth Factor Basement Membrane Matrix (SKU A1048002)
Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified
Gibco™

Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified

Geltrex LDEV-free stem cell-qualified reduced-growth factor basement membrane matrix is a soluble form of basement membrane extracted from murine Engelbreth-Holm-SwarmRead more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A1413302Promo Image5 mL
A1413301Promo Image1 mL
Catalog number A1413302
Price (USD)
388.00
Each
Add to cart
Quantity:
5 mL
Recurring order eligible. Learn more »
Price (USD)
388.00
Each
Add to cart
Ask our AI about this Product

Geltrex LDEV-free stem cell-qualified reduced-growth factor basement membrane matrix is a soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors and is free of viruses, including lactose dehydrogenase elevating virus (LDEV) sometimes found in this type of preparation. Routinely used for attachment and maintenance of human iPSCs or embryonic stem cells, every lot of this matrix is quality control tested using hESCs or iPSCs to help ensure growth and pluripotency of each lot.

Features of Geltrex Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem cell-qualified, include:

  • Every lot qualified using hESC or iPSC, helping reduce the need for customers to lot screen
  • Free of lactose dehydrogenase elevating virus (LDEV), making it safe for all types of cell culture
  • Consistent protein concentration lot-to-lot, helping ensure reproducible results

Gain reliability and consistency in your hESC/iPSC studies

Human hESCs/iPSCs have specific needs for attachment and maintenance and some lots of basement membrane extract may not provide adequate support. Geltrex Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem cell-qualified, has been tested for support of hESC/iPSC growth and pluripotency, minimizing the need to test multiple lots to find one that works. Our consistent manufacturing process helps ensure a protein concentration with very little variation from lot-to-lot, so you can have confidence in every purchase.

Free of LDEV contamination

Recent advances in processing of EHS tumor cells have resulted in virus-free basement membrane extracts. Every lot is tested by tested by RT-PCR methods so you can be confident that every lot is virus-free.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHuman induced pluripotent stem cells (iPSC), primary epithelial cells, endothelial cells, smooth muscle cells
Concentration12 to 18 mg/mL
Endotoxin LevelLow
FormatTube(s)
Product LineGibco™
Product TypeBasement Membrane Matrix
Purity or Quality GradeCell Culture Grade
Quantity5 mL
Shelf Life36 months
Shipping ConditionDry Ice
SpeciesMouse
ClassificationReduced Growth Factor
Culture TypeAdherent Cell Culture
FormFrozen
Serum LevelSerum-free
SterilitySterile
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
1019080CCertificate of AnalysisFeb 06, 2025A1413301, A1413302
1019080BCertificate of AnalysisFeb 06, 2025A1413301, A1413302
1019080ACertificate of AnalysisFeb 06, 2025A1413301, A1413302
1019082DCertificate of AnalysisFeb 05, 2025A1413301, A1413302
1019082ECertificate of AnalysisFeb 05, 2025A1413301, A1413302
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

No, the Ready-To-Use Geltrex matrix was developed for use with the StemPro hESC SFM System. It is not recommended for other PSC media systems.

For other PSC media systems, we recommend the Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413301, A1413302).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601) is a pre-diluted Geltrex product ready to use for coating culture vessels. There is no need to dilute the solution further. The product has also been QC tested using hESCs to ensure that every lot will maintain the attached pluripotency of these cells. The full product description can be found here:

Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601)

Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413301, A1413302) is a "concentrated" version of the Geltrex hESC-Qualified, Ready-To-Use above. You can pre-dilute this concentrated solution to pre-coat the culture vessel to grow hESCs at your desired usage concentration. This Geltrex matrix has also been QC tested using hESCs to ensure every lot will maintain the attached pluripotency of these cells. The full product description can be found here:

Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413301, A1413302)

Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413201, A1413202) is the same as the product above. However, it has not been tested to verify it will support hESC growth and expansion. This is a "general" version and not specific to a cell type or line. A full product description can be found here:

Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413201, A1413202)

Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413201, A1413202)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

We recommend thawing Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. No. 1413301, 1413302) at 4 degrees C before making your stock solution. On ice, make a 1:1 dilution with DMEM/F-12 and aliquot into smaller volumes. Stock solution aliquots should be stored at -20 degrees C or used immediately. Once the stock solution is thawed, it should be used for coating plates immediately.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

We do not have comparison data for the two Geltrex products. We recommend using the Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix for hepatocytes. We have not performed a significant amount of testing using the Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix on these cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

We recommend using Gibco Geltrex LDEV-Free, hESC Qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413301 or A1413302) or Gibco Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A14700) for xeno-free applications.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (8)

Citations & References
Abstract
A comprehensive protocol for efficient differentiation of human NPCs into electrically competent neurons.
Authors:Romito E,Battistella I,Plakhova V,Paplekaj A,Forastieri C,Toffolo E,Musio C,Conti L,Battaglioli E,Rusconi F
Journal:Journal of neuroscience methods
PubMed ID:39053772
Characterizing Pluripotent Stem Cells Using the TaqMan(®) hPSC Scorecard (TM) Panel.
Authors:Fergus J, Quintanilla R, Lakshmipathy U,
Journal:
PubMed ID:25138722
'Rapid technological developments for the efficient generation of footprint-free induced pluripotent stem cells (iPSC) enabled the creation of patient-specific iPSC for downstream applications in drug discovery and regenerative medicine. However, the large number of iPSCs, generated from diverse genetic backgrounds using various methods and culture conditions, created a steep challenge ... More
CRISPR/Cas9 Genome Editing of Human-Induced Pluripotent Stem Cells Followed by Granulocytic Differentiation.
Authors:Dannenmann B, Nasri M, Welte K, Skokowa J
Journal:Methods Mol Biol
PubMed ID:32006418
'Research on patient-derived induced pluripotent stem cells (iPSCs) could immensely benefit from the implementation of CRISPR/Cas9 genome editing of iPSCs, creating unique opportunities such as the establishment of isogenic iPSC lines for disease modeling or personalized patient-specific drug screenings. Here we describe a stepwise protocol of safe, efficient, and selection-free ... More
A Protocol for One-Step Differentiation of Human Induced Pluripotent Stem Cells into Mature Podocytes.
Authors:Murphy C, Feifel E, Jennings P, Gstraunthaler G, Wilmes A
Journal:Methods Mol Biol
PubMed ID:31124107
'Within the glomerulus, podocytes are highly specialized visceral epithelial cells that are part of the glomerular filtration barrier. Human podocyte cell culture is rather challenging for primary or immortalized cells, due to the nonproliferative state of the cells. In addition, rapid dedifferentiation is often observed. Hence, iPSC-derived podocytes offer an ... More
Kinetochore Proteins Have a Post-Mitotic Function in Neurodevelopment.
Authors:Zhao G, Oztan A, Ye Y, Schwarz TL
Journal:Dev Cell
PubMed ID:30827899
'The kinetochore is a complex of proteins, broadly conserved from yeast to man, that resides at the centromere and is essential for chromosome segregation in dividing cells. There are no known functions of the core complex outside of the centromere. We now show that the proteins of the kinetochore have ... More
8 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection