Geltrex™ hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix
Taking your work to the clinic? Check out CTS Recombinant Vitronectin »
Geltrex™ hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix
Gibco™

Geltrex™ hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix

Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane extracted from murine Engelbreth-Holm-SwarmRead more
Have Questions?
Catalog NumberQuantity
A156960150 mL
Catalog number A1569601
Price (USD)
117.00
Each
Add to cart
Quantity:
50 mL
Recurring order eligible. Learn more »
Price (USD)
117.00
Each
Add to cart
Ask our AI about this Product
Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors. This pre-diluted version of the matrix can be taken directly from the refrigerator and dispensed into cell culture plates without further handling. Geltrex hESC-Qualified Ready-To-Use basement membrane matrix is optimized for stem cell culture applications using StemPro hESC SFM. For stem cell culture using the StemFlex Medium or Essential 8 Media System, use Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix.

Geltrex hESC-Qualified Ready-To-Use matrix is:

• Consistent—pre-dilution at a concentration of 1:100 ensures optimal protein concentration for stem cell attachment to standard cell culture plates
• Stable—storage at 4°C eliminates overnight thawing on ice
• Cost effective— the ready-to-use format saves multiple preparation steps and potential for product failure

Reduced variability
Geltrex hESC-Qualified Ready-To-Use matrix is provided as a 1:100 dilution in a buffered solution that can be added directly to cell culture plates for coating. Each lot is prepared at the appropriate protein concentration for optimal stem cell attachment, which minimizes the need to calculate dilution schemes for every lot of matrix.

Reliable
Designed for 4°C storage for the life of the product, this matrix can be kept in the refrigerator along with your cell culture media—no more overnight thawing in clumsy ice baths and no risk of premature gelling. In addition, every lot is functionally qualified using human embryonic stem cells and tested to ensure it is free of lactose dehydrogenase elevating virus (LDEV), making it safe for all cell culture labs.

Cost effective
Geltrex hESC-Qualified Ready-To-Use matrix eliminates multiple steps—no more freezing, thawing, diluting, and aliquoting. This product is a true time saver.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeStem Cells (Hematopoietic, Neural, Mesenchymal, Mouse, Embryonic, iPS - Induced Pluripotent Stem, Human)
Concentration0.12 to 0.18 mg/mL
Endotoxin LevelLow
FormatBottle(s)
Product LineGeltrex™
Product TypeBasement Membrane Matrix
Purity or Quality GradeES Cell-Qualified
Quantity50 mL
Shelf Life18 Months
SpeciesMouse
ClassificationReduced Growth Factor
Culture TypeAdherent Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
Why can Gibco Geltrex Matrix be used for adhesion-related research, as opposed to Gibco AlgiMatrix 3D Culture System which cannot?
Has Gibco Geltrex Matrix been tested with StemPRO hESC serum-free Medium?
Can differentiation of cells occur when using Gibco Geltrex Matrix? Can this be prevented?
What is Gibco Geltrex Matrix?
What is the main application for the Ready-to-Use Gibco Geltrex Matrix?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can I use the Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601) with other PSC media systems (i.e. StemFlex, Essential 8, Essential 8 Flex, etc.)?

No, the Ready-To-Use Geltrex matrix was developed for use with the StemPro hESC SFM System. It is not recommended for other PSC media systems.

For other PSC media systems, we recommend the Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413301, A1413302).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between Geltrex LDEV-Free and Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement?

Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601) is a pre-diluted Geltrex product ready to use for coating culture vessels. There is no need to dilute the solution further. The product has also been QC tested using hESCs to ensure that every lot will maintain the attached pluripotency of these cells. The full product description can be found here:

Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601)

Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413301, A1413302) is a "concentrated" version of the Geltrex hESC-Qualified, Ready-To-Use above. You can pre-dilute this concentrated solution to pre-coat the culture vessel to grow hESCs at your desired usage concentration. This Geltrex matrix has also been QC tested using hESCs to ensure every lot will maintain the attached pluripotency of these cells. The full product description can be found here:

Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413301, A1413302)

Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413201, A1413202) is the same as the product above. However, it has not been tested to verify it will support hESC growth and expansion. This is a "general" version and not specific to a cell type or line. A full product description can be found here:

Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413201, A1413202)

Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413201, A1413202)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the Gentamicin sulfate concentration in Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601)?

The Gentamycin sulfate concentration in Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601) is 10 µg/mL.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What dilution should I use for Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601)?

For stem cells, a 1:100 dilution is the recommended starting dilution for Geltrex hESC, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601). Other (lower) dilutions may be necessary for other primary cell types and should be determined empirically.

Do you have a protocol for coating 96-well plates with Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix?

Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix is in principle a diluted version of the frozen Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix.

Please take a look at page 2 under section “Recover frozen PSCs in complete StemFlex Medium” in the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0016431_StemFlexMediumKit_UG.pdf) for the volume of Geltrex to use to cover a 6-well, 12-well, or 24-well plate.

Basically,
One well of a 24-well plate has a surface area of 2 cm2 and you need 300 µL of diluted Geltrex per well.
One well of a 48-well plate has a surface area of 1 cm2 and you need 150 µL of diluted Geltrex per well.
One well of a 96-well plate has a surface area of 0.32 cm2 and you need 48 µL (˜50 µL) of diluted Geltrex per well.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
1020559ACertificate of AnalysisJul 11, 2025A1569601
1020559CCertificate of AnalysisJul 11, 2025A1569601
1020559FCertificate of AnalysisJul 09, 2025A1569601
1020559DCertificate of AnalysisJul 09, 2025A1569601
1020559ECertificate of AnalysisJul 09, 2025A1569601
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (3)

Citations & References
Abstract
Differentiation of hepatocyte-like cells from human pluripotent stem cells using small molecules.
Authors:Asumda FZ, Hatzistergos KE, Dykxhoorn DM, Jakubski S, Edwards J, Thomas E, Schiff ER
Journal:
PubMed ID:29626713
'A variety of approaches have been developed for the derivation of hepatocyte-like cells from pluripotent stem cells. Currently, most of these strategies employ step-wise differentiation approaches with recombinant growth-factors or small-molecule analogs to recapitulate developmental signaling pathways. Here, we tested the efficacy of a small-molecule based differentiation protocol for the ... More
The arrhythmogenic potential of nerve agents and a cardiac safety profile of antidotes - A proof-of-concept study using human induced pluripotent stem cells derived cardiomyocytes (hiPSC-CM).
Authors:Amend N, Thiermann H, Worek F, Wille T
Journal:Toxicol Lett
PubMed ID:30858091
The global use of organophosphorus compounds (OP) for pest control and nerve agents being used in military conflicts and for assassinations renders intoxications by these agents a public health concern. OP-poisoned patients often suffer from dysrhythmias which may ultimately result in death. In this study, human-induced pluripotent stem cells derived ... More
Modeling of early neural development in vitro by direct neurosphere formation culture of chimpanzee induced pluripotent stem cells.
Authors:Kitajima R, Nakai R, Imamura T, Kameda T, Kozuka D, Hirai H, Ito H, Imai H, Imamura M
Journal:Stem Cell Res
PubMed ID:32151953
Evolutionary developmental biology of our closest living relative, the chimpanzee (Pan troglodytes), is essential for understanding the origin of human traits. However, it is difficult to access developmental events in the chimpanzee in vivo because of technical and ethical restrictions. Induced pluripotent stem cells (iPSCs) offer an alternative in vitro ... More
3 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection