High Select™ Depletion Spin Columns
High Select™ Depletion Spin Columns
High Select™ Depletion Spin Columns
High Select™ Depletion Spin Columns
Thermo Scientific™

High Select™ Depletion Spin Columns

Decrease albumin, immunoglobulins, and other high-abundance proteins from human serum or plasma in preparation for mass spectrometry analysis, or 1D and 2D gel electrophoresis using High Select depletion spin columns or resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct TypePurification Target
A363696 ColumnsMini Spin ColumnTop14 Abundant Protein
A363656 ColumnsMini Spin ColumnHSA/Immunoglobulin
A3636624 ColumnsMini Spin ColumnHSA/Immunoglobulin
A3636710 ColumnsMidi Spin ColumnHSA/Immunoglobulin
A3636850 mLResinHSA/Immunoglobulin
A3637110 ColumnsMidi Spin ColumnTop14 Abundant Protein
A3637250 mLResinTop14 Abundant Protein
A3637024 ColumnsMini Spin ColumnTop14 Abundant Protein
Catalog number A36369
Price (USD)
320.65
Online Exclusive
358.00
Save 37.35 (10%)
Each
-
Add to cart
Quantity:
6 Columns
Product Type:
Mini Spin Column
Purification Target:
Top14 Abundant Protein
Price (USD)
320.65
Online Exclusive
358.00
Save 37.35 (10%)
Each
Add to cart
Ask our AI about this Product
Analysis of human fluids is often complicated by the presence of high concentrations of serum proteins, including albumin and IgG that can make up more than 70% of total serum protein. Removal of albumin, immunoglobulins, and other abundant proteins is essential for the study of low-abundance proteins. Thermo Scientific High Select HSA/Immunoglobulin and Top 14 Abundant Protein depletion spin columns and resins enable depletion of the most abundant proteins in human serum and plasma for improved detection of low-abundance proteins using mass spectrometry analysis or 1D or 2D gel electrophoresis.
Flexible—resin provided in mini and midi column formats as well as bulk for custom filling options
Optimized—removes >95% of IgG and >95% of albumin (HSA/Immunoglobulin); removes >95% of IgG and >95% of albumin, plus 12 other abundant proteins (Top 14)
Fast—process samples in ∼10 minutes
Economical—cost-effective spin columns are priced for single use
Reduced carry-over—one-time use prevents protein carryover and experimental variability

High Select HSA/Immunoglobulin Depletion Resin uses highly specific, immobilized anti-HSA and anti-immuoglobulin antibodies (IgG, IgA, IgM, IgD, and IgE) to remove human serum albumin (HSA) and all major subclasses of immunoglobulins from serum and plasma. High Select Top14 Abundant Protein Depletion Resin uses highly specific immobilized antibodies to remove human serum albumin (HSA), albumin, IgG, IgA, IgM, IgD, IgE, kappa and lambda light chains, alpha-1-acidglycoprotein, alpha-1-antitrypsin, alpha-2-macroglobulin, apolipoprotein A1, fibrinogen, haptoglobin, and transferrin from serum, plasma, or spinal fluids. The resins are provided in an economical and convenient spin column format specifically designed for one-step processing and single use or bulk format for custom filling.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Label or DyeUnlabeled
Purification TargetTop14 Abundant Protein
Quantity6 Columns
Shipping ConditionWet Ice
Workflow StepProtein Depletion
FormatMini Spin Column
Product LineHigh Select™
Product TypeMini Spin Column
Starting MaterialPlasma, Serum
Unit SizeEach
Contents & Storage
Store at 2–8°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3231092Certificate of AnalysisJun 17, 2025A36368
3231085Certificate of AnalysisMay 28, 2025A36372
3212670Certificate of AnalysisMay 21, 2025A36366
3212656Certificate of AnalysisMay 12, 2025A36372
3212436Certificate of AnalysisApr 24, 2025A36370
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link