MagMAX™ DNA Multi-Sample Ultra 2.0 Kit
Elevate your research with expanded sample type claims. Check out new KingFisher validated workflows.
MagMAX™ DNA Multi-Sample Ultra 2.0 Kit
Applied Biosystems™
MagMAX™ DNA Multi-Sample Ultra 2.0 Kit
The MagMAX DNA Multi-Sample Ultra 2.0 Kit is an automatable magnetic bead-based sample preparation technology for the isolation of genomicRead more
Promo ImagePromotion
Promo code:RDNAP25
Get up to 19% Off Premium DNA Purification Solutions!
Save on our wide range of DNA purification solutions, including column-based extraction kits, magnetic beads, organic extraction reagents, and other essentials.Learn More
Have Questions?
Catalog NumberQuantity
A36570Promo Image1 Kit
Catalog number A36570
Price (USD)
599.65
Online Exclusive
658.00
Save 58.35 (9%)
Each
In stock
Add to cart
Quantity:
1 Kit
Recurring order eligible. Learn more »
Price (USD)
599.65
Online Exclusive
658.00
Save 58.35 (9%)
Each
Add to cart
Ask our AI about this Product
The MagMAX DNA Multi-Sample Ultra 2.0 Kit is an automatable magnetic bead-based sample preparation technology for the isolation of genomic DNA (gDNA) from 50 μL to 2 mL of whole blood, saliva, buffy coat, and buccal swabs. The resulting purified gDNA is ideal for a variety of downstream molecular biology applications such as qPCR, next-generation sequencing, and microarray analysis. One kit is sufficient to process 200 50–200 μL samples or 100 200–400 μL samples except for buffy coat samples. For buffy coat only, one kit is sufficient to process 40–60 samples at 50–200 uL input volume. To process up to 200 buffy coat samples, please purchase additional Elution Solution (Cat. No. A36582).
Key features of the MagMAX DNA Multi-Sample Ultra 2.0 Kit:
• Process 6–96 samples in 45 minutes with 5 minutes hands-on time
• Isolate gDNA from 50 μL–2 mL of sample using a simple protocol in SBS microtiter plates
• Skip sample and buffer volume normalization steps for hassle-free processing
• Download validated BindIt software automation scripts for KingFisher Duo Prime and Flex purification systems
• Automation-friendly stand-alone MagMAX DNA Multi-Sample Ultra 2.0 reagent bottle options available
Isolate genomic DNA from multiple sample types with a single sample prep kit
The MagMAX DNA Multi-Sample Ultra 2.0 Kit isolates gDNA from 50 μL–2 mL of whole blood (figure below), fresh and stabilized saliva (figure below), and EDTA/Streck cfDNA tube buffy coat (figure below) samples. Additionally, the same kit can be used to isolate gDNA from buccal swabs and other biological sample types. The MagMAX DNA Multi-Sample Ultra 2.0 Kit maximizes gDNA yields by treating samples with proteinase K (PK) and PK-enhancer followed by an innovative lysis/binding solution to simultaneously lyse cells, denature proteins, and capture gDNA on the high-binding capacity Ultra 2.0 gDNA magnetic binding beads. The Ultra 2.0 Wash I solution removes proteins, detergents, and other impurities from the bound nucleic acid. The gDNA is then released from the beads using a low-TE elution solution.
Fast and hassle-free MagMAX magnetic bead-based gDNA workflows
The MagMAX DNA Multi-Sample Ultra 2.0 Kit combines multiple laborious sample processing steps together into a single step to minimize the time and effort traditionally required by other methods without compromising yield and purity. Process a broad range of sample volume inputs simultaneously without sample normalization or reagent volume adjustments for hassle-free sample prep. Simply match your sample volumes to one of three standard input workflows (50 μL to 400 μL, up to 1 mL, up to 2 mL) and follow the work instructions for either manual or semi-automated sample preparation.
Made for KingFisher purification systems
The MagMAX DNA Multi-Sample Ultra 2.0 Kit is optimized for use on the moderate-throughput KingFisher Duo Prime (Cat. No. 5400110) and the higher-throughput KingFisher Flex (Cat. No. 5400630) purification systems. Users can process up to 400 μL of sample volume in high-throughput SBS 96-well deep-well microtiter plates using the Flex system. For smaller sample batches, users can process 12 samples using the Duo Prime system. Larger sample input volumes (1–2 mL), which are more challenging to process, are purified in SBS 24-well deep-well microtiter plates for either 24 samples on the Flex or 6 samples on the Duo Prime system. KingFisher instrument BindIt software scripts for all sample types and both Flex and Duo Prime systems can be downloaded below and installed onto the KingFisher instruments.
Working with higher throughput sample numbers?
The same MagMAX DNA Multi-Sample Ultra 2.0 Kit reagents are available as automation-friendly stand-alone reagents. MagMAX DNA Multi-Sample Ultra 2.0 stand-alone reagents are individually bottled in larger reagent volumes to make your high-thoughput workflows easier to handle. When purchased together as a set of six stand-alone reagents, users will have sufficient volume of MagMAX DNA Multi-Sample Ultra 2.0 reagents to process 2000 50–200 uL samples or 1000 200–400 uL samples.
MagMAX DNA Ultra Multi-Sample 2.0 Proteinase K (45 mL)
MagMAX DNA Ultra Multi-Sample 2.0 Binding Beads (45 mL)
MagMAX DNA Ultra Multi-Sample 2.0 Wash I Solution (1.1 L)
MagMAX DNA Ultra Multi-Sample 2.0 Lysis/Binding Solution (450 mL)
MagMAX DNA Ultra Multi-Sample 2.0 Elution Solution (120 mL)
MagMAX DNA Ultra Multi-Sample 2.0 Enhancer Solution (45 mL)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
Product LineMagMAX™
Product TypeDNA Multi-Sample Ultra 2.0 Kit
Quantity1 Kit
Shipping ConditionRoom Temperature
FormatKit
Unit SizeEach
Contents & Storage
• 4.5 mL MagMAX DNA Multi-Sample Ultra 2.0 Proteinase K, store at room temperature
• 4.5 mL MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads, store at room temperature
• 110 mL MagMAX DNA Multi-Sample Ultra 2.0 Wash I Solution, store at room temperature
• 45 mL MagMAX DNA Multi-Sample Ultra 2.0 Lysis/Binding Solution, store at room temperature
• 12 mL MagMAX DNA Multi-Sample Ultra 2.0 Elution Solution, store at room temperature
• 4.5 mL MagMAX DNA Multi-Sample Ultra 2.0 Enhancer Solution, store at room temperature
Have questions about this product? Ask our AI assisted search.
What is the level of detection for somatic variants with the Oncomine Myeloid Research Assay GX?
Are the DNA Binding Beads included within the MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (Cat. No. A36570) available as a stand-alone product and in a smaller volume?
What sample controls should I use with the Oncomine Myeloid Research Assay GX?
Saliva samples can be processed with the MagMAX DNA Multi-Sample Ultra 2.0 Kit as well as MagMAX Saliva gDNA Isolation Kit. What is the difference between these kits and which one should I use?
How many primer pools are present in the Oncomine Myeloid Research Assay GX?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Figures
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
3269858Certificate of AnalysisJun 27, 2025A36570
3266868Certificate of AnalysisJun 23, 2025A36570
3266814Certificate of AnalysisJun 23, 2025A36570
3226941Certificate of AnalysisApr 29, 2025A36570
3226834Certificate of AnalysisApr 28, 2025A36570
5 results displayed, search above for a specific certificate
Safety Data Sheets
Scientific Resources
Product Information
Frequently asked questions (FAQs)
We would recommend using one of our MagMAX DNA Multi-Sample Ultra 2.0 Kit or ChargeSwitch gDNA Buccal Cell Kits, which were designed to isolate high-quality genomic DNA from buccal samples. We also offer normalized kits, which will normalize isolated DNA concentration from sample to sample.
We recommend the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) that is optimized for use on the moderate-throughput KingFisher Duo Prime Purification System (Cat. No. 5400110) and the higher-throughput KingFisher Flex Purification System, KingFisher with 96 Deep-well head (Cat. No. 5400630).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
The difference between Proteinase K for MagMAX‐96 DNA Multi‐Sample Kit and Proteinase K for MagMAX DNA Multi‐Sample Ultra Kit is the brand. The former is from Invitrogen legacy and the latter is from Applied Biosystems legacy. Both products have an identical concentration of 100 mg/mL and can be used interchangeably.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center
We offer the MagMAX DNA Multi-Sample Ultra 2.0 Binding Beads (Cat. No. A36579), but the volume is 45 mL. The volume of the beads in the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) is 4.5 mL.
Detection of somatic variants with the Oncomine Myeloid Research Assay GX has been verified down to 5% allele frequency.
Other products to consider
Share catalog number, name or link
1x1 image pixel for data collection